Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 185399760-185400270 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD37
Entrez Gene ID
353322
Gene Name
ankyrin repeat domain 37
Gene Aliases
Lrp2bp
UniGene
Hs.508154
Gene Chromosome Location
Chr.4: 185396686-185400245 on Build GRCh38
Transcripts
NM_181726.3 XM_017008176.1 NM_018359.3 NR_028085.1

Gene Symbol
UFSP2
Entrez Gene ID
55325
Gene Name
UFM1 specific peptidase 2
Gene Aliases
BHD, C4orf20
UniGene
Hs.713548
Gene Chromosome Location
Chr.4: 185399540-185425985 on Build GRCh38
Transcripts
NM_181726.3 XM_017008176.1 NM_018359.3 NR_028085.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1053790 c.467G>T p.R156M Chr.4: 185399764-185399764 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs772438467 Chr.4: 185399788-185399790 on Build GRCh38
rs768905843 Chr.4: 185400001-185400001 on Build GRCh38
rs189059895 Chr.4: 185400007-185400007 on Build GRCh38
rs765518706 Chr.4: 185399977-185399978 on Build GRCh38
rs751602207 Chr.4: 185400015-185400016 on Build GRCh38
rs878875795 Chr.4: 185400142-185400142 on Build GRCh38
rs770157609 Chr.4: 185400017-185400017 on Build GRCh38
rs201799966 Chr.4: 185400024-185400024 on Build GRCh38
rs966370189 Chr.4: 185400099-185400099 on Build GRCh38
rs545340571 Chr.4: 185399927-185399927 on Build GRCh38
rs750267957 Chr.4: 185400063-185400063 on Build GRCh38
rs758328018 Chr.4: 185400115-185400117 on Build GRCh38
rs373722958 Chr.4: 185400080-185400080 on Build GRCh38
rs200207219 Chr.4: 185400005-185400005 on Build GRCh38
rs537169529 Chr.4: 185400103-185400103 on Build GRCh38
rs112860987 Chr.4: 185400113-185400113 on Build GRCh38
rs150877173 Chr.4: 185400062-185400062 on Build GRCh38
rs758542965 Chr.4: 185399987-185399987 on Build GRCh38
rs557089455 Chr.4: 185400115-185400115 on Build GRCh38
rs752309014 Chr.4: 185399817-185399817 on Build GRCh38
rs879231501 Chr.4: 185400242-185400261 on Build GRCh38
rs564896003 Chr.4: 185400186-185400186 on Build GRCh38
rs750447443 Chr.4: 185399978-185399979 on Build GRCh38
rs765877874 Chr.4: 185399806-185399806 on Build GRCh38
rs761957729 Chr.4: 185399973-185399974 on Build GRCh38
rs148425797 Chr.4: 185399958-185399958 on Build GRCh38
rs979223392 Chr.4: 185400181-185400181 on Build GRCh38
rs370459713 Chr.4: 185399807-185399807 on Build GRCh38
rs776107957 Chr.4: 185399805-185399805 on Build GRCh38
rs573004425 Chr.4: 185399972-185399973 on Build GRCh38
rs551361195 Chr.4: 185400075-185400075 on Build GRCh38
rs201769205 Chr.4: 185400049-185400049 on Build GRCh38
rs773771129 Chr.4: 185400023-185400023 on Build GRCh38
rs976339233 Chr.4: 185399868-185399868 on Build GRCh38
rs536815271 Chr.4: 185400197-185400197 on Build GRCh38
rs199667874 Chr.4: 185400042-185400042 on Build GRCh38
rs764502935 Chr.4: 185399970-185399971 on Build GRCh38
rs527587882 Chr.4: 185400061-185400061 on Build GRCh38
rs905626668 Chr.4: 185399961-185399961 on Build GRCh38
rs557620263 Chr.4: 185399813-185399813 on Build GRCh38
rs985979556 Chr.4: 185400123-185400123 on Build GRCh38
rs966927280 Chr.4: 185399879-185399879 on Build GRCh38
rs767302840 Chr.4: 185400039-185400039 on Build GRCh38
rs77278607 Chr.4: 185400128-185400128 on Build GRCh38
rs755958665 Chr.4: 185399826-185399826 on Build GRCh38
rs369003204 Chr.4: 185400077-185400077 on Build GRCh38
rs763286978 Chr.4: 185400003-185400004 on Build GRCh38
rs951755750 Chr.4: 185399801-185399801 on Build GRCh38
rs757608655 Chr.4: 185400206-185400212 on Build GRCh38
rs891713752 Chr.4: 185399894-185399894 on Build GRCh38
rs571550496 Chr.4: 185399876-185399876 on Build GRCh38
rs747229494 Chr.4: 185399991-185399991 on Build GRCh38
rs943874086 Chr.4: 185399845-185399845 on Build GRCh38
rs762550681 Chr.4: 185399785-185399786 on Build GRCh38
rs183872388 Chr.4: 185399978-185399978 on Build GRCh38
rs771405507 Chr.4: 185400033-185400033 on Build GRCh38
rs201136869 Chr.4: 185399816-185399816 on Build GRCh38
rs759245956 Chr.4: 185400038-185400038 on Build GRCh38
rs763404305 Chr.4: 185399791-185399791 on Build GRCh38
rs376863761 Chr.4: 185400036-185400036 on Build GRCh38
rs1013758866 Chr.4: 185400028-185400028 on Build GRCh38
rs747107656 Chr.4: 185399975-185399975 on Build GRCh38
rs753511437 Chr.4: 185399966-185399966 on Build GRCh38
rs755142086 Chr.4: 185400064-185400064 on Build GRCh38
rs997465316 Chr.4: 185400116-185400116 on Build GRCh38
rs1053218 Chr.4: 185399913-185399913 on Build GRCh38
rs573166098 Chr.4: 185399926-185399926 on Build GRCh38
rs781422747 Chr.4: 185400066-185400066 on Build GRCh38
rs748280377 Chr.4: 185400016-185400016 on Build GRCh38
rs917955905 Chr.4: 185400212-185400212 on Build GRCh38
rs937999952 Chr.4: 185399917-185399917 on Build GRCh38
rs553394100 Chr.4: 185400199-185400199 on Build GRCh38
rs373948267 Chr.4: 185399819-185399820 on Build GRCh38
rs1053219 Chr.4: 185399808-185399808 on Build GRCh38
rs777800362 Chr.4: 185399827-185399827 on Build GRCh38
rs371939893 Chr.4: 185399929-185399929 on Build GRCh38
rs763882551 Chr.4: 185400050-185400050 on Build GRCh38
rs990830673 Chr.4: 185400212-185400213 on Build GRCh38

View Full Product Details