Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 185400142-185400658 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD37
Entrez Gene ID
353322
Gene Name
ankyrin repeat domain 37
Gene Aliases
Lrp2bp
UniGene
Hs.508154
Gene Chromosome Location
Chr.4: 185396686-185400245 on Build GRCh38
Transcripts
NM_181726.3 XM_017008176.1 NM_018359.3 NR_028085.1

Gene Symbol
UFSP2
Entrez Gene ID
55325
Gene Name
UFM1 specific peptidase 2
Gene Aliases
BHD, C4orf20
UniGene
Hs.713548
Gene Chromosome Location
Chr.4: 185399540-185425985 on Build GRCh38
Transcripts
NM_181726.3 XM_017008176.1 NM_018359.3 NR_028085.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4412469 c.1324-6A>G p.? Chr.4: 185400484-185400484 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs968505597 Chr.4: 185400617-185400617 on Build GRCh38
rs926975822 Chr.4: 185400337-185400337 on Build GRCh38
rs901547985 Chr.4: 185400359-185400359 on Build GRCh38
rs771618166 Chr.4: 185400368-185400368 on Build GRCh38
rs544351411 Chr.4: 185400429-185400429 on Build GRCh38
rs763657801 Chr.4: 185400401-185400401 on Build GRCh38
rs575214617 Chr.4: 185400308-185400308 on Build GRCh38
rs572741495 Chr.4: 185400532-185400533 on Build GRCh38
rs965630368 Chr.4: 185400250-185400250 on Build GRCh38
rs990928885 Chr.4: 185400479-185400479 on Build GRCh38
rs767664291 Chr.4: 185400432-185400432 on Build GRCh38
rs747983648 Chr.4: 185400501-185400502 on Build GRCh38
rs765178828 Chr.4: 185400415-185400415 on Build GRCh38
rs777769168 Chr.4: 185400358-185400358 on Build GRCh38
rs776156763 Chr.4: 185400524-185400524 on Build GRCh38
rs949398063 Chr.4: 185400247-185400247 on Build GRCh38
rs746178029 Chr.4: 185400486-185400486 on Build GRCh38
rs781063894 Chr.4: 185400490-185400491 on Build GRCh38
rs529079598 Chr.4: 185400542-185400542 on Build GRCh38
rs897427610 Chr.4: 185400409-185400409 on Build GRCh38
rs879231501 Chr.4: 185400242-185400261 on Build GRCh38
rs746270480 Chr.4: 185400371-185400371 on Build GRCh38
rs564896003 Chr.4: 185400186-185400186 on Build GRCh38
rs776022961 Chr.4: 185400387-185400387 on Build GRCh38
rs372836791 Chr.4: 185400439-185400439 on Build GRCh38
rs775222320 Chr.4: 185400386-185400386 on Build GRCh38
rs370248604 Chr.4: 185400385-185400385 on Build GRCh38
rs979223392 Chr.4: 185400181-185400181 on Build GRCh38
rs749351645 Chr.4: 185400478-185400478 on Build GRCh38
rs965400225 Chr.4: 185400546-185400546 on Build GRCh38
rs375238489 Chr.4: 185400521-185400521 on Build GRCh38
rs375981304 Chr.4: 185400470-185400470 on Build GRCh38
rs536815271 Chr.4: 185400197-185400197 on Build GRCh38
rs1038816978 Chr.4: 185400570-185400570 on Build GRCh38
rs576987534 Chr.4: 185400623-185400623 on Build GRCh38
rs901555176 Chr.4: 185400603-185400603 on Build GRCh38
rs3733646 Chr.4: 185400293-185400293 on Build GRCh38
rs757608655 Chr.4: 185400206-185400212 on Build GRCh38
rs931542056 Chr.4: 185400541-185400541 on Build GRCh38
rs942867903 Chr.4: 185400564-185400565 on Build GRCh38
rs895527709 Chr.4: 185400384-185400384 on Build GRCh38
rs756355545 Chr.4: 185400469-185400469 on Build GRCh38
rs1037172741 Chr.4: 185400406-185400406 on Build GRCh38
rs542776845 Chr.4: 185400607-185400607 on Build GRCh38
rs954222321 Chr.4: 185400519-185400519 on Build GRCh38
rs775787335 Chr.4: 185400494-185400494 on Build GRCh38
rs748107400 Chr.4: 185400344-185400344 on Build GRCh38
rs988617768 Chr.4: 185400559-185400559 on Build GRCh38
rs867642986 Chr.4: 185400490-185400490 on Build GRCh38
rs779570982 Chr.4: 185400483-185400483 on Build GRCh38
rs749795266 Chr.4: 185400365-185400365 on Build GRCh38
rs757803330 Chr.4: 185400480-185400480 on Build GRCh38
rs933001637 Chr.4: 185400620-185400620 on Build GRCh38
rs376252704 Chr.4: 185400370-185400370 on Build GRCh38
rs201469027 Chr.4: 185400354-185400354 on Build GRCh38
rs200418126 Chr.4: 185400353-185400353 on Build GRCh38
rs913044033 Chr.4: 185400613-185400613 on Build GRCh38
rs545090918 Chr.4: 185400294-185400294 on Build GRCh38
rs544348416 Chr.4: 185400408-185400408 on Build GRCh38
rs959178532 Chr.4: 185400466-185400466 on Build GRCh38
rs917955905 Chr.4: 185400212-185400212 on Build GRCh38
rs555809622 Chr.4: 185400334-185400335 on Build GRCh38
rs553394100 Chr.4: 185400199-185400199 on Build GRCh38
rs558564595 Chr.4: 185400296-185400296 on Build GRCh38
rs970789449 Chr.4: 185400507-185400507 on Build GRCh38
rs887547619 Chr.4: 185400327-185400327 on Build GRCh38
rs191256886 Chr.4: 185400493-185400493 on Build GRCh38
rs758191031 Chr.4: 185400431-185400431 on Build GRCh38
rs990830673 Chr.4: 185400212-185400213 on Build GRCh38

View Full Product Details