Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 186149036-186149572 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM149A
Entrez Gene ID
25854
Gene Name
family with sequence similarity 149 member A
Gene Aliases
MST119, MSTP119
UniGene
Hs.744961
Gene Chromosome Location
Chr.4: 186104798-186172663 on Build GRCh38
Transcripts
NM_001006655.2 NM_015398.2 XM_005262911.1 XM_005262912.1 XM_005262913.1 XM_011531834.1 XM_011531835.2 XM_011531836.1 XM_017007996.1 XM_017007997.1 XM_017007998.1 XM_017007999.1 XM_017008000.1 XM_017008001.1 XM_017008002.1 XM_017008003.1 XM_017008004.1 XM_017008005.1 XM_017008006.1 XM_017008007.1 XM_017008008.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7207459 c.603T>A p.F201L Chr.4: 186149209-186149209 on Build GRCh38
6389846 c.583G>A p.G195R Chr.4: 186149189-186149189 on Build GRCh38
7302942 c.567-9T>A p.? Chr.4: 186149164-186149164 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs935079066 Chr.4: 186149477-186149477 on Build GRCh38
rs747371456 Chr.4: 186149319-186149319 on Build GRCh38
rs182205568 Chr.4: 186149176-186149176 on Build GRCh38
rs867396030 Chr.4: 186149058-186149058 on Build GRCh38
rs760768417 Chr.4: 186149150-186149150 on Build GRCh38
rs977121640 Chr.4: 186149162-186149162 on Build GRCh38
rs774105585 Chr.4: 186149405-186149405 on Build GRCh38
rs749906321 Chr.4: 186149179-186149179 on Build GRCh38
rs750872236 Chr.4: 186149194-186149194 on Build GRCh38
rs751392358 Chr.4: 186149125-186149125 on Build GRCh38
rs966993377 Chr.4: 186149500-186149500 on Build GRCh38
rs963664051 Chr.4: 186149274-186149274 on Build GRCh38
rs1030499541 Chr.4: 186149488-186149488 on Build GRCh38
rs965214262 Chr.4: 186149085-186149085 on Build GRCh38
rs887741876 Chr.4: 186149315-186149315 on Build GRCh38
rs1051474894 Chr.4: 186149094-186149095 on Build GRCh38
rs764344643 Chr.4: 186149158-186149158 on Build GRCh38
rs112433673 Chr.4: 186149110-186149110 on Build GRCh38
rs761174241 Chr.4: 186149086-186149086 on Build GRCh38
rs201381509 Chr.4: 186149310-186149311 on Build GRCh38
rs560568246 Chr.4: 186149071-186149071 on Build GRCh38
rs755593709 Chr.4: 186149328-186149328 on Build GRCh38
rs550313434 Chr.4: 186149269-186149269 on Build GRCh38
rs1006679069 Chr.4: 186149072-186149072 on Build GRCh38
rs1017780561 Chr.4: 186149078-186149078 on Build GRCh38
rs1035045578 Chr.4: 186149239-186149239 on Build GRCh38
rs754054209 Chr.4: 186149165-186149165 on Build GRCh38
rs544451281 Chr.4: 186149441-186149441 on Build GRCh38
rs929216699 Chr.4: 186149262-186149262 on Build GRCh38
rs1052572280 Chr.4: 186149491-186149491 on Build GRCh38
rs780887121 Chr.4: 186149333-186149333 on Build GRCh38
rs778969598 Chr.4: 186149177-186149177 on Build GRCh38
rs376575506 Chr.4: 186149522-186149522 on Build GRCh38
rs934966027 Chr.4: 186149119-186149119 on Build GRCh38
rs1003587106 Chr.4: 186149238-186149238 on Build GRCh38
rs946967226 Chr.4: 186149520-186149520 on Build GRCh38
rs978298455 Chr.4: 186149502-186149502 on Build GRCh38
rs900568410 Chr.4: 186149424-186149424 on Build GRCh38
rs1047726692 Chr.4: 186149280-186149280 on Build GRCh38
rs948531637 Chr.4: 186149223-186149223 on Build GRCh38
rs564827819 Chr.4: 186149190-186149190 on Build GRCh38
rs567042943 Chr.4: 186149347-186149347 on Build GRCh38
rs530626560 Chr.4: 186149193-186149193 on Build GRCh38
rs191157325 Chr.4: 186149105-186149105 on Build GRCh38
rs780245059 Chr.4: 186149299-186149299 on Build GRCh38
rs1011157311 Chr.4: 186149182-186149182 on Build GRCh38
rs768953893 Chr.4: 186149327-186149327 on Build GRCh38
rs191849868 Chr.4: 186149391-186149391 on Build GRCh38
rs1045522022 Chr.4: 186149235-186149235 on Build GRCh38
rs907157162 Chr.4: 186149254-186149254 on Build GRCh38
rs761477835 Chr.4: 186149146-186149146 on Build GRCh38
rs186857942 Chr.4: 186149203-186149203 on Build GRCh38
rs555913277 Chr.4: 186149361-186149361 on Build GRCh38

View Full Product Details