Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 186155912-186156394 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM149A
Entrez Gene ID
25854
Gene Name
family with sequence similarity 149 member A
Gene Aliases
MST119, MSTP119
UniGene
Hs.744961
Gene Chromosome Location
Chr.4: 186104798-186172663 on Build GRCh38
Transcripts
NM_001006655.2 NM_015398.2 XM_005262911.1 XM_005262912.1 XM_005262913.1 XM_011531834.1 XM_011531835.2 XM_011531836.1 XM_017007996.1 XM_017007997.1 XM_017007998.1 XM_017007999.1 XM_017008000.1 XM_017008001.1 XM_017008002.1 XM_017008003.1 XM_017008004.1 XM_017008005.1 XM_017008006.1 XM_017008007.1 XM_017008008.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3760529 c.436A>G p.K146E Chr.4: 186156052-186156052 on Build GRCh38
6658626 c.1306C>T p.R436C Chr.4: 186156049-186156049 on Build GRCh38
6099784 c.462G>T p.P154P Chr.4: 186156078-186156078 on Build GRCh38
6099786 c.443G>T p.G148V Chr.4: 186156059-186156059 on Build GRCh38
6099785 c.1335G>T p.P445P Chr.4: 186156078-186156078 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368121879 Chr.4: 186156191-186156191 on Build GRCh38
rs368819152 Chr.4: 186156201-186156201 on Build GRCh38
rs1038413407 Chr.4: 186156329-186156329 on Build GRCh38
rs146614253 Chr.4: 186156099-186156099 on Build GRCh38
rs905599413 Chr.4: 186156312-186156312 on Build GRCh38
rs754283932 Chr.4: 186155995-186155995 on Build GRCh38
rs753950218 Chr.4: 186156055-186156055 on Build GRCh38
rs140463493 Chr.4: 186156050-186156050 on Build GRCh38
rs751842987 Chr.4: 186156172-186156172 on Build GRCh38
rs768119890 Chr.4: 186156100-186156100 on Build GRCh38
rs755439404 Chr.4: 186156177-186156177 on Build GRCh38
rs950689691 Chr.4: 186156058-186156058 on Build GRCh38
rs138184420 Chr.4: 186156037-186156037 on Build GRCh38
rs767753016 Chr.4: 186155967-186155967 on Build GRCh38
rs574207344 Chr.4: 186156193-186156193 on Build GRCh38
rs757377346 Chr.4: 186156059-186156059 on Build GRCh38
rs754712900 Chr.4: 186156079-186156079 on Build GRCh38
rs765720647 Chr.4: 186156064-186156064 on Build GRCh38
rs202147474 Chr.4: 186156023-186156023 on Build GRCh38
rs201494456 Chr.4: 186156124-186156124 on Build GRCh38
rs1016291631 Chr.4: 186156369-186156369 on Build GRCh38
rs748063006 Chr.4: 186156039-186156039 on Build GRCh38
rs17884524 Chr.4: 186156024-186156024 on Build GRCh38
rs17881021 Chr.4: 186156015-186156015 on Build GRCh38
rs375098030 Chr.4: 186156034-186156034 on Build GRCh38
rs758748075 Chr.4: 186156029-186156029 on Build GRCh38
rs148413995 Chr.4: 186156327-186156327 on Build GRCh38
rs144604546 Chr.4: 186156102-186156102 on Build GRCh38
rs62350489 Chr.4: 186156354-186156354 on Build GRCh38
rs1011246932 Chr.4: 186155957-186155957 on Build GRCh38
rs745650578 Chr.4: 186156235-186156235 on Build GRCh38
rs762037425 Chr.4: 186156241-186156241 on Build GRCh38
rs200916497 Chr.4: 186156184-186156184 on Build GRCh38
rs980999587 Chr.4: 186156011-186156011 on Build GRCh38
rs114480943 Chr.4: 186156051-186156051 on Build GRCh38
rs766730836 Chr.4: 186156167-186156167 on Build GRCh38
rs773881614 Chr.4: 186156233-186156233 on Build GRCh38
rs779207239 Chr.4: 186156010-186156010 on Build GRCh38
rs17881380 Chr.4: 186156000-186156000 on Build GRCh38
rs769481879 Chr.4: 186156094-186156094 on Build GRCh38
rs572376870 Chr.4: 186156242-186156242 on Build GRCh38
rs1002746731 Chr.4: 186156320-186156320 on Build GRCh38
rs62350488 Chr.4: 186156272-186156272 on Build GRCh38
rs556222486 Chr.4: 186156113-186156113 on Build GRCh38
rs139661959 Chr.4: 186156068-186156068 on Build GRCh38
rs761877863 Chr.4: 186156136-186156136 on Build GRCh38
rs377314469 Chr.4: 186156220-186156220 on Build GRCh38
rs756211875 Chr.4: 186155993-186155993 on Build GRCh38
rs192140289 Chr.4: 186156002-186156002 on Build GRCh38
rs752308639 Chr.4: 186156206-186156206 on Build GRCh38
rs372208223 Chr.4: 186156114-186156114 on Build GRCh38
rs370895680 Chr.4: 186156226-186156226 on Build GRCh38
rs764161823 Chr.4: 186155994-186155994 on Build GRCh38
rs7670550 Chr.4: 186156239-186156239 on Build GRCh38
rs61259112 Chr.4: 186156360-186156361 on Build GRCh38
rs147196705 Chr.4: 186156046-186156046 on Build GRCh38
rs554040476 Chr.4: 186156067-186156067 on Build GRCh38
rs746222066 Chr.4: 186156013-186156013 on Build GRCh38
rs376227303 Chr.4: 186156078-186156078 on Build GRCh38
rs533339417 Chr.4: 186156308-186156308 on Build GRCh38
rs764269790 Chr.4: 186156117-186156117 on Build GRCh38
rs747635923 Chr.4: 186156081-186156081 on Build GRCh38
rs149745987 Chr.4: 186156080-186156080 on Build GRCh38
rs750887144 Chr.4: 186156154-186156154 on Build GRCh38
rs780974220 Chr.4: 186156198-186156198 on Build GRCh38
rs17880686 Chr.4: 186155978-186155978 on Build GRCh38
rs752462255 Chr.4: 186156200-186156200 on Build GRCh38
rs4862653 Chr.4: 186156052-186156052 on Build GRCh38
rs13114283 Chr.4: 186156283-186156283 on Build GRCh38
rs769752801 Chr.4: 186155978-186155979 on Build GRCh38
rs892777267 Chr.4: 186156253-186156253 on Build GRCh38
rs764111506 Chr.4: 186156053-186156053 on Build GRCh38
rs752916885 Chr.4: 186155992-186155992 on Build GRCh38
rs776769122 Chr.4: 186156240-186156240 on Build GRCh38
rs141088571 Chr.4: 186156137-186156137 on Build GRCh38
rs746650008 Chr.4: 186156035-186156035 on Build GRCh38
rs762755273 Chr.4: 186156219-186156219 on Build GRCh38
rs745898966 Chr.4: 186156279-186156283 on Build GRCh38
rs17886141 Chr.4: 186156012-186156012 on Build GRCh38
rs368472968 Chr.4: 186156077-186156077 on Build GRCh38
rs563753051 Chr.4: 186156353-186156353 on Build GRCh38
rs768225420 Chr.4: 186156036-186156036 on Build GRCh38
rs777381311 Chr.4: 186156052-186156052 on Build GRCh38
rs397941639 Chr.4: 186156366-186156367 on Build GRCh38
rs763394753 Chr.4: 186156156-186156156 on Build GRCh38
rs144229720 Chr.4: 186156049-186156049 on Build GRCh38

View Full Product Details