Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 186596852-186597357 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAT1
Entrez Gene ID
2195
Gene Name
FAT atypical cadherin 1
Gene Aliases
CDHF7, CDHR8, FAT, ME5, hFat1
UniGene
Hs.481371
Gene Chromosome Location
Chr.4: 186587783-186726915 on Build GRCh38
Transcripts
NM_005245.3 XM_005262834.2 XM_005262835.2 XM_006714139.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4584981 c.12641C>A p.A4214D Chr.4: 186596899-186596899 on Build GRCh38
6957349 c.12640G>A p.A4214T Chr.4: 186596900-186596900 on Build GRCh38
6913449 c.12462_12463insA p.H4155fs*50 Chr.4: 186597077-186597078 on Build GRCh38
6931703 c.12551G>A p.G4184E Chr.4: 186596989-186596989 on Build GRCh38
5985207 c.12641C>T p.A4214V Chr.4: 186596899-186596899 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1046322764 Chr.4: 186597019-186597019 on Build GRCh38
rs201891217 Chr.4: 186597189-186597189 on Build GRCh38
rs947097391 Chr.4: 186597162-186597162 on Build GRCh38
rs774694931 Chr.4: 186597013-186597013 on Build GRCh38
rs185345294 Chr.4: 186597224-186597224 on Build GRCh38
rs750605265 Chr.4: 186596980-186596982 on Build GRCh38
rs902227655 Chr.4: 186596950-186596950 on Build GRCh38
rs1026399548 Chr.4: 186597029-186597029 on Build GRCh38
rs752274557 Chr.4: 186597221-186597221 on Build GRCh38
rs964862087 Chr.4: 186597317-186597317 on Build GRCh38
rs200934696 Chr.4: 186597111-186597111 on Build GRCh38
rs200185840 Chr.4: 186596932-186596932 on Build GRCh38
rs559086104 Chr.4: 186596899-186596899 on Build GRCh38
rs768789360 Chr.4: 186597187-186597187 on Build GRCh38
rs539779886 Chr.4: 186596968-186596968 on Build GRCh38
rs996240859 Chr.4: 186597330-186597330 on Build GRCh38
rs201960763 Chr.4: 186596918-186596918 on Build GRCh38
rs781295226 Chr.4: 186597061-186597061 on Build GRCh38
rs1016280976 Chr.4: 186597313-186597313 on Build GRCh38
rs753311624 Chr.4: 186596934-186596934 on Build GRCh38
rs184149651 Chr.4: 186597072-186597072 on Build GRCh38
rs756398778 Chr.4: 186596939-186596939 on Build GRCh38
rs748975832 Chr.4: 186596876-186596876 on Build GRCh38
rs187023550 Chr.4: 186597139-186597139 on Build GRCh38
rs777396007 Chr.4: 186596871-186596871 on Build GRCh38
rs762260849 Chr.4: 186597192-186597192 on Build GRCh38
rs760729090 Chr.4: 186597186-186597186 on Build GRCh38
rs760747643 Chr.4: 186597102-186597102 on Build GRCh38
rs776531396 Chr.4: 186597009-186597009 on Build GRCh38
rs368427985 Chr.4: 186597220-186597220 on Build GRCh38
rs745413276 Chr.4: 186596885-186596885 on Build GRCh38
rs777571361 Chr.4: 186597157-186597157 on Build GRCh38
rs745311419 Chr.4: 186596981-186596981 on Build GRCh38
rs768737467 Chr.4: 186597006-186597006 on Build GRCh38
rs773802069 Chr.4: 186596883-186596883 on Build GRCh38
rs779569229 Chr.4: 186597045-186597045 on Build GRCh38
rs1016376045 Chr.4: 186597085-186597085 on Build GRCh38
rs766743632 Chr.4: 186596902-186596902 on Build GRCh38
rs764617344 Chr.4: 186597026-186597026 on Build GRCh38
rs765693916 Chr.4: 186597195-186597195 on Build GRCh38
rs754510259 Chr.4: 186597215-186597215 on Build GRCh38
rs754596378 Chr.4: 186597153-186597153 on Build GRCh38
rs369894626 Chr.4: 186597022-186597022 on Build GRCh38
rs35694170 Chr.4: 186596973-186596973 on Build GRCh38
rs761708072 Chr.4: 186597010-186597010 on Build GRCh38
rs1044056564 Chr.4: 186597165-186597165 on Build GRCh38
rs72716245 Chr.4: 186597018-186597018 on Build GRCh38
rs922925202 Chr.4: 186596905-186596905 on Build GRCh38
rs748132913 Chr.4: 186597156-186597156 on Build GRCh38
rs757631042 Chr.4: 186597037-186597037 on Build GRCh38
rs775508916 Chr.4: 186596888-186596888 on Build GRCh38
rs200322175 Chr.4: 186596984-186596984 on Build GRCh38
rs369440829 Chr.4: 186597000-186597000 on Build GRCh38
rs772269631 Chr.4: 186597179-186597179 on Build GRCh38
rs374263517 Chr.4: 186597214-186597214 on Build GRCh38
rs375994752 Chr.4: 186597119-186597119 on Build GRCh38
rs372348852 Chr.4: 186596901-186596901 on Build GRCh38
rs749064405 Chr.4: 186597164-186597164 on Build GRCh38
rs191917852 Chr.4: 186597155-186597155 on Build GRCh38
rs767899323 Chr.4: 186596933-186596933 on Build GRCh38
rs566899991 Chr.4: 186597281-186597281 on Build GRCh38
rs750733656 Chr.4: 186597199-186597199 on Build GRCh38
rs772201790 Chr.4: 186597093-186597093 on Build GRCh38
rs770402320 Chr.4: 186596877-186596877 on Build GRCh38
rs763597855 Chr.4: 186597105-186597105 on Build GRCh38
rs778521969 Chr.4: 186597041-186597041 on Build GRCh38
rs779582047 Chr.4: 186597125-186597125 on Build GRCh38
rs746533501 Chr.4: 186597051-186597051 on Build GRCh38
rs377151791 Chr.4: 186597021-186597021 on Build GRCh38
rs775588951 Chr.4: 186597180-186597180 on Build GRCh38
rs1006246644 Chr.4: 186597297-186597297 on Build GRCh38
rs899252716 Chr.4: 186597329-186597329 on Build GRCh38
rs765684772 Chr.4: 186597106-186597106 on Build GRCh38
rs751626816 Chr.4: 186596917-186596917 on Build GRCh38
rs60404529 Chr.4: 186597042-186597042 on Build GRCh38
rs754134795 Chr.4: 186597032-186597032 on Build GRCh38
rs935510544 Chr.4: 186597152-186597152 on Build GRCh38
rs190301534 Chr.4: 186597225-186597225 on Build GRCh38
rs367784038 Chr.4: 186596966-186596966 on Build GRCh38
rs780327089 Chr.4: 186597271-186597271 on Build GRCh38
rs544799226 Chr.4: 186596970-186596970 on Build GRCh38
rs775598947 Chr.4: 186597095-186597095 on Build GRCh38
rs768651990 Chr.4: 186597054-186597054 on Build GRCh38
rs376106855 Chr.4: 186597043-186597043 on Build GRCh38
rs1035943462 Chr.4: 186596990-186596990 on Build GRCh38
rs754398558 Chr.4: 186597104-186597104 on Build GRCh38
rs72716244 Chr.4: 186596887-186596887 on Build GRCh38
rs199692977 Chr.4: 186597074-186597074 on Build GRCh38
rs370606960 Chr.4: 186597208-186597208 on Build GRCh38
rs749675349 Chr.4: 186596967-186596967 on Build GRCh38
rs368645432 Chr.4: 186596900-186596900 on Build GRCh38
rs747946682 Chr.4: 186597062-186597062 on Build GRCh38
rs75737957 Chr.4: 186597276-186597276 on Build GRCh38
rs747017523 Chr.4: 186597001-186597001 on Build GRCh38

View Full Product Details