Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 186597601-186598101 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAT1
Entrez Gene ID
2195
Gene Name
FAT atypical cadherin 1
Gene Aliases
CDHF7, CDHR8, FAT, ME5, hFat1
UniGene
Hs.481371
Gene Chromosome Location
Chr.4: 186587783-186726915 on Build GRCh38
Transcripts
NM_005245.3 XM_005262834.2 XM_005262835.2 XM_006714139.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4123861 c.12245A>G p.Y4082C Chr.4: 186597984-186597984 on Build GRCh38
6099753 c.12215G>A p.G4072E Chr.4: 186598014-186598014 on Build GRCh38
6922210 c.12253C>G p.Q4085E Chr.4: 186597976-186597976 on Build GRCh38
6976486 c.12164C>A p.P4055Q Chr.4: 186598065-186598065 on Build GRCh38
6099754 c.12215G>T p.G4072V Chr.4: 186598014-186598014 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs545481607 Chr.4: 186598053-186598053 on Build GRCh38
rs768136150 Chr.4: 186598041-186598041 on Build GRCh38
rs979859223 Chr.4: 186597729-186597729 on Build GRCh38
rs547687281 Chr.4: 186597830-186597830 on Build GRCh38
rs779847763 Chr.4: 186597889-186597889 on Build GRCh38
rs371227366 Chr.4: 186597984-186597984 on Build GRCh38
rs376771050 Chr.4: 186597777-186597777 on Build GRCh38
rs2306990 Chr.4: 186597822-186597822 on Build GRCh38
rs201796025 Chr.4: 186597657-186597657 on Build GRCh38
rs960026866 Chr.4: 186597761-186597761 on Build GRCh38
rs569242636 Chr.4: 186597906-186597906 on Build GRCh38
rs371249526 Chr.4: 186597948-186597948 on Build GRCh38
rs751396320 Chr.4: 186598015-186598015 on Build GRCh38
rs760226790 Chr.4: 186597815-186597815 on Build GRCh38
rs372923940 Chr.4: 186597841-186597841 on Build GRCh38
rs748249672 Chr.4: 186597767-186597767 on Build GRCh38
rs1035560651 Chr.4: 186598011-186598011 on Build GRCh38
rs757467460 Chr.4: 186598022-186598022 on Build GRCh38
rs77814178 Chr.4: 186597962-186597962 on Build GRCh38
rs767305041 Chr.4: 186597716-186597716 on Build GRCh38
rs778058808 Chr.4: 186598017-186598017 on Build GRCh38
rs761400229 Chr.4: 186597990-186597990 on Build GRCh38
rs377052861 Chr.4: 186597724-186597724 on Build GRCh38
rs750552688 Chr.4: 186598073-186598073 on Build GRCh38
rs375588095 Chr.4: 186597656-186597656 on Build GRCh38
rs573837982 Chr.4: 186597652-186597652 on Build GRCh38
rs772540738 Chr.4: 186598031-186598031 on Build GRCh38
rs767217120 Chr.4: 186597802-186597802 on Build GRCh38
rs961687769 Chr.4: 186598020-186598020 on Build GRCh38
rs561788984 Chr.4: 186598076-186598076 on Build GRCh38
rs371819771 Chr.4: 186597696-186597696 on Build GRCh38
rs780055518 Chr.4: 186597932-186597934 on Build GRCh38
rs752904545 Chr.4: 186597836-186597836 on Build GRCh38
rs771493652 Chr.4: 186597932-186597932 on Build GRCh38
rs745695748 Chr.4: 186597746-186597746 on Build GRCh38
rs201130322 Chr.4: 186598044-186598044 on Build GRCh38
rs749054777 Chr.4: 186597649-186597649 on Build GRCh38
rs759980028 Chr.4: 186597798-186597798 on Build GRCh38
rs376155526 Chr.4: 186597936-186597936 on Build GRCh38
rs143744017 Chr.4: 186598043-186598043 on Build GRCh38
rs200611830 Chr.4: 186598072-186598072 on Build GRCh38
rs1280097 Chr.4: 186598052-186598052 on Build GRCh38
rs747406113 Chr.4: 186598037-186598037 on Build GRCh38
rs376617534 Chr.4: 186597638-186597638 on Build GRCh38
rs775938826 Chr.4: 186597975-186597975 on Build GRCh38
rs890546842 Chr.4: 186597855-186597855 on Build GRCh38
rs180899930 Chr.4: 186597926-186597926 on Build GRCh38
rs757044814 Chr.4: 186597732-186597732 on Build GRCh38
rs773654248 Chr.4: 186597688-186597688 on Build GRCh38
rs753411930 Chr.4: 186597819-186597819 on Build GRCh38
rs890632292 Chr.4: 186597919-186597919 on Build GRCh38
rs773774314 Chr.4: 186597653-186597653 on Build GRCh38
rs1037664539 Chr.4: 186597847-186597847 on Build GRCh38
rs774367320 Chr.4: 186598065-186598065 on Build GRCh38
rs1030902519 Chr.4: 186597640-186597640 on Build GRCh38
rs2306989 Chr.4: 186597717-186597717 on Build GRCh38
rs765147751 Chr.4: 186597991-186597991 on Build GRCh38
rs746415591 Chr.4: 186598025-186598025 on Build GRCh38
rs763373898 Chr.4: 186597690-186597690 on Build GRCh38
rs758627924 Chr.4: 186597828-186597828 on Build GRCh38
rs538368483 Chr.4: 186597909-186597909 on Build GRCh38
rs755127492 Chr.4: 186597747-186597747 on Build GRCh38
rs748231872 Chr.4: 186597681-186597681 on Build GRCh38
rs969118045 Chr.4: 186597741-186597741 on Build GRCh38
rs576041146 Chr.4: 186598068-186598068 on Build GRCh38
rs369681913 Chr.4: 186597736-186597736 on Build GRCh38
rs779558478 Chr.4: 186598024-186598024 on Build GRCh38
rs755592521 Chr.4: 186597719-186597719 on Build GRCh38
rs371989627 Chr.4: 186598016-186598016 on Build GRCh38
rs367812758 Chr.4: 186598014-186598014 on Build GRCh38
rs115420826 Chr.4: 186597905-186597905 on Build GRCh38
rs780242823 Chr.4: 186598032-186598032 on Build GRCh38
rs1280096 Chr.4: 186597826-186597826 on Build GRCh38
rs749811366 Chr.4: 186597785-186597785 on Build GRCh38
rs188472180 Chr.4: 186597833-186597833 on Build GRCh38
rs1009373100 Chr.4: 186597858-186597858 on Build GRCh38
rs769835915 Chr.4: 186597772-186597772 on Build GRCh38
rs371027115 Chr.4: 186597637-186597637 on Build GRCh38
rs116628547 Chr.4: 186597720-186597720 on Build GRCh38
rs776652560 Chr.4: 186597664-186597664 on Build GRCh38
rs759213300 Chr.4: 186597715-186597715 on Build GRCh38
rs999664921 Chr.4: 186597923-186597923 on Build GRCh38
rs762709565 Chr.4: 186598012-186598012 on Build GRCh38
rs760293297 Chr.4: 186597964-186597964 on Build GRCh38
rs200661419 Chr.4: 186597968-186597968 on Build GRCh38
rs138084499 Chr.4: 186598021-186598021 on Build GRCh38
rs758250575 Chr.4: 186597831-186597831 on Build GRCh38
rs763551872 Chr.4: 186597817-186597817 on Build GRCh38
rs758495731 Chr.4: 186597856-186597857 on Build GRCh38
rs374812339 Chr.4: 186597992-186597992 on Build GRCh38
rs538167941 Chr.4: 186597699-186597699 on Build GRCh38
rs150584542 Chr.4: 186598064-186598064 on Build GRCh38
rs375107697 Chr.4: 186597705-186597705 on Build GRCh38
rs1052457943 Chr.4: 186597888-186597888 on Build GRCh38
rs773113119 Chr.4: 186598047-186598047 on Build GRCh38
rs940602352 Chr.4: 186597786-186597786 on Build GRCh38
rs770281172 Chr.4: 186597683-186597683 on Build GRCh38
rs774814395 Chr.4: 186597792-186597792 on Build GRCh38
rs749414274 Chr.4: 186597929-186597929 on Build GRCh38
rs202166446 Chr.4: 186597762-186597762 on Build GRCh38
rs769337342 Chr.4: 186598045-186598045 on Build GRCh38
rs775340406 Chr.4: 186597810-186597810 on Build GRCh38
rs757379580 Chr.4: 186598071-186598071 on Build GRCh38

View Full Product Details