Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 189961365-189961910 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRG1
Entrez Gene ID
2483
Gene Name
FSHD region gene 1
Gene Aliases
FRG1A, FSG1
UniGene
Hs.203772
Gene Chromosome Location
Chr.4: 189940819-189963204 on Build GRCh38
Transcripts
NM_004477.2 XM_017007958.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6100224 c.644G>A p.S215N Chr.4: 189961836-189961836 on Build GRCh38
1131140 c.683G>T p.S228I Chr.4: 189961875-189961875 on Build GRCh38
3132181 c.630-6T>C p.? Chr.4: 189961816-189961816 on Build GRCh38
4597442 c.692T>C p.L231P Chr.4: 189961884-189961884 on Build GRCh38
1716000 c.704G>A p.R235Q Chr.4: 189961896-189961896 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs75590332 Chr.4: 189961843-189961843 on Build GRCh38
rs948616130 Chr.4: 189961826-189961826 on Build GRCh38
rs948563749 Chr.4: 189961533-189961533 on Build GRCh38
rs779579309 Chr.4: 189961724-189961725 on Build GRCh38
rs977230742 Chr.4: 189961714-189961714 on Build GRCh38
rs142297472 Chr.4: 189961564-189961564 on Build GRCh38
rs541610551 Chr.4: 189961683-189961683 on Build GRCh38
rs1031834491 Chr.4: 189961717-189961717 on Build GRCh38
rs575016515 Chr.4: 189961782-189961782 on Build GRCh38
rs536675534 Chr.4: 189961553-189961553 on Build GRCh38
rs769294402 Chr.4: 189961464-189961464 on Build GRCh38
rs796396919 Chr.4: 189961532-189961532 on Build GRCh38
rs915842096 Chr.4: 189961801-189961801 on Build GRCh38
rs878969536 Chr.4: 189961671-189961672 on Build GRCh38
rs560717739 Chr.4: 189961840-189961840 on Build GRCh38
rs1005831888 Chr.4: 189961409-189961410 on Build GRCh38
rs755380808 Chr.4: 189961816-189961816 on Build GRCh38
rs78046499 Chr.4: 189961754-189961754 on Build GRCh38
rs142128033 Chr.4: 189961409-189961410 on Build GRCh38
rs879110637 Chr.4: 189961691-189961691 on Build GRCh38
rs577530518 Chr.4: 189961676-189961676 on Build GRCh38
rs112946446 Chr.4: 189961875-189961875 on Build GRCh38
rs955711656 Chr.4: 189961699-189961699 on Build GRCh38
rs565478494 Chr.4: 189961411-189961411 on Build GRCh38
rs374630014 Chr.4: 189961867-189961867 on Build GRCh38
rs201978002 Chr.4: 189961852-189961852 on Build GRCh38
rs201210198 Chr.4: 189961726-189961726 on Build GRCh38
rs78181874 Chr.4: 189961818-189961818 on Build GRCh38
rs755496611 Chr.4: 189961793-189961793 on Build GRCh38
rs370890306 Chr.4: 189961563-189961563 on Build GRCh38
rs1029428875 Chr.4: 189961446-189961446 on Build GRCh38
rs879170972 Chr.4: 189961873-189961873 on Build GRCh38
rs201058826 Chr.4: 189961773-189961773 on Build GRCh38
rs796645389 Chr.4: 189961494-189961494 on Build GRCh38
rs184246550 Chr.4: 189961558-189961558 on Build GRCh38
rs112995708 Chr.4: 189961649-189961649 on Build GRCh38
rs544072877 Chr.4: 189961410-189961410 on Build GRCh38
rs375714692 Chr.4: 189961762-189961762 on Build GRCh38
rs78987873 Chr.4: 189961539-189961539 on Build GRCh38
rs757055455 Chr.4: 189961740-189961740 on Build GRCh38
rs773602053 Chr.4: 189961823-189961825 on Build GRCh38
rs577468777 Chr.4: 189961418-189961418 on Build GRCh38
rs531160609 Chr.4: 189961868-189961868 on Build GRCh38
rs73024974 Chr.4: 189961390-189961390 on Build GRCh38
rs753099278 Chr.4: 189961838-189961838 on Build GRCh38
rs376110574 Chr.4: 189961781-189961782 on Build GRCh38
rs564292001 Chr.4: 189961493-189961493 on Build GRCh38
rs772043026 Chr.4: 189961880-189961880 on Build GRCh38
rs781611568 Chr.4: 189961410-189961410 on Build GRCh38
rs1017200785 Chr.4: 189961595-189961595 on Build GRCh38
rs763097514 Chr.4: 189961804-189961804 on Build GRCh38
rs145922262 Chr.4: 189961409-189961409 on Build GRCh38
rs78402326 Chr.4: 189961439-189961439 on Build GRCh38
rs542648111 Chr.4: 189961807-189961807 on Build GRCh38
rs113609235 Chr.4: 189961770-189961770 on Build GRCh38
rs570180997 Chr.4: 189961607-189961607 on Build GRCh38
rs547683952 Chr.4: 189961537-189961537 on Build GRCh38
rs1045513472 Chr.4: 189961534-189961534 on Build GRCh38
rs750583178 Chr.4: 189961785-189961785 on Build GRCh38
rs117300383 Chr.4: 189961430-189961430 on Build GRCh38
rs373222273 Chr.4: 189961756-189961756 on Build GRCh38
rs887150878 Chr.4: 189961567-189961567 on Build GRCh38
rs373191894 Chr.4: 189961565-189961565 on Build GRCh38
rs796202483 Chr.4: 189961488-189961488 on Build GRCh38
rs549610185 Chr.4: 189961481-189961481 on Build GRCh38
rs796305272 Chr.4: 189961491-189961491 on Build GRCh38
rs113116556 Chr.4: 189961794-189961794 on Build GRCh38
rs144407668 Chr.4: 189961555-189961555 on Build GRCh38
rs527304793 Chr.4: 189961470-189961470 on Build GRCh38
rs376452352 Chr.4: 189961502-189961502 on Build GRCh38
rs543121899 Chr.4: 189961872-189961872 on Build GRCh38
rs537547783 Chr.4: 189961627-189961627 on Build GRCh38
rs79806655 Chr.4: 189961496-189961496 on Build GRCh38
rs561072040 Chr.4: 189961476-189961476 on Build GRCh38
rs878866930 Chr.4: 189961687-189961687 on Build GRCh38
rs776918341 Chr.4: 189961772-189961772 on Build GRCh38
rs541756244 Chr.4: 189961420-189961420 on Build GRCh38
rs199588911 Chr.4: 189961871-189961871 on Build GRCh38
rs951439299 Chr.4: 189961405-189961405 on Build GRCh38
rs879034752 Chr.4: 189961701-189961701 on Build GRCh38
rs900053001 Chr.4: 189961646-189961646 on Build GRCh38
rs35503121 Chr.4: 189961418-189961418 on Build GRCh38
rs997032606 Chr.4: 189961648-189961648 on Build GRCh38
rs76995424 Chr.4: 189961444-189961444 on Build GRCh38
rs532133652 Chr.4: 189961531-189961531 on Build GRCh38
rs377313353 Chr.4: 189961572-189961572 on Build GRCh38
rs111325833 Chr.4: 189961645-189961645 on Build GRCh38
rs879188786 Chr.4: 189961688-189961688 on Build GRCh38
rs191654402 Chr.4: 189961471-189961471 on Build GRCh38
rs559054039 Chr.4: 189961665-189961665 on Build GRCh38
rs112377574 Chr.4: 189961641-189961641 on Build GRCh38
rs187536295 Chr.4: 189961391-189961391 on Build GRCh38
rs531296272 Chr.4: 189961477-189961477 on Build GRCh38
rs377281594 Chr.4: 189961700-189961700 on Build GRCh38
rs112880159 Chr.4: 189961846-189961846 on Build GRCh38
rs751957929 Chr.4: 189961809-189961809 on Build GRCh38
rs113447994 Chr.4: 189961596-189961596 on Build GRCh38
rs977511710 Chr.4: 189961450-189961450 on Build GRCh38
rs553657626 Chr.4: 189961706-189961706 on Build GRCh38
rs373100464 Chr.4: 189961684-189961684 on Build GRCh38
rs957060823 Chr.4: 189961724-189961724 on Build GRCh38
rs770099816 Chr.4: 189961780-189961781 on Build GRCh38
rs796980548 Chr.4: 189961556-189961556 on Build GRCh38
rs370592074 Chr.4: 189961588-189961588 on Build GRCh38
rs565806094 Chr.4: 189961545-189961545 on Build GRCh38
rs200007941 Chr.4: 189961884-189961884 on Build GRCh38
rs114577831 Chr.4: 189961456-189961456 on Build GRCh38
rs867397318 Chr.4: 189961886-189961886 on Build GRCh38

View Full Product Details