Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 311188-311676 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AHRR
Entrez Gene ID
57491
Gene Name
aryl-hydrocarbon receptor repressor
Gene Aliases
AHH, AHHR, bHLHe77
UniGene
Hs.50823
Gene Chromosome Location
Chr.5: 304176-438291 on Build GRCh38
Transcripts
NM_001242412.1 NM_020731.4 NM_001267556.1 NM_001267557.1 NM_001267558.1 NM_001267559.1 NM_013232.3

Gene Symbol
PDCD6
Entrez Gene ID
10016
Gene Name
programmed cell death 6
Gene Aliases
ALG-2, ALG2, PEF1B
UniGene
Hs.50823
Gene Chromosome Location
Chr.5: 271621-314974 on Build GRCh38
Transcripts
NM_001242412.1 NM_020731.4 NM_001267556.1 NM_001267557.1 NM_001267558.1 NM_001267559.1 NM_013232.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6742281 c.469G>A p.V157I Chr.5: 311394-311394 on Build GRCh38
449516 c.438T>C p.I146I Chr.5: 311363-311363 on Build GRCh38
7172352 c.382G>C p.D128H Chr.5: 311307-311307 on Build GRCh38
4548959 c.460G>A p.G154S Chr.5: 311385-311385 on Build GRCh38
6470260 c.472C>T p.L158L Chr.5: 311397-311397 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs956580463 Chr.5: 311211-311211 on Build GRCh38
rs145654625 Chr.5: 311337-311337 on Build GRCh38
rs878926845 Chr.5: 311500-311500 on Build GRCh38
rs757147897 Chr.5: 311624-311624 on Build GRCh38
rs371761537 Chr.5: 311421-311421 on Build GRCh38
rs560642997 Chr.5: 311470-311470 on Build GRCh38
rs192286129 Chr.5: 311250-311250 on Build GRCh38
rs779365929 Chr.5: 311369-311369 on Build GRCh38
rs775859679 Chr.5: 311431-311431 on Build GRCh38
rs376713344 Chr.5: 311245-311245 on Build GRCh38
rs535589799 Chr.5: 311560-311560 on Build GRCh38
rs757656555 Chr.5: 311365-311365 on Build GRCh38
rs371288200 Chr.5: 311394-311394 on Build GRCh38
rs996549656 Chr.5: 311364-311364 on Build GRCh38
rs775930052 Chr.5: 311243-311244 on Build GRCh38
rs373194951 Chr.5: 311288-311288 on Build GRCh38
rs755197615 Chr.5: 311398-311398 on Build GRCh38
rs557839530 Chr.5: 311249-311249 on Build GRCh38
rs896467274 Chr.5: 311577-311577 on Build GRCh38
rs376242079 Chr.5: 311321-311321 on Build GRCh38
rs200791638 Chr.5: 311408-311408 on Build GRCh38
rs899386742 Chr.5: 311639-311639 on Build GRCh38
rs768155115 Chr.5: 311393-311393 on Build GRCh38
rs1029082968 Chr.5: 311391-311391 on Build GRCh38
rs112251704 Chr.5: 311438-311439 on Build GRCh38
rs79431533 Chr.5: 311482-311482 on Build GRCh38
rs375867873 Chr.5: 311407-311407 on Build GRCh38
rs187459406 Chr.5: 311557-311557 on Build GRCh38
rs767684079 Chr.5: 311352-311352 on Build GRCh38
rs756330213 Chr.5: 311357-311357 on Build GRCh38
rs753821970 Chr.5: 311445-311445 on Build GRCh38
rs751807620 Chr.5: 311242-311242 on Build GRCh38
rs764291004 Chr.5: 311438-311438 on Build GRCh38
rs550683412 Chr.5: 311619-311619 on Build GRCh38
rs1037927314 Chr.5: 311608-311608 on Build GRCh38
rs940856306 Chr.5: 311587-311587 on Build GRCh38
rs746930161 Chr.5: 311564-311564 on Build GRCh38
rs1022186273 Chr.5: 311494-311494 on Build GRCh38
rs2244029 Chr.5: 311363-311363 on Build GRCh38
rs772846590 Chr.5: 311332-311332 on Build GRCh38
rs201748307 Chr.5: 311318-311318 on Build GRCh38
rs760922697 Chr.5: 311437-311437 on Build GRCh38
rs771582574 Chr.5: 311272-311272 on Build GRCh38
rs779088571 Chr.5: 311255-311255 on Build GRCh38
rs745555751 Chr.5: 311268-311268 on Build GRCh38
rs767614009 Chr.5: 311429-311431 on Build GRCh38
rs562584100 Chr.5: 311397-311397 on Build GRCh38
rs932020533 Chr.5: 311641-311641 on Build GRCh38
rs183545231 Chr.5: 311353-311353 on Build GRCh38
rs368897410 Chr.5: 311370-311370 on Build GRCh38
rs775090904 Chr.5: 311284-311284 on Build GRCh38
rs752867008 Chr.5: 311246-311246 on Build GRCh38
rs538181362 Chr.5: 311602-311602 on Build GRCh38
rs890449668 Chr.5: 311650-311651 on Build GRCh38
rs776613586 Chr.5: 311299-311299 on Build GRCh38
rs1032938242 Chr.5: 311343-311343 on Build GRCh38
rs939124517 Chr.5: 311556-311556 on Build GRCh38
rs6555160 Chr.5: 311554-311554 on Build GRCh38
rs373470365 Chr.5: 311243-311243 on Build GRCh38
rs956508663 Chr.5: 311460-311460 on Build GRCh38
rs969083521 Chr.5: 311552-311552 on Build GRCh38
rs368651363 Chr.5: 311366-311366 on Build GRCh38
rs929994581 Chr.5: 311226-311226 on Build GRCh38
rs752777850 Chr.5: 311455-311456 on Build GRCh38
rs746657841 Chr.5: 311389-311389 on Build GRCh38
rs75013020 Chr.5: 311443-311443 on Build GRCh38
rs540250017 Chr.5: 311251-311251 on Build GRCh38
rs1050829403 Chr.5: 311642-311642 on Build GRCh38
rs6555159 Chr.5: 311447-311447 on Build GRCh38
rs989383782 Chr.5: 311241-311241 on Build GRCh38
rs758155579 Chr.5: 311463-311463 on Build GRCh38
rs927746903 Chr.5: 311555-311555 on Build GRCh38
rs779736494 Chr.5: 311372-311372 on Build GRCh38
rs113680699 Chr.5: 311430-311430 on Build GRCh38
rs34421451 Chr.5: 311253-311253 on Build GRCh38
rs989430992 Chr.5: 311488-311488 on Build GRCh38
rs773182042 Chr.5: 311420-311420 on Build GRCh38

View Full Product Details