Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 443063-443560 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOC3
Entrez Gene ID
11336
Gene Name
exocyst complex component 3
Gene Aliases
SEC6, SEC6L1, Sec6p
UniGene
Hs.481464
Gene Chromosome Location
Chr.5: 443184-471938 on Build GRCh38
Transcripts
NM_007277.4 XM_011513948.2 XM_011513951.2 XM_017009000.1 XM_017009001.1 XM_017009002.1 XM_017009003.1 XM_017009004.1 XM_017009005.1 XM_017009006.1 NR_126522.1

Gene Symbol
EXOC3-AS1
Entrez Gene ID
116349
Gene Name
EXOC3 antisense RNA 1
Gene Aliases
C5orf55
UniGene
Hs.446702
Gene Chromosome Location
Chr.5: 441835-443160 on Build GRCh38
Transcripts
NM_007277.4 XM_011513948.2 XM_011513951.2 XM_017009000.1 XM_017009001.1 XM_017009002.1 XM_017009003.1 XM_017009004.1 XM_017009005.1 XM_017009006.1 NR_126522.1

SNP Detail

SNP ID Chromosome Location
rs916662576 Chr.5: 443115-443115 on Build GRCh38
rs553176185 Chr.5: 443262-443262 on Build GRCh38
rs530239020 Chr.5: 443171-443171 on Build GRCh38
rs956161473 Chr.5: 443478-443478 on Build GRCh38
rs34861347 Chr.5: 443460-443460 on Build GRCh38
rs866602655 Chr.5: 443352-443352 on Build GRCh38
rs1041490109 Chr.5: 443391-443391 on Build GRCh38
rs374353264 Chr.5: 443241-443241 on Build GRCh38
rs543092291 Chr.5: 443428-443428 on Build GRCh38
rs11134040 Chr.5: 443250-443250 on Build GRCh38
rs758457229 Chr.5: 443391-443391 on Build GRCh38
rs958212614 Chr.5: 443099-443099 on Build GRCh38
rs998267332 Chr.5: 443083-443083 on Build GRCh38
rs182219748 Chr.5: 443229-443229 on Build GRCh38
rs925427321 Chr.5: 443098-443098 on Build GRCh38
rs933722007 Chr.5: 443385-443385 on Build GRCh38
rs1039939911 Chr.5: 443349-443349 on Build GRCh38
rs926803497 Chr.5: 443236-443236 on Build GRCh38
rs373080314 Chr.5: 443259-443259 on Build GRCh38
rs565082033 Chr.5: 443091-443091 on Build GRCh38
rs865967090 Chr.5: 443464-443464 on Build GRCh38
rs938132772 Chr.5: 443238-443238 on Build GRCh38
rs958159081 Chr.5: 443244-443244 on Build GRCh38
rs369669320 Chr.5: 443165-443165 on Build GRCh38
rs548664886 Chr.5: 443199-443199 on Build GRCh38
rs1046828206 Chr.5: 443119-443119 on Build GRCh38
rs554396122 Chr.5: 443326-443326 on Build GRCh38
rs11274307 Chr.5: 443258-443278 on Build GRCh38
rs999756730 Chr.5: 443232-443233 on Build GRCh38
rs1020362513 Chr.5: 443223-443223 on Build GRCh38
rs532427489 Chr.5: 443117-443117 on Build GRCh38
rs558906544 Chr.5: 443217-443217 on Build GRCh38
rs922407996 Chr.5: 443513-443513 on Build GRCh38
rs929883849 Chr.5: 443164-443165 on Build GRCh38
rs115554641 Chr.5: 443121-443121 on Build GRCh38
rs564856372 Chr.5: 443450-443450 on Build GRCh38
rs1022373116 Chr.5: 443419-443419 on Build GRCh38
rs368689128 Chr.5: 443253-443253 on Build GRCh38
rs967990472 Chr.5: 443421-443421 on Build GRCh38
rs1049496189 Chr.5: 443538-443538 on Build GRCh38
rs190573874 Chr.5: 443088-443088 on Build GRCh38
rs537076783 Chr.5: 443206-443206 on Build GRCh38
rs11134039 Chr.5: 443247-443247 on Build GRCh38
rs989582035 Chr.5: 443240-443240 on Build GRCh38
rs752151701 Chr.5: 443378-443378 on Build GRCh38
rs918566828 Chr.5: 443299-443299 on Build GRCh38
rs867023807 Chr.5: 443114-443114 on Build GRCh38
rs1006691299 Chr.5: 443188-443188 on Build GRCh38
rs1032609508 Chr.5: 443238-443239 on Build GRCh38
rs978372183 Chr.5: 443095-443095 on Build GRCh38
rs534941835 Chr.5: 443256-443256 on Build GRCh38
rs2672739 Chr.5: 443332-443332 on Build GRCh38
rs1039420868 Chr.5: 443198-443198 on Build GRCh38
rs370708378 Chr.5: 443342-443342 on Build GRCh38
rs990989345 Chr.5: 443250-443251 on Build GRCh38
rs909809073 Chr.5: 443327-443327 on Build GRCh38
rs1030541365 Chr.5: 443086-443086 on Build GRCh38
rs1012536584 Chr.5: 443440-443440 on Build GRCh38
rs1024211885 Chr.5: 443451-443451 on Build GRCh38
rs781596781 Chr.5: 443205-443206 on Build GRCh38
rs542004313 Chr.5: 443271-443271 on Build GRCh38
rs879175367 Chr.5: 443110-443110 on Build GRCh38
rs970945910 Chr.5: 443523-443523 on Build GRCh38
rs867284203 Chr.5: 443226-443226 on Build GRCh38
rs976724648 Chr.5: 443490-443490 on Build GRCh38
rs924506002 Chr.5: 443463-443463 on Build GRCh38
rs932539337 Chr.5: 443536-443536 on Build GRCh38
rs781287119 Chr.5: 443221-443259 on Build GRCh38
rs929995433 Chr.5: 443302-443302 on Build GRCh38
rs894059885 Chr.5: 443433-443433 on Build GRCh38
rs1023847605 Chr.5: 443282-443282 on Build GRCh38
rs532203396 Chr.5: 443107-443107 on Build GRCh38
rs796743116 Chr.5: 443241-443262 on Build GRCh38
rs990876513 Chr.5: 443101-443101 on Build GRCh38
rs971214578 Chr.5: 443283-443283 on Build GRCh38
rs570116571 Chr.5: 443205-443205 on Build GRCh38

View Full Product Details