Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 461868-462373 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOC3
Entrez Gene ID
11336
Gene Name
exocyst complex component 3
Gene Aliases
SEC6, SEC6L1, Sec6p
UniGene
Hs.481464
Gene Chromosome Location
Chr.5: 443184-471938 on Build GRCh38
Transcripts
NM_007277.4 XM_011513948.2 XM_011513949.2 XM_011513950.2 XM_011513951.2 XM_017009000.1 XM_017009001.1 XM_017009002.1 XM_017009003.1 XM_017009004.1 XM_017009005.1 XM_017009006.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6011726 c.1543G>C p.E515Q Chr.5: 462197-462197 on Build GRCh38
1068518 c.1605G>A p.A535A Chr.5: 462259-462259 on Build GRCh38
7124373 c.1564C>G p.P522A Chr.5: 462218-462218 on Build GRCh38
6011727 c.1609G>C p.E537Q Chr.5: 462263-462263 on Build GRCh38
5142755 c.1597G>A p.A533T Chr.5: 462251-462251 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781688902 Chr.5: 461932-461932 on Build GRCh38
rs962674255 Chr.5: 462014-462014 on Build GRCh38
rs766737756 Chr.5: 462100-462100 on Build GRCh38
rs759033187 Chr.5: 461974-461974 on Build GRCh38
rs767463397 Chr.5: 462042-462042 on Build GRCh38
rs764035656 Chr.5: 461993-461993 on Build GRCh38
rs563248749 Chr.5: 462333-462333 on Build GRCh38
rs754663864 Chr.5: 462311-462311 on Build GRCh38
rs748155022 Chr.5: 462248-462248 on Build GRCh38
rs943364609 Chr.5: 462032-462032 on Build GRCh38
rs768319821 Chr.5: 461948-461948 on Build GRCh38
rs201209420 Chr.5: 462238-462238 on Build GRCh38
rs370123001 Chr.5: 462330-462330 on Build GRCh38
rs749591372 Chr.5: 462065-462065 on Build GRCh38
rs371466267 Chr.5: 462344-462344 on Build GRCh38
rs142927762 Chr.5: 462250-462250 on Build GRCh38
rs774053027 Chr.5: 462219-462219 on Build GRCh38
rs771794323 Chr.5: 462256-462256 on Build GRCh38
rs769786148 Chr.5: 462326-462326 on Build GRCh38
rs894022430 Chr.5: 462144-462144 on Build GRCh38
rs774844859 Chr.5: 462086-462086 on Build GRCh38
rs774011095 Chr.5: 462077-462077 on Build GRCh38
rs765128719 Chr.5: 462140-462140 on Build GRCh38
rs370047867 Chr.5: 462080-462080 on Build GRCh38
rs776670266 Chr.5: 462126-462126 on Build GRCh38
rs770185735 Chr.5: 461941-461941 on Build GRCh38
rs767806146 Chr.5: 462254-462254 on Build GRCh38
rs376838648 Chr.5: 462023-462023 on Build GRCh38
rs754386448 Chr.5: 462204-462204 on Build GRCh38
rs896590272 Chr.5: 462203-462203 on Build GRCh38
rs867277355 Chr.5: 462255-462255 on Build GRCh38
rs373753725 Chr.5: 462095-462095 on Build GRCh38
rs756621662 Chr.5: 462105-462105 on Build GRCh38
rs770675110 Chr.5: 462337-462337 on Build GRCh38
rs755045310 Chr.5: 462040-462040 on Build GRCh38
rs771658490 Chr.5: 462083-462083 on Build GRCh38
rs575131206 Chr.5: 462125-462125 on Build GRCh38
rs1015616434 Chr.5: 462212-462212 on Build GRCh38
rs770586478 Chr.5: 462070-462070 on Build GRCh38
rs752411447 Chr.5: 462324-462325 on Build GRCh38
rs556077599 Chr.5: 462227-462227 on Build GRCh38
rs758696298 Chr.5: 462021-462021 on Build GRCh38
rs753906965 Chr.5: 462263-462263 on Build GRCh38
rs192396555 Chr.5: 461975-461975 on Build GRCh38
rs767569822 Chr.5: 462154-462154 on Build GRCh38
rs764053862 Chr.5: 462094-462094 on Build GRCh38
rs753678939 Chr.5: 462017-462017 on Build GRCh38
rs779522652 Chr.5: 462220-462220 on Build GRCh38
rs759871234 Chr.5: 462148-462148 on Build GRCh38
rs539451787 Chr.5: 462007-462007 on Build GRCh38
rs1034983336 Chr.5: 461935-461935 on Build GRCh38
rs749679505 Chr.5: 462112-462112 on Build GRCh38
rs776701887 Chr.5: 462345-462345 on Build GRCh38
rs755535337 Chr.5: 462329-462329 on Build GRCh38
rs183905893 Chr.5: 462044-462044 on Build GRCh38
rs373334871 Chr.5: 461911-461911 on Build GRCh38
rs372707852 Chr.5: 462251-462251 on Build GRCh38
rs771617010 Chr.5: 462118-462118 on Build GRCh38
rs745588491 Chr.5: 462117-462117 on Build GRCh38
rs757743642 Chr.5: 462205-462205 on Build GRCh38
rs781391611 Chr.5: 462103-462103 on Build GRCh38
rs757582135 Chr.5: 462113-462113 on Build GRCh38
rs377494437 Chr.5: 461947-461947 on Build GRCh38
rs779396819 Chr.5: 462210-462210 on Build GRCh38
rs760860067 Chr.5: 462257-462257 on Build GRCh38
rs759003166 Chr.5: 462082-462082 on Build GRCh38
rs367875026 Chr.5: 462102-462102 on Build GRCh38
rs375770333 Chr.5: 462273-462273 on Build GRCh38
rs780916912 Chr.5: 462320-462320 on Build GRCh38
rs760503809 Chr.5: 461982-461982 on Build GRCh38
rs886946471 Chr.5: 462092-462092 on Build GRCh38
rs371766523 Chr.5: 462120-462120 on Build GRCh38
rs760759046 Chr.5: 462087-462087 on Build GRCh38
rs764619730 Chr.5: 462187-462187 on Build GRCh38
rs780433692 Chr.5: 462291-462291 on Build GRCh38
rs773857614 Chr.5: 461968-461968 on Build GRCh38
rs746548341 Chr.5: 462224-462224 on Build GRCh38
rs890972 Chr.5: 462350-462350 on Build GRCh38
rs867803757 Chr.5: 461961-461961 on Build GRCh38
rs754675803 Chr.5: 462065-462066 on Build GRCh38
rs746149777 Chr.5: 462214-462214 on Build GRCh38
rs747595341 Chr.5: 462323-462323 on Build GRCh38
rs752716798 Chr.5: 461981-461981 on Build GRCh38
rs751460700 Chr.5: 461951-461952 on Build GRCh38
rs758648199 Chr.5: 462282-462282 on Build GRCh38
rs1019013793 Chr.5: 462262-462262 on Build GRCh38
rs764212752 Chr.5: 462258-462258 on Build GRCh38
rs987621686 Chr.5: 462247-462247 on Build GRCh38
rs374688550 Chr.5: 462041-462041 on Build GRCh38
rs779121173 Chr.5: 462114-462114 on Build GRCh38
rs772871065 Chr.5: 462143-462143 on Build GRCh38
rs765603186 Chr.5: 462274-462274 on Build GRCh38
rs751305470 Chr.5: 462310-462310 on Build GRCh38
rs748491930 Chr.5: 461940-461940 on Build GRCh38
rs762301900 Chr.5: 462026-462026 on Build GRCh38
rs761810326 Chr.5: 462138-462138 on Build GRCh38
rs1002302504 Chr.5: 461895-461895 on Build GRCh38
rs775067205 Chr.5: 462119-462119 on Build GRCh38
rs914111346 Chr.5: 462101-462101 on Build GRCh38
rs774114571 Chr.5: 462338-462338 on Build GRCh38
rs778178495 Chr.5: 462047-462047 on Build GRCh38
rs906305158 Chr.5: 462110-462110 on Build GRCh38
rs750819644 Chr.5: 462276-462276 on Build GRCh38
rs767212984 Chr.5: 462091-462092 on Build GRCh38
rs370666944 Chr.5: 461953-461953 on Build GRCh38
rs374263956 Chr.5: 462184-462184 on Build GRCh38
rs371456245 Chr.5: 462027-462027 on Build GRCh38
rs920594476 Chr.5: 462347-462347 on Build GRCh38
rs778198726 Chr.5: 462108-462108 on Build GRCh38
rs766055431 Chr.5: 461914-461914 on Build GRCh38

View Full Product Details