Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 10258026-10258527 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCT5
Entrez Gene ID
22948
Gene Name
chaperonin containing TCP1 subunit 5
Gene Aliases
CCT-epsilon, CCTE, HEL-S-69, PNAS-102, TCP-1-epsilon
UniGene
Hs.1600
Gene Chromosome Location
Chr.5: 10249921-10266412 on Build GRCh38
Transcripts
NM_001306153.1 NM_001306154.1 NM_001306155.1 NM_001306156.1 NM_012073.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4749834 c.682G>A p.V228M Chr.5: 10258262-10258262 on Build GRCh38
6220226 c.841G>A p.E281K Chr.5: 10258503-10258503 on Build GRCh38
1059308 c.681C>T p.G227G Chr.5: 10258261-10258261 on Build GRCh38
6101463 c.741G>C p.K247N Chr.5: 10258403-10258403 on Build GRCh38
7283221 c.601C>T p.R201W Chr.5: 10258181-10258181 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs555534209 Chr.5: 10258108-10258108 on Build GRCh38
rs754962890 Chr.5: 10258498-10258498 on Build GRCh38
rs200819422 Chr.5: 10258410-10258410 on Build GRCh38
rs774961022 Chr.5: 10258424-10258424 on Build GRCh38
rs780247734 Chr.5: 10258368-10258368 on Build GRCh38
rs758906555 Chr.5: 10258418-10258418 on Build GRCh38
rs367653103 Chr.5: 10258201-10258201 on Build GRCh38
rs989835897 Chr.5: 10258468-10258468 on Build GRCh38
rs370943521 Chr.5: 10258098-10258098 on Build GRCh38
rs1020599801 Chr.5: 10258475-10258475 on Build GRCh38
rs749838217 Chr.5: 10258070-10258070 on Build GRCh38
rs781347459 Chr.5: 10258476-10258476 on Build GRCh38
rs766038337 Chr.5: 10258159-10258159 on Build GRCh38
rs770463902 Chr.5: 10258280-10258280 on Build GRCh38
rs764583584 Chr.5: 10258218-10258218 on Build GRCh38
rs751271784 Chr.5: 10258176-10258176 on Build GRCh38
rs762850699 Chr.5: 10258272-10258272 on Build GRCh38
rs758504808 Chr.5: 10258255-10258255 on Build GRCh38
rs754551015 Chr.5: 10258198-10258198 on Build GRCh38
rs759287448 Chr.5: 10258301-10258301 on Build GRCh38
rs762233070 Chr.5: 10258329-10258329 on Build GRCh38
rs757831877 Chr.5: 10258072-10258072 on Build GRCh38
rs57137339 Chr.5: 10258331-10258331 on Build GRCh38
rs753662828 Chr.5: 10258087-10258087 on Build GRCh38
rs372716481 Chr.5: 10258364-10258364 on Build GRCh38
rs765737541 Chr.5: 10258440-10258440 on Build GRCh38
rs145578766 Chr.5: 10258231-10258231 on Build GRCh38
rs376225755 Chr.5: 10258081-10258081 on Build GRCh38
rs769754440 Chr.5: 10258267-10258267 on Build GRCh38
rs11557655 Chr.5: 10258299-10258299 on Build GRCh38
rs201804455 Chr.5: 10258229-10258229 on Build GRCh38
rs925661175 Chr.5: 10258250-10258250 on Build GRCh38
rs753033282 Chr.5: 10258060-10258060 on Build GRCh38
rs767023577 Chr.5: 10258310-10258310 on Build GRCh38
rs11557649 Chr.5: 10258400-10258400 on Build GRCh38
rs745979403 Chr.5: 10258247-10258247 on Build GRCh38
rs773084151 Chr.5: 10258268-10258268 on Build GRCh38
rs967299283 Chr.5: 10258478-10258478 on Build GRCh38
rs775816493 Chr.5: 10258085-10258085 on Build GRCh38
rs372629115 Chr.5: 10258295-10258295 on Build GRCh38
rs368338119 Chr.5: 10258262-10258262 on Build GRCh38
rs898527531 Chr.5: 10258241-10258241 on Build GRCh38
rs539649252 Chr.5: 10258082-10258082 on Build GRCh38
rs750814476 Chr.5: 10258449-10258449 on Build GRCh38
rs763613936 Chr.5: 10258471-10258471 on Build GRCh38
rs771812955 Chr.5: 10258374-10258374 on Build GRCh38
rs542323956 Chr.5: 10258421-10258421 on Build GRCh38
rs755231384 Chr.5: 10258377-10258377 on Build GRCh38
rs766945390 Chr.5: 10258185-10258185 on Build GRCh38
rs758298076 Chr.5: 10258312-10258312 on Build GRCh38
rs746803733 Chr.5: 10258091-10258091 on Build GRCh38
rs768544770 Chr.5: 10258093-10258093 on Build GRCh38
rs751689306 Chr.5: 10258479-10258479 on Build GRCh38
rs922131591 Chr.5: 10258289-10258289 on Build GRCh38
rs755384536 Chr.5: 10258415-10258415 on Build GRCh38
rs754237380 Chr.5: 10258313-10258313 on Build GRCh38
rs762408037 Chr.5: 10258156-10258156 on Build GRCh38
rs886422880 Chr.5: 10258376-10258376 on Build GRCh38
rs754143874 Chr.5: 10258228-10258228 on Build GRCh38
rs932840381 Chr.5: 10258086-10258086 on Build GRCh38
rs542783054 Chr.5: 10258163-10258163 on Build GRCh38
rs781230840 Chr.5: 10258380-10258380 on Build GRCh38
rs761866534 Chr.5: 10258101-10258101 on Build GRCh38
rs769622374 Chr.5: 10258399-10258399 on Build GRCh38
rs144280885 Chr.5: 10258406-10258406 on Build GRCh38
rs747958037 Chr.5: 10258397-10258397 on Build GRCh38
rs563335974 Chr.5: 10258066-10258066 on Build GRCh38
rs933579352 Chr.5: 10258328-10258328 on Build GRCh38
rs768757632 Chr.5: 10258257-10258257 on Build GRCh38
rs535459086 Chr.5: 10258261-10258261 on Build GRCh38
rs978444316 Chr.5: 10258157-10258157 on Build GRCh38
rs772711272 Chr.5: 10258128-10258128 on Build GRCh38
rs11557648 Chr.5: 10258441-10258441 on Build GRCh38
rs772430738 Chr.5: 10258083-10258083 on Build GRCh38

View Full Product Details