Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 10261441-10261953 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCT5
Entrez Gene ID
22948
Gene Name
chaperonin containing TCP1 subunit 5
Gene Aliases
CCT-epsilon, CCTE, HEL-S-69, PNAS-102, TCP-1-epsilon
UniGene
Hs.1600
Gene Chromosome Location
Chr.5: 10249921-10266412 on Build GRCh38
Transcripts
NM_001306153.1 NM_001306154.1 NM_001306155.1 NM_001306156.1 NM_012073.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
319327 c.994-1G>T p.? Chr.5: 10261559-10261559 on Build GRCh38
6623310 c.1001C>T p.A334V Chr.5: 10261567-10261567 on Build GRCh38
3993883 c.1073T>G p.L358R Chr.5: 10261639-10261639 on Build GRCh38
1059310 c.1061G>A p.G354D Chr.5: 10261627-10261627 on Build GRCh38
7360165 c.1086C>G p.I362M Chr.5: 10261652-10261652 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147989324 Chr.5: 10261641-10261641 on Build GRCh38
rs773314762 Chr.5: 10261698-10261698 on Build GRCh38
rs377234917 Chr.5: 10261727-10261727 on Build GRCh38
rs758282324 Chr.5: 10261513-10261513 on Build GRCh38
rs750220310 Chr.5: 10261511-10261511 on Build GRCh38
rs776768547 Chr.5: 10261583-10261583 on Build GRCh38
rs756903385 Chr.5: 10261822-10261822 on Build GRCh38
rs1016532745 Chr.5: 10261611-10261611 on Build GRCh38
rs780454592 Chr.5: 10261826-10261826 on Build GRCh38
rs761888593 Chr.5: 10261661-10261661 on Build GRCh38
rs769955803 Chr.5: 10261858-10261858 on Build GRCh38
rs913771814 Chr.5: 10261719-10261719 on Build GRCh38
rs111255743 Chr.5: 10261575-10261575 on Build GRCh38
rs755604055 Chr.5: 10261794-10261794 on Build GRCh38
rs781696673 Chr.5: 10261856-10261856 on Build GRCh38
rs114634318 Chr.5: 10261625-10261625 on Build GRCh38
rs1040445862 Chr.5: 10261818-10261818 on Build GRCh38
rs751586130 Chr.5: 10261616-10261616 on Build GRCh38
rs765442933 Chr.5: 10261595-10261595 on Build GRCh38
rs780097467 Chr.5: 10261521-10261521 on Build GRCh38
rs768768696 Chr.5: 10261572-10261572 on Build GRCh38
rs747585890 Chr.5: 10261654-10261654 on Build GRCh38
rs750191898 Chr.5: 10261605-10261605 on Build GRCh38
rs746173214 Chr.5: 10261555-10261555 on Build GRCh38
rs762786367 Chr.5: 10261612-10261612 on Build GRCh38
rs747569772 Chr.5: 10261473-10261473 on Build GRCh38
rs771215606 Chr.5: 10261872-10261872 on Build GRCh38
rs1030087457 Chr.5: 10261901-10261901 on Build GRCh38
rs754583997 Chr.5: 10261530-10261530 on Build GRCh38
rs886572164 Chr.5: 10261913-10261913 on Build GRCh38
rs140095139 Chr.5: 10261617-10261617 on Build GRCh38
rs747317644 Chr.5: 10261853-10261853 on Build GRCh38
rs143444882 Chr.5: 10261636-10261636 on Build GRCh38
rs951622250 Chr.5: 10261608-10261608 on Build GRCh38
rs199805135 Chr.5: 10261613-10261613 on Build GRCh38
rs532046384 Chr.5: 10261888-10261888 on Build GRCh38
rs763264040 Chr.5: 10261756-10261756 on Build GRCh38
rs775929559 Chr.5: 10261560-10261560 on Build GRCh38
rs550566762 Chr.5: 10261923-10261923 on Build GRCh38
rs997301746 Chr.5: 10261488-10261488 on Build GRCh38
rs759362220 Chr.5: 10261878-10261878 on Build GRCh38
rs777440322 Chr.5: 10261638-10261638 on Build GRCh38
rs777159469 Chr.5: 10261590-10261590 on Build GRCh38
rs767375395 Chr.5: 10261894-10261894 on Build GRCh38
rs770267509 Chr.5: 10261782-10261782 on Build GRCh38
rs146087813 Chr.5: 10261871-10261872 on Build GRCh38
rs2244717 Chr.5: 10261854-10261854 on Build GRCh38
rs773531721 Chr.5: 10261688-10261688 on Build GRCh38
rs772476426 Chr.5: 10261649-10261649 on Build GRCh38
rs769126593 Chr.5: 10261478-10261478 on Build GRCh38
rs936622660 Chr.5: 10261864-10261864 on Build GRCh38
rs759556512 Chr.5: 10261742-10261742 on Build GRCh38
rs943014723 Chr.5: 10261811-10261811 on Build GRCh38
rs748579621 Chr.5: 10261857-10261857 on Build GRCh38
rs769929695 Chr.5: 10261687-10261687 on Build GRCh38
rs375319644 Chr.5: 10261535-10261535 on Build GRCh38
rs998269335 Chr.5: 10261584-10261584 on Build GRCh38
rs11557657 Chr.5: 10261730-10261730 on Build GRCh38
rs552462975 Chr.5: 10261925-10261925 on Build GRCh38
rs892350838 Chr.5: 10261467-10261467 on Build GRCh38
rs776587777 Chr.5: 10261657-10261657 on Build GRCh38
rs749578962 Chr.5: 10261868-10261868 on Build GRCh38
rs772601071 Chr.5: 10261556-10261556 on Build GRCh38
rs773610039 Chr.5: 10261828-10261829 on Build GRCh38
rs761152363 Chr.5: 10261565-10261565 on Build GRCh38
rs773160689 Chr.5: 10261860-10261860 on Build GRCh38
rs539339127 Chr.5: 10261554-10261554 on Build GRCh38
rs1052278149 Chr.5: 10261464-10261464 on Build GRCh38
rs946603531 Chr.5: 10261725-10261725 on Build GRCh38
rs141675330 Chr.5: 10261652-10261652 on Build GRCh38
rs752189712 Chr.5: 10261634-10261634 on Build GRCh38
rs139449863 Chr.5: 10261689-10261689 on Build GRCh38
rs751669557 Chr.5: 10261875-10261875 on Build GRCh38
rs1039094195 Chr.5: 10261866-10261866 on Build GRCh38
rs368250896 Chr.5: 10261644-10261644 on Build GRCh38
rs780816888 Chr.5: 10261531-10261531 on Build GRCh38
rs917728034 Chr.5: 10261787-10261787 on Build GRCh38
rs752178580 Chr.5: 10261782-10261782 on Build GRCh38
rs368816698 Chr.5: 10261769-10261769 on Build GRCh38
rs753533342 Chr.5: 10261796-10261796 on Build GRCh38
rs746538247 Chr.5: 10261525-10261525 on Build GRCh38
rs1043777692 Chr.5: 10261512-10261512 on Build GRCh38
rs769470413 Chr.5: 10261543-10261543 on Build GRCh38
rs190901402 Chr.5: 10261466-10261466 on Build GRCh38
rs765584260 Chr.5: 10261656-10261656 on Build GRCh38
rs182779563 Chr.5: 10261873-10261873 on Build GRCh38

View Full Product Details