Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 10760779-10761306 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DAP
Entrez Gene ID
1611
Gene Name
death associated protein
Gene Aliases
-
UniGene
Hs.75189
Gene Chromosome Location
Chr.5: 10679230-10761275 on Build GRCh38
Transcripts
NM_001291963.1 NM_004394.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5508944 c.44A>C p.H15P Chr.5: 10761025-10761025 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs966021064 Chr.5: 10761212-10761212 on Build GRCh38
rs535471661 Chr.5: 10760936-10760936 on Build GRCh38
rs998333578 Chr.5: 10761217-10761217 on Build GRCh38
rs369001210 Chr.5: 10761033-10761033 on Build GRCh38
rs976903083 Chr.5: 10760934-10760934 on Build GRCh38
rs768231070 Chr.5: 10761063-10761063 on Build GRCh38
rs904222978 Chr.5: 10761201-10761201 on Build GRCh38
rs969322765 Chr.5: 10761238-10761238 on Build GRCh38
rs955765457 Chr.5: 10760826-10760826 on Build GRCh38
rs551186435 Chr.5: 10760819-10760819 on Build GRCh38
rs575523678 Chr.5: 10760994-10760994 on Build GRCh38
rs775921278 Chr.5: 10761082-10761082 on Build GRCh38
rs866975493 Chr.5: 10761263-10761263 on Build GRCh38
rs985733833 Chr.5: 10761059-10761059 on Build GRCh38
rs999433200 Chr.5: 10760841-10760841 on Build GRCh38
rs1032672532 Chr.5: 10760851-10760851 on Build GRCh38
rs147507510 Chr.5: 10761231-10761231 on Build GRCh38
rs770017957 Chr.5: 10761000-10761001 on Build GRCh38
rs945771129 Chr.5: 10761265-10761265 on Build GRCh38
rs773244654 Chr.5: 10761072-10761072 on Build GRCh38
rs776377093 Chr.5: 10761065-10761065 on Build GRCh38
rs1002648252 Chr.5: 10760808-10760808 on Build GRCh38
rs562325050 Chr.5: 10761258-10761258 on Build GRCh38
rs201354802 Chr.5: 10761041-10761041 on Build GRCh38
rs1013558088 Chr.5: 10760835-10760835 on Build GRCh38
rs780067899 Chr.5: 10761056-10761056 on Build GRCh38
rs895205331 Chr.5: 10760824-10760824 on Build GRCh38
rs778935780 Chr.5: 10760967-10760967 on Build GRCh38
rs891521085 Chr.5: 10761186-10761186 on Build GRCh38
rs957921816 Chr.5: 10760886-10760886 on Build GRCh38
rs760011276 Chr.5: 10761025-10761025 on Build GRCh38
rs909885948 Chr.5: 10761233-10761233 on Build GRCh38
rs544175556 Chr.5: 10761031-10761031 on Build GRCh38
rs905778966 Chr.5: 10761169-10761169 on Build GRCh38
rs920144076 Chr.5: 10760811-10760811 on Build GRCh38
rs929960313 Chr.5: 10760942-10760942 on Build GRCh38
rs753926112 Chr.5: 10761098-10761098 on Build GRCh38
rs539601049 Chr.5: 10760871-10760871 on Build GRCh38
rs1018856398 Chr.5: 10761060-10761060 on Build GRCh38
rs776363765 Chr.5: 10761006-10761006 on Build GRCh38
rs760768339 Chr.5: 10761089-10761089 on Build GRCh38
rs774636302 Chr.5: 10761078-10761078 on Build GRCh38
rs537331369 Chr.5: 10760817-10760817 on Build GRCh38
rs1003025609 Chr.5: 10760975-10760976 on Build GRCh38
rs1007145601 Chr.5: 10761203-10761203 on Build GRCh38
rs938495726 Chr.5: 10761177-10761177 on Build GRCh38
rs868793253 Chr.5: 10760907-10760907 on Build GRCh38
rs984060894 Chr.5: 10761143-10761143 on Build GRCh38
rs765488331 Chr.5: 10761113-10761113 on Build GRCh38
rs1048386582 Chr.5: 10760943-10760943 on Build GRCh38
rs766612036 Chr.5: 10761010-10761010 on Build GRCh38
rs981189479 Chr.5: 10761256-10761256 on Build GRCh38
rs753961777 Chr.5: 10761040-10761040 on Build GRCh38
rs763398169 Chr.5: 10761007-10761007 on Build GRCh38
rs1010013910 Chr.5: 10761189-10761189 on Build GRCh38
rs968977330 Chr.5: 10760887-10760887 on Build GRCh38
rs902447316 Chr.5: 10760840-10760840 on Build GRCh38
rs761995566 Chr.5: 10761105-10761105 on Build GRCh38
rs10037087 Chr.5: 10760959-10760959 on Build GRCh38
rs931453407 Chr.5: 10760815-10760815 on Build GRCh38
rs984206622 Chr.5: 10761232-10761232 on Build GRCh38
rs542644316 Chr.5: 10761257-10761257 on Build GRCh38
rs772108942 Chr.5: 10760991-10760991 on Build GRCh38
rs1035410156 Chr.5: 10760992-10760992 on Build GRCh38
rs1018487992 Chr.5: 10761210-10761210 on Build GRCh38
rs1042731625 Chr.5: 10761197-10761197 on Build GRCh38
rs550997909 Chr.5: 10761278-10761278 on Build GRCh38
rs758206159 Chr.5: 10761120-10761120 on Build GRCh38
rs941968649 Chr.5: 10760953-10760953 on Build GRCh38
rs572971500 Chr.5: 10760931-10760931 on Build GRCh38
rs909917101 Chr.5: 10761144-10761144 on Build GRCh38
rs745658094 Chr.5: 10761048-10761048 on Build GRCh38
rs560313097 Chr.5: 10761229-10761229 on Build GRCh38
rs758474315 Chr.5: 10761051-10761051 on Build GRCh38
rs774748089 Chr.5: 10761021-10761021 on Build GRCh38
rs769466384 Chr.5: 10761077-10761077 on Build GRCh38
rs746530109 Chr.5: 10761058-10761058 on Build GRCh38
rs745755705 Chr.5: 10760977-10760977 on Build GRCh38
rs1044332030 Chr.5: 10761161-10761161 on Build GRCh38
rs987139728 Chr.5: 10760913-10760913 on Build GRCh38
rs1035759505 Chr.5: 10760821-10760821 on Build GRCh38
rs941947304 Chr.5: 10761159-10761159 on Build GRCh38
rs11740244 Chr.5: 10760897-10760897 on Build GRCh38
rs905734061 Chr.5: 10760961-10760961 on Build GRCh38
rs540383972 Chr.5: 10761079-10761079 on Build GRCh38
rs568752790 Chr.5: 10760810-10760810 on Build GRCh38
rs768425295 Chr.5: 10761004-10761004 on Build GRCh38
rs267928 Chr.5: 10760838-10760838 on Build GRCh38
rs267927 Chr.5: 10761070-10761070 on Build GRCh38
rs887674188 Chr.5: 10760945-10760945 on Build GRCh38
rs772574208 Chr.5: 10761080-10761080 on Build GRCh38
rs996400872 Chr.5: 10760889-10760889 on Build GRCh38
rs764316925 Chr.5: 10761038-10761038 on Build GRCh38
rs946673714 Chr.5: 10761261-10761271 on Build GRCh38
rs778618456 Chr.5: 10761042-10761042 on Build GRCh38
rs1009882050 Chr.5: 10761039-10761039 on Build GRCh38
rs756123267 Chr.5: 10761035-10761035 on Build GRCh38
rs746509981 Chr.5: 10760998-10760998 on Build GRCh38
rs930062702 Chr.5: 10761096-10761096 on Build GRCh38
rs531274676 Chr.5: 10761264-10761264 on Build GRCh38

View Full Product Details