Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 10971759-10972269 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTNND2
Entrez Gene ID
1501
Gene Name
catenin delta 2
Gene Aliases
GT24, NPRAP
UniGene
Hs.314543
Gene Chromosome Location
Chr.5: 10971840-11904067 on Build GRCh38
Transcripts
NM_001288715.1 NM_001288716.1 NM_001288717.1 NM_001332.3 NR_109988.1

SNP Detail

SNP ID Chromosome Location
rs867169195 Chr.5: 10971943-10971943 on Build GRCh38
rs901604646 Chr.5: 10972194-10972194 on Build GRCh38
rs928752794 Chr.5: 10972230-10972230 on Build GRCh38
rs961500922 Chr.5: 10971902-10971902 on Build GRCh38
rs75879762 Chr.5: 10972228-10972228 on Build GRCh38
rs998631816 Chr.5: 10972002-10972002 on Build GRCh38
rs948850081 Chr.5: 10972081-10972081 on Build GRCh38
rs902983761 Chr.5: 10971790-10971790 on Build GRCh38
rs576079733 Chr.5: 10972227-10972227 on Build GRCh38
rs928698598 Chr.5: 10972102-10972102 on Build GRCh38
rs1023310869 Chr.5: 10972089-10972089 on Build GRCh38
rs776297486 Chr.5: 10971900-10971900 on Build GRCh38
rs762702164 Chr.5: 10972032-10972032 on Build GRCh38
rs1005984467 Chr.5: 10972101-10972101 on Build GRCh38
rs1012860021 Chr.5: 10971834-10971834 on Build GRCh38
rs934637222 Chr.5: 10971940-10971940 on Build GRCh38
rs961429164 Chr.5: 10972125-10972125 on Build GRCh38
rs888923311 Chr.5: 10971998-10971998 on Build GRCh38
rs972317635 Chr.5: 10971824-10971824 on Build GRCh38
rs932886148 Chr.5: 10971937-10971937 on Build GRCh38
rs558184039 Chr.5: 10971810-10971810 on Build GRCh38
rs948796529 Chr.5: 10971910-10971910 on Build GRCh38
rs866718923 Chr.5: 10972178-10972178 on Build GRCh38
rs852601 Chr.5: 10971945-10971945 on Build GRCh38
rs545492105 Chr.5: 10972090-10972090 on Build GRCh38
rs1022139674 Chr.5: 10971977-10971977 on Build GRCh38
rs537489756 Chr.5: 10972190-10972190 on Build GRCh38
rs1035575780 Chr.5: 10971823-10971823 on Build GRCh38
rs867877925 Chr.5: 10972054-10972054 on Build GRCh38
rs371570303 Chr.5: 10972211-10972211 on Build GRCh38
rs555702537 Chr.5: 10971959-10971959 on Build GRCh38
rs908621540 Chr.5: 10971901-10971901 on Build GRCh38
rs192063416 Chr.5: 10972075-10972075 on Build GRCh38
rs564174684 Chr.5: 10972128-10972128 on Build GRCh38
rs1007339220 Chr.5: 10972162-10972162 on Build GRCh38
rs1053131688 Chr.5: 10972112-10972112 on Build GRCh38
rs1022961020 Chr.5: 10971857-10971857 on Build GRCh38
rs769397361 Chr.5: 10971934-10971934 on Build GRCh38
rs956119841 Chr.5: 10972247-10972247 on Build GRCh38
rs747733533 Chr.5: 10971913-10971913 on Build GRCh38
rs10075852 Chr.5: 10972229-10972229 on Build GRCh38
rs780469853 Chr.5: 10971793-10971793 on Build GRCh38
rs573924831 Chr.5: 10972004-10972004 on Build GRCh38
rs868394291 Chr.5: 10972225-10972225 on Build GRCh38
rs567307358 Chr.5: 10972115-10972115 on Build GRCh38
rs530149009 Chr.5: 10971941-10971941 on Build GRCh38
rs922803133 Chr.5: 10971922-10971922 on Build GRCh38
rs984980077 Chr.5: 10971893-10971893 on Build GRCh38
rs985418892 Chr.5: 10972055-10972055 on Build GRCh38
rs768235307 Chr.5: 10971852-10971852 on Build GRCh38
rs147003964 Chr.5: 10972011-10972011 on Build GRCh38
rs976829039 Chr.5: 10972195-10972195 on Build GRCh38
rs965664903 Chr.5: 10971798-10971798 on Build GRCh38
rs888977359 Chr.5: 10972147-10972147 on Build GRCh38
rs1050006035 Chr.5: 10971939-10971939 on Build GRCh38
rs571704003 Chr.5: 10972080-10972080 on Build GRCh38
rs922771789 Chr.5: 10972204-10972204 on Build GRCh38
rs537614614 Chr.5: 10971958-10971958 on Build GRCh38
rs916102617 Chr.5: 10972065-10972065 on Build GRCh38
rs61752754 Chr.5: 10972053-10972053 on Build GRCh38
rs1040137684 Chr.5: 10972163-10972163 on Build GRCh38
rs902951747 Chr.5: 10971989-10971989 on Build GRCh38
rs1025993741 Chr.5: 10972220-10972220 on Build GRCh38
rs1044017575 Chr.5: 10972127-10972127 on Build GRCh38
rs747554257 Chr.5: 10971830-10971830 on Build GRCh38
rs952857412 Chr.5: 10971871-10971871 on Build GRCh38
rs946589318 Chr.5: 10972102-10972116 on Build GRCh38
rs1056966203 Chr.5: 10972008-10972008 on Build GRCh38
rs1042775621 Chr.5: 10971952-10971952 on Build GRCh38
rs1045828021 Chr.5: 10971914-10971914 on Build GRCh38

View Full Product Details