Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 10981622-10982120 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTNND2
Entrez Gene ID
1501
Gene Name
catenin delta 2
Gene Aliases
GT24, NPRAP
UniGene
Hs.314543
Gene Chromosome Location
Chr.5: 10971840-11904067 on Build GRCh38
Transcripts
NM_001288715.1 NM_001288716.1 NM_001288717.1 NM_001332.3 NR_109988.1 XM_005248251.3 XM_005248252.2 XM_005248253.1 XM_011513967.2 XM_017009072.1 XM_017009073.1 XM_017009074.1 XM_017009075.1 XM_017009076.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6206184 c.3352A>G p.T1118A Chr.5: 10981838-10981838 on Build GRCh38
4410546 c.3357A>G p.G1119G Chr.5: 10981833-10981833 on Build GRCh38
359997 c.3402C>T p.D1134D Chr.5: 10981788-10981788 on Build GRCh38
1310427 c.3369G>A p.L1123L Chr.5: 10981821-10981821 on Build GRCh38
236080 c.3417G>T p.Q1139H Chr.5: 10981773-10981773 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371533826 Chr.5: 10981987-10981987 on Build GRCh38
rs61752667 Chr.5: 10981857-10981857 on Build GRCh38
rs754236397 Chr.5: 10981880-10981880 on Build GRCh38
rs147220528 Chr.5: 10981795-10981795 on Build GRCh38
rs199829191 Chr.5: 10981767-10981768 on Build GRCh38
rs111641324 Chr.5: 10982069-10982069 on Build GRCh38
rs749877746 Chr.5: 10981776-10981776 on Build GRCh38
rs995369540 Chr.5: 10982010-10982010 on Build GRCh38
rs778104952 Chr.5: 10981874-10981874 on Build GRCh38
rs201402185 Chr.5: 10981822-10981822 on Build GRCh38
rs746839369 Chr.5: 10981819-10981819 on Build GRCh38
rs906620229 Chr.5: 10982083-10982083 on Build GRCh38
rs61752666 Chr.5: 10982022-10982022 on Build GRCh38
rs764766060 Chr.5: 10981774-10981774 on Build GRCh38
rs1046853980 Chr.5: 10982070-10982070 on Build GRCh38
rs751125091 Chr.5: 10981861-10981861 on Build GRCh38
rs1000818589 Chr.5: 10981653-10981653 on Build GRCh38
rs959324966 Chr.5: 10982023-10982023 on Build GRCh38
rs761786077 Chr.5: 10981836-10981836 on Build GRCh38
rs536170795 Chr.5: 10981955-10981955 on Build GRCh38
rs763233981 Chr.5: 10981726-10981727 on Build GRCh38
rs1054194070 Chr.5: 10981655-10981655 on Build GRCh38
rs559180150 Chr.5: 10982061-10982061 on Build GRCh38
rs868638164 Chr.5: 10981790-10981790 on Build GRCh38
rs369415203 Chr.5: 10981926-10981926 on Build GRCh38
rs762302358 Chr.5: 10981858-10981858 on Build GRCh38
rs899737412 Chr.5: 10981644-10981644 on Build GRCh38
rs766070070 Chr.5: 10981860-10981860 on Build GRCh38
rs373010050 Chr.5: 10981673-10981676 on Build GRCh38
rs1041822358 Chr.5: 10981811-10981811 on Build GRCh38
rs762724120 Chr.5: 10981787-10981787 on Build GRCh38
rs369587014 Chr.5: 10981808-10981808 on Build GRCh38
rs776498983 Chr.5: 10981762-10981762 on Build GRCh38
rs573198951 Chr.5: 10982089-10982089 on Build GRCh38
rs145274994 Chr.5: 10981780-10981780 on Build GRCh38
rs367742942 Chr.5: 10981755-10981755 on Build GRCh38
rs373647754 Chr.5: 10981744-10981744 on Build GRCh38
rs780077041 Chr.5: 10981818-10981818 on Build GRCh38
rs912296284 Chr.5: 10981973-10981973 on Build GRCh38
rs761154163 Chr.5: 10981867-10981867 on Build GRCh38
rs919552189 Chr.5: 10981934-10981934 on Build GRCh38
rs962569721 Chr.5: 10981727-10981727 on Build GRCh38
rs1029799535 Chr.5: 10981674-10981674 on Build GRCh38
rs187779487 Chr.5: 10981992-10981992 on Build GRCh38
rs546417917 Chr.5: 10981701-10981701 on Build GRCh38
rs764595657 Chr.5: 10981877-10981877 on Build GRCh38
rs61752668 Chr.5: 10981659-10981659 on Build GRCh38
rs761342342 Chr.5: 10981770-10981770 on Build GRCh38
rs749670384 Chr.5: 10981904-10981904 on Build GRCh38
rs879204282 Chr.5: 10981759-10981759 on Build GRCh38
rs936208139 Chr.5: 10981850-10981850 on Build GRCh38
rs371572646 Chr.5: 10981800-10981800 on Build GRCh38
rs1055095440 Chr.5: 10981852-10981852 on Build GRCh38
rs769704199 Chr.5: 10981853-10981853 on Build GRCh38
rs996882379 Chr.5: 10981863-10981863 on Build GRCh38
rs768396526 Chr.5: 10981821-10981821 on Build GRCh38
rs951093160 Chr.5: 10981935-10981935 on Build GRCh38
rs991857611 Chr.5: 10982036-10982036 on Build GRCh38
rs61754547 Chr.5: 10981806-10981806 on Build GRCh38
rs575186578 Chr.5: 10981888-10981888 on Build GRCh38
rs765662790 Chr.5: 10981893-10981893 on Build GRCh38
rs374159285 Chr.5: 10981712-10981712 on Build GRCh38
rs753111084 Chr.5: 10981796-10981796 on Build GRCh38
rs1003957649 Chr.5: 10981966-10981966 on Build GRCh38
rs2228958 Chr.5: 10981809-10981809 on Build GRCh38
rs982916306 Chr.5: 10981954-10981954 on Build GRCh38
rs571416843 Chr.5: 10981797-10981797 on Build GRCh38
rs921137813 Chr.5: 10982026-10982026 on Build GRCh38
rs943795924 Chr.5: 10981977-10981977 on Build GRCh38
rs936533214 Chr.5: 10982053-10982053 on Build GRCh38
rs892430325 Chr.5: 10982068-10982068 on Build GRCh38
rs757809631 Chr.5: 10981801-10981801 on Build GRCh38
rs917244002 Chr.5: 10982060-10982060 on Build GRCh38
rs866148702 Chr.5: 10981828-10981828 on Build GRCh38

View Full Product Details