Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 13690881-13691423 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAH5
Entrez Gene ID
1767
Gene Name
dynein axonemal heavy chain 5
Gene Aliases
CILD3, DNAHC5, HL1, KTGNR, PCD
UniGene
Hs.212360
Gene Chromosome Location
Chr.5: 13690328-14011829 on Build GRCh38
Transcripts
NM_001369.2 XM_005248262.3 XM_017009177.1 XM_017009178.1 XM_017009179.1 XM_017009185.1 XM_017009186.1 XM_017009188.1

SNP Detail

SNP ID Chromosome Location
rs753667496 Chr.5: 13691124-13691125 on Build GRCh38
rs184119517 Chr.5: 13690986-13690986 on Build GRCh38
rs983453056 Chr.5: 13691150-13691150 on Build GRCh38
rs1039255620 Chr.5: 13691108-13691108 on Build GRCh38
rs952142607 Chr.5: 13691147-13691147 on Build GRCh38
rs932494192 Chr.5: 13690949-13690949 on Build GRCh38
rs532882575 Chr.5: 13691080-13691080 on Build GRCh38
rs1016690439 Chr.5: 13691277-13691277 on Build GRCh38
rs146040448 Chr.5: 13691209-13691209 on Build GRCh38
rs1024219283 Chr.5: 13691208-13691208 on Build GRCh38
rs77289648 Chr.5: 13691335-13691335 on Build GRCh38
rs750104927 Chr.5: 13691100-13691100 on Build GRCh38
rs972466962 Chr.5: 13691019-13691019 on Build GRCh38
rs891770594 Chr.5: 13691167-13691167 on Build GRCh38
rs556194663 Chr.5: 13691165-13691165 on Build GRCh38
rs893593258 Chr.5: 13690980-13690980 on Build GRCh38
rs914976995 Chr.5: 13691266-13691266 on Build GRCh38
rs370671074 Chr.5: 13690926-13690926 on Build GRCh38
rs2896111 Chr.5: 13690936-13690936 on Build GRCh38
rs990989406 Chr.5: 13691117-13691117 on Build GRCh38
rs979941002 Chr.5: 13690940-13690940 on Build GRCh38
rs907843260 Chr.5: 13691157-13691157 on Build GRCh38
rs972489046 Chr.5: 13691287-13691287 on Build GRCh38
rs909632608 Chr.5: 13690962-13690962 on Build GRCh38
rs756162621 Chr.5: 13691136-13691136 on Build GRCh38
rs902150578 Chr.5: 13691324-13691324 on Build GRCh38
rs779396186 Chr.5: 13691301-13691301 on Build GRCh38
rs962398942 Chr.5: 13691012-13691012 on Build GRCh38
rs904693129 Chr.5: 13691111-13691111 on Build GRCh38
rs551833742 Chr.5: 13691001-13691001 on Build GRCh38
rs918232808 Chr.5: 13691031-13691031 on Build GRCh38
rs566197457 Chr.5: 13690938-13690938 on Build GRCh38
rs933671345 Chr.5: 13691060-13691060 on Build GRCh38
rs1003220552 Chr.5: 13691333-13691333 on Build GRCh38
rs902748089 Chr.5: 13691023-13691023 on Build GRCh38
rs962533590 Chr.5: 13691285-13691285 on Build GRCh38
rs1014143705 Chr.5: 13690919-13690919 on Build GRCh38
rs1025840401 Chr.5: 13691293-13691293 on Build GRCh38
rs1001410221 Chr.5: 13691275-13691275 on Build GRCh38
rs886059932 Chr.5: 13691182-13691182 on Build GRCh38
rs531264344 Chr.5: 13690918-13690918 on Build GRCh38
rs932481352 Chr.5: 13691224-13691224 on Build GRCh38
rs1014693228 Chr.5: 13691198-13691198 on Build GRCh38
rs868041475 Chr.5: 13691091-13691091 on Build GRCh38
rs764924502 Chr.5: 13691093-13691093 on Build GRCh38
rs569999464 Chr.5: 13691097-13691097 on Build GRCh38
rs1049581524 Chr.5: 13691265-13691265 on Build GRCh38
rs1042159329 Chr.5: 13691322-13691322 on Build GRCh38
rs570689259 Chr.5: 13691025-13691025 on Build GRCh38
rs1049427705 Chr.5: 13690968-13690968 on Build GRCh38
rs142535984 Chr.5: 13691103-13691103 on Build GRCh38
rs562393427 Chr.5: 13690911-13690911 on Build GRCh38
rs565332942 Chr.5: 13690989-13690989 on Build GRCh38

View Full Product Details