Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 14693169-14693671 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OTULIN
Entrez Gene ID
90268
Gene Name
OTU deubiquitinase with linear linkage specificity
Gene Aliases
FAM105B, GUM
UniGene
Hs.406335
Gene Chromosome Location
Chr.5: 14660794-14716552 on Build GRCh38
Transcripts
NM_138348.4 XM_011514151.2 XM_011514154.2 XM_017010015.1

SNP Detail

SNP ID Chromosome Location
rs557054061 Chr.5: 14693330-14693330 on Build GRCh38
rs774068010 Chr.5: 14693249-14693249 on Build GRCh38
rs975115847 Chr.5: 14693463-14693463 on Build GRCh38
rs577859773 Chr.5: 14693260-14693260 on Build GRCh38
rs987503007 Chr.5: 14693413-14693413 on Build GRCh38
rs17363829 Chr.5: 14693289-14693289 on Build GRCh38
rs886684496 Chr.5: 14693594-14693594 on Build GRCh38
rs945315255 Chr.5: 14693454-14693454 on Build GRCh38
rs926159606 Chr.5: 14693235-14693235 on Build GRCh38
rs1001899746 Chr.5: 14693334-14693334 on Build GRCh38
rs1049730557 Chr.5: 14693584-14693584 on Build GRCh38
rs143563486 Chr.5: 14693438-14693438 on Build GRCh38
rs886713106 Chr.5: 14693248-14693248 on Build GRCh38
rs542421560 Chr.5: 14693377-14693377 on Build GRCh38
rs1027891937 Chr.5: 14693229-14693229 on Build GRCh38
rs974803145 Chr.5: 14693307-14693307 on Build GRCh38
rs796129732 Chr.5: 14693430-14693430 on Build GRCh38
rs915144184 Chr.5: 14693438-14693438 on Build GRCh38
rs953917008 Chr.5: 14693385-14693385 on Build GRCh38
rs989750279 Chr.5: 14693394-14693394 on Build GRCh38
rs977718365 Chr.5: 14693455-14693455 on Build GRCh38
rs528027993 Chr.5: 14693361-14693361 on Build GRCh38
rs561269859 Chr.5: 14693393-14693393 on Build GRCh38
rs1005101585 Chr.5: 14693643-14693643 on Build GRCh38
rs987450711 Chr.5: 14693195-14693195 on Build GRCh38
rs541462203 Chr.5: 14693618-14693618 on Build GRCh38
rs891570884 Chr.5: 14693250-14693250 on Build GRCh38
rs936191469 Chr.5: 14693242-14693242 on Build GRCh38
rs939058336 Chr.5: 14693509-14693509 on Build GRCh38
rs894915455 Chr.5: 14693575-14693575 on Build GRCh38
rs898370855 Chr.5: 14693369-14693369 on Build GRCh38
rs936215744 Chr.5: 14693519-14693519 on Build GRCh38
rs760333252 Chr.5: 14693628-14693628 on Build GRCh38
rs930807670 Chr.5: 14693579-14693579 on Build GRCh38
rs927617222 Chr.5: 14693484-14693484 on Build GRCh38
rs1016852913 Chr.5: 14693358-14693358 on Build GRCh38
rs148351338 Chr.5: 14693197-14693197 on Build GRCh38
rs1054894705 Chr.5: 14693518-14693518 on Build GRCh38
rs573030948 Chr.5: 14693563-14693563 on Build GRCh38
rs192386213 Chr.5: 14693285-14693285 on Build GRCh38
rs1030098913 Chr.5: 14693481-14693481 on Build GRCh38
rs535656925 Chr.5: 14693585-14693585 on Build GRCh38
rs943646597 Chr.5: 14693212-14693212 on Build GRCh38
rs975014634 Chr.5: 14693233-14693233 on Build GRCh38
rs1025407110 Chr.5: 14693383-14693383 on Build GRCh38
rs25951 Chr.5: 14693371-14693371 on Build GRCh38

View Full Product Details