Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 14705358-14705903 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKH
Entrez Gene ID
56172
Gene Name
ANKH inorganic pyrophosphate transport regulator
Gene Aliases
ANK, CCAL2, CMDJ, CPPDD, HANK, MANK
UniGene
Hs.156727
Gene Chromosome Location
Chr.5: 14704800-14871778 on Build GRCh38
Transcripts
NM_054027.4 XM_017009644.1 XM_011514151.2

Gene Symbol
OTULIN
Entrez Gene ID
90268
Gene Name
OTU deubiquitinase with linear linkage specificity
Gene Aliases
FAM105B, GUM
UniGene
Hs.406335
Gene Chromosome Location
Chr.5: 14660794-14716552 on Build GRCh38
Transcripts
NM_054027.4 XM_017009644.1 XM_011514151.2

SNP Detail

SNP ID Chromosome Location
rs995090592 Chr.5: 14705670-14705670 on Build GRCh38
rs553005034 Chr.5: 14705480-14705480 on Build GRCh38
rs964482029 Chr.5: 14705768-14705768 on Build GRCh38
rs913125811 Chr.5: 14705829-14705829 on Build GRCh38
rs145549693 Chr.5: 14705429-14705429 on Build GRCh38
rs190396675 Chr.5: 14705535-14705535 on Build GRCh38
rs143143398 Chr.5: 14705679-14705679 on Build GRCh38
rs772887866 Chr.5: 14705412-14705412 on Build GRCh38
rs966871990 Chr.5: 14705873-14705873 on Build GRCh38
rs79739542 Chr.5: 14705738-14705738 on Build GRCh38
rs754212859 Chr.5: 14705858-14705858 on Build GRCh38
rs982417870 Chr.5: 14705457-14705457 on Build GRCh38
rs35934788 Chr.5: 14705813-14705814 on Build GRCh38
rs529421049 Chr.5: 14705644-14705644 on Build GRCh38
rs1012794359 Chr.5: 14705420-14705420 on Build GRCh38
rs997891213 Chr.5: 14705752-14705752 on Build GRCh38
rs757746663 Chr.5: 14705867-14705867 on Build GRCh38
rs1002644163 Chr.5: 14705630-14705630 on Build GRCh38
rs1055125011 Chr.5: 14705611-14705611 on Build GRCh38
rs140985943 Chr.5: 14705578-14705578 on Build GRCh38
rs974850836 Chr.5: 14705769-14705769 on Build GRCh38
rs921056952 Chr.5: 14705490-14705490 on Build GRCh38
rs183658975 Chr.5: 14705718-14705718 on Build GRCh38
rs1055579550 Chr.5: 14705879-14705879 on Build GRCh38
rs367603371 Chr.5: 14705750-14705750 on Build GRCh38
rs976486174 Chr.5: 14705482-14705482 on Build GRCh38
rs766189058 Chr.5: 14705565-14705565 on Build GRCh38
rs913073195 Chr.5: 14705577-14705577 on Build GRCh38
rs990025092 Chr.5: 14705834-14705834 on Build GRCh38
rs988699639 Chr.5: 14705822-14705822 on Build GRCh38
rs747544921 Chr.5: 14705491-14705491 on Build GRCh38
rs925641099 Chr.5: 14705776-14705776 on Build GRCh38
rs26307 Chr.5: 14705556-14705556 on Build GRCh38
rs943049364 Chr.5: 14705471-14705471 on Build GRCh38
rs188487726 Chr.5: 14705806-14705806 on Build GRCh38
rs1055124794 Chr.5: 14705636-14705636 on Build GRCh38
rs139038211 Chr.5: 14705711-14705712 on Build GRCh38
rs868333155 Chr.5: 14705710-14705710 on Build GRCh38
rs987227757 Chr.5: 14705426-14705426 on Build GRCh38
rs561871600 Chr.5: 14705389-14705389 on Build GRCh38
rs764392423 Chr.5: 14705797-14705797 on Build GRCh38
rs758843802 Chr.5: 14705413-14705413 on Build GRCh38
rs116362523 Chr.5: 14705438-14705438 on Build GRCh38
rs1045528669 Chr.5: 14705838-14705838 on Build GRCh38
rs950874844 Chr.5: 14705689-14705689 on Build GRCh38
rs1015288456 Chr.5: 14705459-14705459 on Build GRCh38
rs1046481539 Chr.5: 14705599-14705599 on Build GRCh38
rs568862998 Chr.5: 14705552-14705552 on Build GRCh38
rs939381160 Chr.5: 14705604-14705604 on Build GRCh38
rs1013132945 Chr.5: 14705676-14705676 on Build GRCh38
rs953828126 Chr.5: 14705810-14705810 on Build GRCh38
rs187007081 Chr.5: 14705472-14705472 on Build GRCh38
rs886060070 Chr.5: 14705604-14705604 on Build GRCh38
rs557456192 Chr.5: 14705737-14705737 on Build GRCh38
rs769747533 Chr.5: 14705407-14705407 on Build GRCh38
rs895237230 Chr.5: 14705663-14705663 on Build GRCh38
rs1052925740 Chr.5: 14705557-14705557 on Build GRCh38
rs557160307 Chr.5: 14705860-14705860 on Build GRCh38
rs527718949 Chr.5: 14705534-14705534 on Build GRCh38
rs1042345021 Chr.5: 14705568-14705568 on Build GRCh38
rs566229423 Chr.5: 14705696-14705696 on Build GRCh38
rs928071760 Chr.5: 14705872-14705872 on Build GRCh38
rs568320530 Chr.5: 14705609-14705609 on Build GRCh38
rs1028451825 Chr.5: 14705807-14705807 on Build GRCh38

View Full Product Details