Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 14709616-14710126 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKH
Entrez Gene ID
56172
Gene Name
ANKH inorganic pyrophosphate transport regulator
Gene Aliases
ANK, CCAL2, CMDJ, CPPDD, HANK, MANK
UniGene
Hs.156727
Gene Chromosome Location
Chr.5: 14704800-14871778 on Build GRCh38
Transcripts
NM_054027.4 XM_017009644.1 XM_011514151.2

Gene Symbol
OTULIN
Entrez Gene ID
90268
Gene Name
OTU deubiquitinase with linear linkage specificity
Gene Aliases
FAM105B, GUM
UniGene
Hs.406335
Gene Chromosome Location
Chr.5: 14660794-14716552 on Build GRCh38
Transcripts
NM_054027.4 XM_017009644.1 XM_011514151.2

SNP Detail

SNP ID Chromosome Location
rs979495956 Chr.5: 14709683-14709683 on Build GRCh38
rs944637312 Chr.5: 14709790-14709790 on Build GRCh38
rs59784310 Chr.5: 14709925-14709925 on Build GRCh38
rs771287493 Chr.5: 14709846-14709846 on Build GRCh38
rs950491108 Chr.5: 14709739-14709739 on Build GRCh38
rs550094209 Chr.5: 14709959-14709962 on Build GRCh38
rs572028652 Chr.5: 14709788-14709789 on Build GRCh38
rs955791722 Chr.5: 14709880-14709880 on Build GRCh38
rs529069059 Chr.5: 14709897-14709897 on Build GRCh38
rs572785716 Chr.5: 14709774-14709774 on Build GRCh38
rs896380779 Chr.5: 14709770-14709770 on Build GRCh38
rs533669123 Chr.5: 14709953-14709953 on Build GRCh38
rs746470299 Chr.5: 14709756-14709756 on Build GRCh38
rs571146745 Chr.5: 14709755-14709755 on Build GRCh38
rs935458214 Chr.5: 14710003-14710003 on Build GRCh38
rs564201800 Chr.5: 14710087-14710087 on Build GRCh38
rs935158554 Chr.5: 14709706-14709706 on Build GRCh38
rs558959596 Chr.5: 14709686-14709686 on Build GRCh38
rs372796134 Chr.5: 14710055-14710055 on Build GRCh38
rs191464917 Chr.5: 14709827-14709827 on Build GRCh38
rs1034641791 Chr.5: 14709891-14709891 on Build GRCh38
rs917775927 Chr.5: 14710049-14710049 on Build GRCh38
rs1044728417 Chr.5: 14709775-14709775 on Build GRCh38
rs540164481 Chr.5: 14709793-14709793 on Build GRCh38
rs935636334 Chr.5: 14710041-14710041 on Build GRCh38
rs138050705 Chr.5: 14710005-14710005 on Build GRCh38
rs975056712 Chr.5: 14710008-14710008 on Build GRCh38
rs886060080 Chr.5: 14709843-14709843 on Build GRCh38
rs779663715 Chr.5: 14709742-14709742 on Build GRCh38
rs907357224 Chr.5: 14710096-14710096 on Build GRCh38
rs376296536 Chr.5: 14710050-14710050 on Build GRCh38
rs550363280 Chr.5: 14710092-14710092 on Build GRCh38
rs886060081 Chr.5: 14709872-14709875 on Build GRCh38
rs925362846 Chr.5: 14709978-14709978 on Build GRCh38
rs10570 Chr.5: 14709997-14709997 on Build GRCh38
rs921891628 Chr.5: 14710025-14710025 on Build GRCh38
rs1018198398 Chr.5: 14709919-14709919 on Build GRCh38
rs962733675 Chr.5: 14709676-14709676 on Build GRCh38
rs182918257 Chr.5: 14709859-14709859 on Build GRCh38
rs886060082 Chr.5: 14709986-14709986 on Build GRCh38
rs1054095058 Chr.5: 14709825-14709825 on Build GRCh38
rs773388808 Chr.5: 14709766-14709770 on Build GRCh38
rs988360785 Chr.5: 14710015-14710015 on Build GRCh38
rs1045009753 Chr.5: 14710083-14710083 on Build GRCh38
rs758220812 Chr.5: 14709738-14709738 on Build GRCh38
rs951455159 Chr.5: 14709958-14709958 on Build GRCh38
rs75992532 Chr.5: 14709814-14709814 on Build GRCh38
rs776755741 Chr.5: 14709707-14709707 on Build GRCh38
rs911693760 Chr.5: 14709985-14709985 on Build GRCh38
rs945770575 Chr.5: 14710081-14710081 on Build GRCh38
rs747738518 Chr.5: 14709845-14709845 on Build GRCh38
rs192691329 Chr.5: 14709975-14709975 on Build GRCh38
rs1005939556 Chr.5: 14709792-14709792 on Build GRCh38
rs771338338 Chr.5: 14709814-14709818 on Build GRCh38
rs78290648 Chr.5: 14709710-14709710 on Build GRCh38
rs918713028 Chr.5: 14709702-14709702 on Build GRCh38
rs1002181297 Chr.5: 14709883-14709883 on Build GRCh38
rs187733448 Chr.5: 14709927-14709927 on Build GRCh38
rs25990 Chr.5: 14709926-14709926 on Build GRCh38
rs1025838224 Chr.5: 14709864-14709864 on Build GRCh38
rs1010377250 Chr.5: 14709836-14709836 on Build GRCh38
rs912962154 Chr.5: 14710078-14710078 on Build GRCh38
rs935701557 Chr.5: 14709821-14709821 on Build GRCh38
rs754691127 Chr.5: 14709771-14709771 on Build GRCh38
rs759984300 Chr.5: 14709847-14709848 on Build GRCh38
rs576801623 Chr.5: 14709813-14709813 on Build GRCh38
rs748218900 Chr.5: 14709687-14709688 on Build GRCh38

View Full Product Details