Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 15500106-15500594 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXL7
Entrez Gene ID
23194
Gene Name
F-box and leucine rich repeat protein 7
Gene Aliases
FBL6, FBL7
UniGene
Hs.433057
Gene Chromosome Location
Chr.5: 15500196-15939796 on Build GRCh38
Transcripts
NM_012304.4

SNP Detail

SNP ID Chromosome Location
rs886284424 Chr.5: 15500177-15500177 on Build GRCh38
rs902407748 Chr.5: 15500391-15500391 on Build GRCh38
rs1037082752 Chr.5: 15500338-15500338 on Build GRCh38
rs908124019 Chr.5: 15500306-15500306 on Build GRCh38
rs560658914 Chr.5: 15500540-15500540 on Build GRCh38
rs377379955 Chr.5: 15500140-15500140 on Build GRCh38
rs978333270 Chr.5: 15500146-15500146 on Build GRCh38
rs370614214 Chr.5: 15500330-15500330 on Build GRCh38
rs370307411 Chr.5: 15500465-15500465 on Build GRCh38
rs962580230 Chr.5: 15500467-15500467 on Build GRCh38
rs569013257 Chr.5: 15500193-15500193 on Build GRCh38
rs952161094 Chr.5: 15500426-15500426 on Build GRCh38
rs540091616 Chr.5: 15500561-15500562 on Build GRCh38
rs1012699898 Chr.5: 15500423-15500423 on Build GRCh38
rs112289589 Chr.5: 15500198-15500198 on Build GRCh38
rs940564534 Chr.5: 15500183-15500183 on Build GRCh38
rs966644887 Chr.5: 15500202-15500202 on Build GRCh38
rs549492404 Chr.5: 15500214-15500214 on Build GRCh38
rs550537153 Chr.5: 15500181-15500181 on Build GRCh38
rs542654103 Chr.5: 15500548-15500548 on Build GRCh38
rs910955028 Chr.5: 15500560-15500560 on Build GRCh38
rs575790249 Chr.5: 15500468-15500468 on Build GRCh38
rs928241649 Chr.5: 15500150-15500150 on Build GRCh38
rs139767597 Chr.5: 15500228-15500228 on Build GRCh38
rs542353119 Chr.5: 15500505-15500505 on Build GRCh38
rs1023668720 Chr.5: 15500424-15500424 on Build GRCh38
rs978479720 Chr.5: 15500210-15500210 on Build GRCh38
rs546148027 Chr.5: 15500151-15500151 on Build GRCh38
rs960740268 Chr.5: 15500422-15500422 on Build GRCh38
rs372983518 Chr.5: 15500154-15500154 on Build GRCh38
rs976369733 Chr.5: 15500537-15500537 on Build GRCh38
rs535016943 Chr.5: 15500234-15500234 on Build GRCh38
rs768896296 Chr.5: 15500175-15500175 on Build GRCh38
rs372622055 Chr.5: 15500415-15500415 on Build GRCh38
rs1044142810 Chr.5: 15500352-15500352 on Build GRCh38
rs997965163 Chr.5: 15500173-15500173 on Build GRCh38
rs1010730886 Chr.5: 15500386-15500386 on Build GRCh38
rs886733222 Chr.5: 15500160-15500160 on Build GRCh38
rs578197287 Chr.5: 15500318-15500318 on Build GRCh38
rs1049496926 Chr.5: 15500372-15500372 on Build GRCh38
rs922223097 Chr.5: 15500541-15500541 on Build GRCh38
rs889843678 Chr.5: 15500563-15500563 on Build GRCh38
rs930811847 Chr.5: 15500262-15500262 on Build GRCh38
rs1040303459 Chr.5: 15500527-15500527 on Build GRCh38
rs1035396561 Chr.5: 15500400-15500400 on Build GRCh38
rs553373000 Chr.5: 15500280-15500280 on Build GRCh38
rs916707132 Chr.5: 15500235-15500235 on Build GRCh38
rs539858244 Chr.5: 15500569-15500569 on Build GRCh38
rs572576226 Chr.5: 15500564-15500564 on Build GRCh38
rs1028746814 Chr.5: 15500554-15500554 on Build GRCh38
rs999460270 Chr.5: 15500398-15500398 on Build GRCh38
rs900849945 Chr.5: 15500361-15500361 on Build GRCh38
rs916330183 Chr.5: 15500155-15500155 on Build GRCh38
rs984908753 Chr.5: 15500427-15500427 on Build GRCh38
rs1043549954 Chr.5: 15500562-15500562 on Build GRCh38
rs984959588 Chr.5: 15500265-15500265 on Build GRCh38
rs1022161655 Chr.5: 15500388-15500388 on Build GRCh38
rs532402258 Chr.5: 15500170-15500170 on Build GRCh38
rs966906187 Chr.5: 15500145-15500145 on Build GRCh38
rs933702357 Chr.5: 15500371-15500371 on Build GRCh38
rs1022396333 Chr.5: 15500572-15500572 on Build GRCh38
rs966832791 Chr.5: 15500576-15500576 on Build GRCh38
rs940972790 Chr.5: 15500317-15500317 on Build GRCh38
rs375952615 Chr.5: 15500162-15500162 on Build GRCh38
rs930860792 Chr.5: 15500148-15500148 on Build GRCh38
rs528066589 Chr.5: 15500132-15500132 on Build GRCh38
rs889573351 Chr.5: 15500375-15500375 on Build GRCh38

View Full Product Details