Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 26890348-26890862 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDH9
Entrez Gene ID
1007
Gene Name
cadherin 9
Gene Aliases
-
UniGene
Hs.272212
Gene Chromosome Location
Chr.5: 26880600-27038582 on Build GRCh38
Transcripts
NM_016279.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
449509 c.1389A>G p.I463M Chr.5: 26890429-26890429 on Build GRCh38
6485037 c.1342C>A p.R448R Chr.5: 26890476-26890476 on Build GRCh38
5455392 c.1355C>A p.P452H Chr.5: 26890463-26890463 on Build GRCh38
1436949 c.1343G>A p.R448Q Chr.5: 26890475-26890475 on Build GRCh38
6546104 c.1284C>A p.D428E Chr.5: 26890534-26890534 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs534822627 Chr.5: 26890753-26890753 on Build GRCh38
rs777158238 Chr.5: 26890429-26890429 on Build GRCh38
rs989987613 Chr.5: 26890587-26890587 on Build GRCh38
rs773321321 Chr.5: 26890614-26890614 on Build GRCh38
rs772484794 Chr.5: 26890449-26890449 on Build GRCh38
rs977493505 Chr.5: 26890836-26890836 on Build GRCh38
rs957387667 Chr.5: 26890553-26890553 on Build GRCh38
rs867091469 Chr.5: 26890465-26890465 on Build GRCh38
rs867376080 Chr.5: 26890518-26890518 on Build GRCh38
rs761070646 Chr.5: 26890456-26890456 on Build GRCh38
rs555002432 Chr.5: 26890550-26890550 on Build GRCh38
rs967093278 Chr.5: 26890831-26890831 on Build GRCh38
rs747656352 Chr.5: 26890414-26890414 on Build GRCh38
rs189637992 Chr.5: 26890646-26890646 on Build GRCh38
rs769934258 Chr.5: 26890605-26890605 on Build GRCh38
rs1034969550 Chr.5: 26890751-26890751 on Build GRCh38
rs777294478 Chr.5: 26890543-26890543 on Build GRCh38
rs772604317 Chr.5: 26890548-26890548 on Build GRCh38
rs898012490 Chr.5: 26890762-26890762 on Build GRCh38
rs201441998 Chr.5: 26890432-26890432 on Build GRCh38
rs775903704 Chr.5: 26890452-26890452 on Build GRCh38
rs780978039 Chr.5: 26890532-26890532 on Build GRCh38
rs907830063 Chr.5: 26890535-26890536 on Build GRCh38
rs898558124 Chr.5: 26890686-26890686 on Build GRCh38
rs761984153 Chr.5: 26890595-26890595 on Build GRCh38
rs776900416 Chr.5: 26890458-26890458 on Build GRCh38
rs1013804803 Chr.5: 26890785-26890785 on Build GRCh38
rs1046690150 Chr.5: 26890814-26890814 on Build GRCh38
rs766274426 Chr.5: 26890483-26890483 on Build GRCh38
rs745823439 Chr.5: 26890406-26890407 on Build GRCh38
rs967246029 Chr.5: 26890612-26890612 on Build GRCh38
rs768954953 Chr.5: 26890551-26890551 on Build GRCh38
rs971053594 Chr.5: 26890477-26890477 on Build GRCh38
rs751604509 Chr.5: 26890635-26890635 on Build GRCh38
rs769483643 Chr.5: 26890424-26890424 on Build GRCh38
rs142357370 Chr.5: 26890484-26890484 on Build GRCh38
rs758273778 Chr.5: 26890398-26890398 on Build GRCh38
rs751366845 Chr.5: 26890523-26890523 on Build GRCh38
rs531407008 Chr.5: 26890666-26890666 on Build GRCh38
rs746317937 Chr.5: 26890446-26890446 on Build GRCh38
rs1050897854 Chr.5: 26890676-26890676 on Build GRCh38
rs373897251 Chr.5: 26890391-26890391 on Build GRCh38
rs942136743 Chr.5: 26890737-26890737 on Build GRCh38
rs781029442 Chr.5: 26890411-26890411 on Build GRCh38
rs368339950 Chr.5: 26890404-26890404 on Build GRCh38
rs148731332 Chr.5: 26890476-26890476 on Build GRCh38
rs1006389749 Chr.5: 26890389-26890389 on Build GRCh38
rs967021056 Chr.5: 26890581-26890581 on Build GRCh38
rs754653524 Chr.5: 26890407-26890407 on Build GRCh38
rs182661360 Chr.5: 26890759-26890759 on Build GRCh38
rs866075314 Chr.5: 26890533-26890533 on Build GRCh38
rs994147740 Chr.5: 26890695-26890695 on Build GRCh38
rs775554905 Chr.5: 26890631-26890632 on Build GRCh38
rs759556480 Chr.5: 26890639-26890639 on Build GRCh38
rs754886766 Chr.5: 26890530-26890530 on Build GRCh38
rs765294287 Chr.5: 26890468-26890468 on Build GRCh38
rs112654898 Chr.5: 26890637-26890637 on Build GRCh38
rs756785120 Chr.5: 26890603-26890603 on Build GRCh38
rs914263625 Chr.5: 26890562-26890562 on Build GRCh38
rs751026716 Chr.5: 26890457-26890457 on Build GRCh38
rs766334351 Chr.5: 26890634-26890634 on Build GRCh38
rs776986066 Chr.5: 26890568-26890568 on Build GRCh38
rs562858081 Chr.5: 26890710-26890710 on Build GRCh38
rs982020288 Chr.5: 26890448-26890448 on Build GRCh38
rs372072724 Chr.5: 26890405-26890405 on Build GRCh38
rs775122116 Chr.5: 26890396-26890396 on Build GRCh38
rs762037380 Chr.5: 26890463-26890463 on Build GRCh38
rs909277907 Chr.5: 26890723-26890723 on Build GRCh38
rs533639381 Chr.5: 26890481-26890481 on Build GRCh38
rs755827991 Chr.5: 26890538-26890538 on Build GRCh38
rs1034095488 Chr.5: 26890535-26890535 on Build GRCh38
rs1047020105 Chr.5: 26890696-26890696 on Build GRCh38
rs762981423 Chr.5: 26890627-26890627 on Build GRCh38
rs980224696 Chr.5: 26890500-26890500 on Build GRCh38
rs771846324 Chr.5: 26890539-26890539 on Build GRCh38

View Full Product Details