Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 34042683-34043178 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1QTNF3
Entrez Gene ID
114899
Gene Name
C1q and tumor necrosis factor related protein 3
Gene Aliases
C1ATNF3, CORCS, CORS, CORS-26, CORS26, CTRP3
UniGene
Hs.171929
Gene Chromosome Location
Chr.5: 34017858-34043266 on Build GRCh38
Transcripts
NM_030945.3 NM_181435.5 NR_037951.1

Gene Symbol
C1QTNF3-AMACR
Entrez Gene ID
100534612
Gene Name
C1QTNF3-AMACR readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.171929
Gene Chromosome Location
Chr.5: 33986986-34124528 on Build GRCh38
Transcripts
NM_030945.3 NM_181435.5 NR_037951.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1620247 c.62T>A p.L21Q Chr.5: 34043064-34043064 on Build GRCh38
1578548 c.195_196delTG p.G66fs*4 Chr.5: 34042930-34042931 on Build GRCh38
3615542 c.261C>T p.P87P Chr.5: 34042865-34042865 on Build GRCh38
3827954 c.159G>A p.R53R Chr.5: 34042967-34042967 on Build GRCh38
6484130 c.111G>C p.V37V Chr.5: 34043015-34043015 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs984690475 Chr.5: 34042751-34042753 on Build GRCh38
rs1042538186 Chr.5: 34042915-34042915 on Build GRCh38
rs769863322 Chr.5: 34042849-34042849 on Build GRCh38
rs769355920 Chr.5: 34042992-34042992 on Build GRCh38
rs769321574 Chr.5: 34043092-34043092 on Build GRCh38
rs754006287 Chr.5: 34042949-34042949 on Build GRCh38
rs756327553 Chr.5: 34042868-34042868 on Build GRCh38
rs143587337 Chr.5: 34042976-34042976 on Build GRCh38
rs775986621 Chr.5: 34043130-34043130 on Build GRCh38
rs761099252 Chr.5: 34043134-34043134 on Build GRCh38
rs747745802 Chr.5: 34042985-34042985 on Build GRCh38
rs113668504 Chr.5: 34042870-34042870 on Build GRCh38
rs755392798 Chr.5: 34042841-34042841 on Build GRCh38
rs1023613583 Chr.5: 34043041-34043041 on Build GRCh38
rs780118939 Chr.5: 34042905-34042905 on Build GRCh38
rs933041044 Chr.5: 34042924-34042924 on Build GRCh38
rs376644937 Chr.5: 34042980-34042980 on Build GRCh38
rs764509132 Chr.5: 34043136-34043136 on Build GRCh38
rs373831651 Chr.5: 34043020-34043020 on Build GRCh38
rs376405283 Chr.5: 34043048-34043048 on Build GRCh38
rs112167551 Chr.5: 34042761-34042762 on Build GRCh38
rs141837290 Chr.5: 34042939-34042939 on Build GRCh38
rs754835283 Chr.5: 34043075-34043075 on Build GRCh38
rs762830778 Chr.5: 34043052-34043052 on Build GRCh38
rs752992491 Chr.5: 34042933-34042933 on Build GRCh38
rs758481865 Chr.5: 34042850-34042850 on Build GRCh38
rs144445678 Chr.5: 34042871-34042871 on Build GRCh38
rs759897017 Chr.5: 34042926-34042926 on Build GRCh38
rs752124283 Chr.5: 34042918-34042918 on Build GRCh38
rs72343197 Chr.5: 34042772-34042773 on Build GRCh38
rs748777817 Chr.5: 34043118-34043118 on Build GRCh38
rs752972223 Chr.5: 34042865-34042865 on Build GRCh38
rs147817904 Chr.5: 34043084-34043084 on Build GRCh38
rs750607069 Chr.5: 34042836-34042838 on Build GRCh38
rs386403521 Chr.5: 34042775-34042776 on Build GRCh38
rs531769358 Chr.5: 34042872-34042872 on Build GRCh38
rs368754050 Chr.5: 34043010-34043010 on Build GRCh38
rs760178805 Chr.5: 34042794-34042794 on Build GRCh38
rs751575080 Chr.5: 34042941-34042943 on Build GRCh38
rs188876397 Chr.5: 34042901-34042901 on Build GRCh38
rs745730855 Chr.5: 34042883-34042883 on Build GRCh38
rs73761687 Chr.5: 34042761-34042761 on Build GRCh38
rs761440829 Chr.5: 34042907-34042907 on Build GRCh38
rs369846327 Chr.5: 34042800-34042800 on Build GRCh38
rs765416002 Chr.5: 34043039-34043039 on Build GRCh38
rs763605813 Chr.5: 34042819-34042819 on Build GRCh38
rs190492922 Chr.5: 34042826-34042826 on Build GRCh38
rs767994826 Chr.5: 34042928-34042928 on Build GRCh38
rs201504297 Chr.5: 34042947-34042947 on Build GRCh38
rs981292320 Chr.5: 34042795-34042795 on Build GRCh38
rs573985404 Chr.5: 34042764-34042765 on Build GRCh38
rs762143540 Chr.5: 34043141-34043141 on Build GRCh38
rs771905676 Chr.5: 34042890-34042890 on Build GRCh38
rs199873478 Chr.5: 34042895-34042895 on Build GRCh38
rs780020256 Chr.5: 34042714-34042714 on Build GRCh38
rs1051912707 Chr.5: 34042931-34042931 on Build GRCh38
rs6859593 Chr.5: 34043140-34043140 on Build GRCh38
rs753383673 Chr.5: 34042803-34042803 on Build GRCh38
rs371214613 Chr.5: 34043074-34043074 on Build GRCh38
rs57826552 Chr.5: 34042777-34042778 on Build GRCh38
rs772597219 Chr.5: 34042993-34042993 on Build GRCh38
rs781072048 Chr.5: 34043078-34043078 on Build GRCh38
rs538468468 Chr.5: 34042793-34042794 on Build GRCh38
rs768533163 Chr.5: 34042981-34042981 on Build GRCh38
rs750447263 Chr.5: 34043047-34043047 on Build GRCh38
rs745884570 Chr.5: 34042974-34042974 on Build GRCh38
rs900266305 Chr.5: 34042891-34042891 on Build GRCh38
rs952056918 Chr.5: 34042745-34042745 on Build GRCh38
rs775933704 Chr.5: 34043015-34043015 on Build GRCh38
rs555686985 Chr.5: 34042762-34042764 on Build GRCh38
rs113809897 Chr.5: 34042760-34042760 on Build GRCh38
rs544070128 Chr.5: 34043036-34043036 on Build GRCh38
rs73761686 Chr.5: 34042759-34042759 on Build GRCh38
rs971247603 Chr.5: 34043088-34043088 on Build GRCh38
rs1005132863 Chr.5: 34042961-34042961 on Build GRCh38
rs567065604 Chr.5: 34043014-34043014 on Build GRCh38
rs760151163 Chr.5: 34042996-34042996 on Build GRCh38
rs776323791 Chr.5: 34042906-34042906 on Build GRCh38
rs775222105 Chr.5: 34042775-34042775 on Build GRCh38
rs755006748 Chr.5: 34043106-34043106 on Build GRCh38
rs770423787 Chr.5: 34043122-34043122 on Build GRCh38
rs72108923 Chr.5: 34042762-34042763 on Build GRCh38
rs776445792 Chr.5: 34042982-34042982 on Build GRCh38
rs779889392 Chr.5: 34043073-34043073 on Build GRCh38
rs765344573 Chr.5: 34043145-34043145 on Build GRCh38
rs768399020 Chr.5: 34042903-34042903 on Build GRCh38
rs757338339 Chr.5: 34042967-34042967 on Build GRCh38
rs199553003 Chr.5: 34043035-34043035 on Build GRCh38
rs772829497 Chr.5: 34043095-34043095 on Build GRCh38
rs990954680 Chr.5: 34042713-34042713 on Build GRCh38
rs200960868 Chr.5: 34042975-34042975 on Build GRCh38
rs764791785 Chr.5: 34042908-34042908 on Build GRCh38
rs751972062 Chr.5: 34042828-34042828 on Build GRCh38

View Full Product Details