Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 37813540-37814045 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GDNF
Entrez Gene ID
2668
Gene Name
glial cell derived neurotrophic factor
Gene Aliases
ATF1, ATF2, HFB1-GDNF, HSCR3
UniGene
Hs.248114
Gene Chromosome Location
Chr.5: 37812677-37840044 on Build GRCh38
Transcripts
NM_000514.3 NM_001190468.1 NM_001190469.1 NM_001278098.1 NM_199231.2

SNP Detail

SNP ID Chromosome Location
rs76445038 Chr.5: 37813580-37813580 on Build GRCh38
rs996403481 Chr.5: 37813838-37813838 on Build GRCh38
rs886466471 Chr.5: 37813796-37813796 on Build GRCh38
rs890459008 Chr.5: 37813808-37813808 on Build GRCh38
rs916052708 Chr.5: 37813651-37813651 on Build GRCh38
rs998616288 Chr.5: 37813873-37813873 on Build GRCh38
rs79582724 Chr.5: 37813584-37813584 on Build GRCh38
rs979962420 Chr.5: 37813867-37813867 on Build GRCh38
rs200330062 Chr.5: 37813952-37813952 on Build GRCh38
rs1005125379 Chr.5: 37813929-37813929 on Build GRCh38
rs1037895741 Chr.5: 37813705-37813705 on Build GRCh38
rs1035741263 Chr.5: 37813875-37813875 on Build GRCh38
rs199775384 Chr.5: 37813864-37813864 on Build GRCh38
rs886060590 Chr.5: 37813592-37813592 on Build GRCh38
rs541881147 Chr.5: 37813995-37813995 on Build GRCh38
rs991180936 Chr.5: 37813938-37813938 on Build GRCh38
rs934306077 Chr.5: 37813659-37813659 on Build GRCh38
rs869091796 Chr.5: 37813583-37813584 on Build GRCh38
rs149012858 Chr.5: 37813860-37813860 on Build GRCh38
rs115322727 Chr.5: 37813866-37813866 on Build GRCh38
rs996844389 Chr.5: 37813996-37813996 on Build GRCh38
rs530864008 Chr.5: 37813637-37813637 on Build GRCh38
rs548428619 Chr.5: 37813852-37813852 on Build GRCh38
rs916884433 Chr.5: 37813863-37813863 on Build GRCh38
rs899791064 Chr.5: 37813849-37813849 on Build GRCh38
rs886060591 Chr.5: 37813724-37813724 on Build GRCh38
rs145848373 Chr.5: 37813850-37813850 on Build GRCh38
rs1057070384 Chr.5: 37813693-37813693 on Build GRCh38
rs1008078517 Chr.5: 37813833-37813833 on Build GRCh38
rs1022490029 Chr.5: 37813840-37813840 on Build GRCh38
rs898982193 Chr.5: 37813738-37813738 on Build GRCh38
rs188617599 Chr.5: 37813958-37813958 on Build GRCh38
rs924286132 Chr.5: 37813642-37813642 on Build GRCh38
rs538387510 Chr.5: 37813889-37813889 on Build GRCh38
rs904043405 Chr.5: 37814023-37814023 on Build GRCh38
rs374825523 Chr.5: 37813961-37813961 on Build GRCh38
rs370174553 Chr.5: 37813650-37813650 on Build GRCh38
rs556575483 Chr.5: 37813900-37813900 on Build GRCh38
rs977482220 Chr.5: 37813622-37813622 on Build GRCh38
rs750653484 Chr.5: 37813856-37813856 on Build GRCh38
rs765390997 Chr.5: 37813721-37813721 on Build GRCh38
rs886060592 Chr.5: 37813863-37813863 on Build GRCh38
rs577405600 Chr.5: 37813888-37813888 on Build GRCh38
rs916146097 Chr.5: 37813941-37813941 on Build GRCh38
rs942162199 Chr.5: 37813658-37813658 on Build GRCh38
rs781582573 Chr.5: 37814018-37814018 on Build GRCh38
rs4869550 Chr.5: 37813597-37813597 on Build GRCh38
rs147083416 Chr.5: 37813869-37813869 on Build GRCh38
rs886060589 Chr.5: 37813584-37813584 on Build GRCh38
rs886060593 Chr.5: 37813938-37813938 on Build GRCh38
rs4869551 Chr.5: 37813610-37813610 on Build GRCh38
rs878873257 Chr.5: 37813694-37813694 on Build GRCh38
rs1051878013 Chr.5: 37814003-37814003 on Build GRCh38
rs149881166 Chr.5: 37813657-37813657 on Build GRCh38
rs1015617127 Chr.5: 37813943-37813943 on Build GRCh38
rs928326839 Chr.5: 37813624-37813624 on Build GRCh38
rs1021780779 Chr.5: 37813592-37813592 on Build GRCh38
rs886060594 Chr.5: 37813994-37813994 on Build GRCh38
rs1039612423 Chr.5: 37813845-37813845 on Build GRCh38
rs144538407 Chr.5: 37813926-37813927 on Build GRCh38
rs1040778372 Chr.5: 37813969-37813969 on Build GRCh38
rs963555970 Chr.5: 37813954-37813954 on Build GRCh38
rs965338577 Chr.5: 37813828-37813828 on Build GRCh38
rs924336732 Chr.5: 37813846-37813846 on Build GRCh38
rs960237435 Chr.5: 37813626-37813626 on Build GRCh38
rs929442590 Chr.5: 37813724-37813724 on Build GRCh38
rs547628776 Chr.5: 37813753-37813753 on Build GRCh38
rs11111 Chr.5: 37814000-37814000 on Build GRCh38
rs568543354 Chr.5: 37813788-37813788 on Build GRCh38
rs900476741 Chr.5: 37813976-37813976 on Build GRCh38
rs748498805 Chr.5: 37814024-37814024 on Build GRCh38
rs919442110 Chr.5: 37813962-37813962 on Build GRCh38

View Full Product Details