Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 37815134-37815641 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GDNF
Entrez Gene ID
2668
Gene Name
glial cell derived neurotrophic factor
Gene Aliases
ATF1, ATF2, HFB1-GDNF, HSCR3
UniGene
Hs.248114
Gene Chromosome Location
Chr.5: 37812677-37840044 on Build GRCh38
Transcripts
NM_000514.3 NM_001190468.1 NM_001190469.1 NM_001278098.1 NM_199231.2 XM_011514030.2 XM_017009337.1

SNP Detail

SNP ID Chromosome Location
rs138074154 Chr.5: 37815388-37815388 on Build GRCh38
rs376098758 Chr.5: 37815541-37815541 on Build GRCh38
rs997471865 Chr.5: 37815313-37815313 on Build GRCh38
rs988610131 Chr.5: 37815404-37815404 on Build GRCh38
rs926872946 Chr.5: 37815515-37815523 on Build GRCh38
rs1036030610 Chr.5: 37815304-37815304 on Build GRCh38
rs992189832 Chr.5: 37815191-37815191 on Build GRCh38
rs558003481 Chr.5: 37815312-37815312 on Build GRCh38
rs931512760 Chr.5: 37815519-37815519 on Build GRCh38
rs767561340 Chr.5: 37815299-37815299 on Build GRCh38
rs932243767 Chr.5: 37815308-37815308 on Build GRCh38
rs141395105 Chr.5: 37815201-37815201 on Build GRCh38
rs771093670 Chr.5: 37815215-37815215 on Build GRCh38
rs981068338 Chr.5: 37815440-37815440 on Build GRCh38
rs868643238 Chr.5: 37815520-37815520 on Build GRCh38
rs1028896890 Chr.5: 37815326-37815327 on Build GRCh38
rs45535335 Chr.5: 37815558-37815558 on Build GRCh38
rs45611434 Chr.5: 37815288-37815288 on Build GRCh38
rs979925306 Chr.5: 37815232-37815232 on Build GRCh38
rs888991758 Chr.5: 37815366-37815366 on Build GRCh38
rs1047945291 Chr.5: 37815293-37815293 on Build GRCh38
rs1013591730 Chr.5: 37815447-37815447 on Build GRCh38
rs896597971 Chr.5: 37815435-37815435 on Build GRCh38
rs949681730 Chr.5: 37815175-37815175 on Build GRCh38
rs150577324 Chr.5: 37815488-37815490 on Build GRCh38
rs1040055321 Chr.5: 37815302-37815302 on Build GRCh38
rs1043861797 Chr.5: 37815353-37815353 on Build GRCh38
rs567404383 Chr.5: 37815210-37815210 on Build GRCh38
rs1036491605 Chr.5: 37815570-37815570 on Build GRCh38
rs13178132 Chr.5: 37815526-37815526 on Build GRCh38
rs951956149 Chr.5: 37815573-37815573 on Build GRCh38
rs926820676 Chr.5: 37815194-37815194 on Build GRCh38
rs1024989592 Chr.5: 37815421-37815421 on Build GRCh38
rs944357950 Chr.5: 37815300-37815300 on Build GRCh38
rs775849560 Chr.5: 37815615-37815615 on Build GRCh38
rs1000692685 Chr.5: 37815373-37815373 on Build GRCh38
rs970004881 Chr.5: 37815217-37815217 on Build GRCh38
rs565976844 Chr.5: 37815237-37815237 on Build GRCh38
rs11952257 Chr.5: 37815523-37815523 on Build GRCh38
rs11952258 Chr.5: 37815529-37815529 on Build GRCh38
rs1023676494 Chr.5: 37815451-37815451 on Build GRCh38
rs983325701 Chr.5: 37815582-37815582 on Build GRCh38
rs886060603 Chr.5: 37815518-37815523 on Build GRCh38
rs981400561 Chr.5: 37815190-37815190 on Build GRCh38
rs1030979770 Chr.5: 37815179-37815179 on Build GRCh38
rs576329269 Chr.5: 37815355-37815355 on Build GRCh38
rs759593973 Chr.5: 37815295-37815295 on Build GRCh38
rs371311257 Chr.5: 37815301-37815301 on Build GRCh38
rs560077980 Chr.5: 37815549-37815549 on Build GRCh38
rs960709756 Chr.5: 37815186-37815186 on Build GRCh38
rs753233921 Chr.5: 37815305-37815305 on Build GRCh38
rs907626843 Chr.5: 37815588-37815588 on Build GRCh38
rs975685452 Chr.5: 37815592-37815592 on Build GRCh38
rs904038862 Chr.5: 37815380-37815380 on Build GRCh38
rs770256327 Chr.5: 37815612-37815612 on Build GRCh38
rs574469451 Chr.5: 37815515-37815515 on Build GRCh38
rs764521758 Chr.5: 37815609-37815609 on Build GRCh38
rs984262092 Chr.5: 37815547-37815547 on Build GRCh38
rs1031490841 Chr.5: 37815420-37815420 on Build GRCh38
rs777231466 Chr.5: 37815488-37815493 on Build GRCh38
rs944493852 Chr.5: 37815589-37815589 on Build GRCh38
rs537204707 Chr.5: 37815378-37815378 on Build GRCh38
rs940399996 Chr.5: 37815294-37815294 on Build GRCh38

View Full Product Details