Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 39285137-39285637 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9
Entrez Gene ID
735
Gene Name
complement component 9
Gene Aliases
ARMD15, C9D
UniGene
Hs.654443
Gene Chromosome Location
Chr.5: 39284275-39364553 on Build GRCh38
Transcripts

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1695697 c.1675A>T p.K559* Chr.5: 39285204-39285204 on Build GRCh38
6616769 c.1656C>A p.A552A Chr.5: 39285223-39285223 on Build GRCh38
6513004 c.1650G>T p.L550F Chr.5: 39285229-39285229 on Build GRCh38
5403488 c.1665C>T p.F555F Chr.5: 39285214-39285214 on Build GRCh38
1068064 c.1671T>C p.N557N Chr.5: 39285208-39285208 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375839761 Chr.5: 39285242-39285242 on Build GRCh38
rs975541822 Chr.5: 39285297-39285297 on Build GRCh38
rs995283569 Chr.5: 39285520-39285520 on Build GRCh38
rs769962112 Chr.5: 39285413-39285413 on Build GRCh38
rs375687695 Chr.5: 39285387-39285387 on Build GRCh38
rs1003456005 Chr.5: 39285446-39285446 on Build GRCh38
rs947249432 Chr.5: 39285335-39285335 on Build GRCh38
rs964845357 Chr.5: 39285572-39285572 on Build GRCh38
rs987669767 Chr.5: 39285303-39285303 on Build GRCh38
rs55791193 Chr.5: 39285574-39285574 on Build GRCh38
rs565504592 Chr.5: 39285408-39285408 on Build GRCh38
rs773994692 Chr.5: 39285234-39285234 on Build GRCh38
rs1012975550 Chr.5: 39285500-39285500 on Build GRCh38
rs374757245 Chr.5: 39285541-39285541 on Build GRCh38
rs747455321 Chr.5: 39285301-39285301 on Build GRCh38
rs532117706 Chr.5: 39285354-39285354 on Build GRCh38
rs963474531 Chr.5: 39285511-39285511 on Build GRCh38
rs141988743 Chr.5: 39285214-39285214 on Build GRCh38
rs369783130 Chr.5: 39285225-39285225 on Build GRCh38
rs373112281 Chr.5: 39285181-39285181 on Build GRCh38
rs564637052 Chr.5: 39285340-39285340 on Build GRCh38
rs762159491 Chr.5: 39285202-39285202 on Build GRCh38
rs1027158520 Chr.5: 39285532-39285532 on Build GRCh38
rs973695404 Chr.5: 39285291-39285291 on Build GRCh38
rs1042039059 Chr.5: 39285364-39285364 on Build GRCh38
rs780317875 Chr.5: 39285174-39285174 on Build GRCh38
rs912081931 Chr.5: 39285545-39285545 on Build GRCh38
rs548616913 Chr.5: 39285501-39285501 on Build GRCh38
rs376433843 Chr.5: 39285179-39285179 on Build GRCh38
rs1034826418 Chr.5: 39285461-39285461 on Build GRCh38
rs530392152 Chr.5: 39285498-39285498 on Build GRCh38
rs768195604 Chr.5: 39285224-39285224 on Build GRCh38
rs887239586 Chr.5: 39285551-39285551 on Build GRCh38
rs777986322 Chr.5: 39285195-39285195 on Build GRCh38
rs760746886 Chr.5: 39285276-39285276 on Build GRCh38
rs141471456 Chr.5: 39285236-39285236 on Build GRCh38
rs191338806 Chr.5: 39285530-39285530 on Build GRCh38
rs771406248 Chr.5: 39285208-39285208 on Build GRCh38
rs776807144 Chr.5: 39285348-39285348 on Build GRCh38
rs750215685 Chr.5: 39285272-39285272 on Build GRCh38
rs773290245 Chr.5: 39285463-39285463 on Build GRCh38
rs756112365 Chr.5: 39285475-39285475 on Build GRCh38
rs188200607 Chr.5: 39285508-39285508 on Build GRCh38
rs747555176 Chr.5: 39285203-39285203 on Build GRCh38
rs369297852 Chr.5: 39285271-39285271 on Build GRCh38
rs762407914 Chr.5: 39285220-39285220 on Build GRCh38
rs1020261331 Chr.5: 39285605-39285605 on Build GRCh38
rs375344631 Chr.5: 39285190-39285190 on Build GRCh38
rs779153625 Chr.5: 39285189-39285189 on Build GRCh38
rs183108180 Chr.5: 39285162-39285162 on Build GRCh38
rs539072742 Chr.5: 39285382-39285382 on Build GRCh38
rs150275549 Chr.5: 39285227-39285227 on Build GRCh38
rs142450396 Chr.5: 39285465-39285465 on Build GRCh38
rs987682623 Chr.5: 39285545-39285545 on Build GRCh38
rs367587173 Chr.5: 39285304-39285304 on Build GRCh38
rs943102078 Chr.5: 39285321-39285321 on Build GRCh38
rs772422977 Chr.5: 39285194-39285194 on Build GRCh38
rs1004480420 Chr.5: 39285603-39285603 on Build GRCh38
rs898908553 Chr.5: 39285513-39285513 on Build GRCh38
rs946404388 Chr.5: 39285359-39285359 on Build GRCh38
rs766372878 Chr.5: 39285284-39285284 on Build GRCh38
rs974894167 Chr.5: 39285595-39285595 on Build GRCh38
rs138480043 Chr.5: 39285209-39285209 on Build GRCh38
rs919524750 Chr.5: 39285295-39285295 on Build GRCh38
rs951040419 Chr.5: 39285543-39285543 on Build GRCh38
rs991502142 Chr.5: 39285231-39285231 on Build GRCh38

View Full Product Details