Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 41142094-41142634 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C6
Entrez Gene ID
729
Gene Name
complement component 6
Gene Aliases
-
UniGene
Hs.481992
Gene Chromosome Location
Chr.5: 41142116-41261486 on Build GRCh38
Transcripts
NM_000065.3 NM_001115131.2 XM_005248357.2 XM_006714496.3 XM_011514114.2 XM_011514115.2 XM_011514116.2 XM_011514117.2 XM_011514118.2 XM_011514119.2 XM_011514121.2 XM_017009818.1 XM_017009819.1

SNP Detail

SNP ID Chromosome Location
rs969436040 Chr.5: 41142607-41142607 on Build GRCh38
rs73750287 Chr.5: 41142188-41142188 on Build GRCh38
rs776821708 Chr.5: 41142513-41142513 on Build GRCh38
rs901547232 Chr.5: 41142496-41142496 on Build GRCh38
rs968243724 Chr.5: 41142132-41142132 on Build GRCh38
rs191402179 Chr.5: 41142289-41142289 on Build GRCh38
rs537049045 Chr.5: 41142128-41142128 on Build GRCh38
rs555488091 Chr.5: 41142613-41142613 on Build GRCh38
rs150777213 Chr.5: 41142167-41142167 on Build GRCh38
rs1045994554 Chr.5: 41142506-41142506 on Build GRCh38
rs984836772 Chr.5: 41142319-41142319 on Build GRCh38
rs564613317 Chr.5: 41142334-41142334 on Build GRCh38
rs1029188759 Chr.5: 41142131-41142131 on Build GRCh38
rs780430648 Chr.5: 41142433-41142433 on Build GRCh38
rs776929039 Chr.5: 41142347-41142369 on Build GRCh38
rs543192104 Chr.5: 41142193-41142193 on Build GRCh38
rs772974738 Chr.5: 41142565-41142565 on Build GRCh38
rs1034351928 Chr.5: 41142545-41142545 on Build GRCh38
rs563497043 Chr.5: 41142391-41142391 on Build GRCh38
rs952037576 Chr.5: 41142550-41142550 on Build GRCh38
rs922444529 Chr.5: 41142608-41142608 on Build GRCh38
rs376366014 Chr.5: 41142281-41142281 on Build GRCh38
rs548097850 Chr.5: 41142486-41142486 on Build GRCh38
rs1025075164 Chr.5: 41142601-41142601 on Build GRCh38
rs1034340933 Chr.5: 41142309-41142309 on Build GRCh38
rs1001516967 Chr.5: 41142540-41142540 on Build GRCh38
rs955513262 Chr.5: 41142610-41142610 on Build GRCh38
rs9200 Chr.5: 41142504-41142504 on Build GRCh38
rs1010496635 Chr.5: 41142194-41142194 on Build GRCh38
rs1423415 Chr.5: 41142583-41142583 on Build GRCh38
rs762649576 Chr.5: 41142175-41142175 on Build GRCh38
rs549858178 Chr.5: 41142404-41142404 on Build GRCh38
rs952154304 Chr.5: 41142314-41142314 on Build GRCh38
rs972675816 Chr.5: 41142377-41142377 on Build GRCh38
rs761207427 Chr.5: 41142164-41142164 on Build GRCh38
rs531644988 Chr.5: 41142368-41142368 on Build GRCh38
rs1033219348 Chr.5: 41142189-41142189 on Build GRCh38
rs1049508794 Chr.5: 41142154-41142154 on Build GRCh38
rs73750288 Chr.5: 41142249-41142249 on Build GRCh38
rs927215528 Chr.5: 41142394-41142394 on Build GRCh38
rs184933625 Chr.5: 41142129-41142129 on Build GRCh38
rs996408625 Chr.5: 41142130-41142130 on Build GRCh38
rs1054173341 Chr.5: 41142490-41142490 on Build GRCh38
rs529234037 Chr.5: 41142482-41142482 on Build GRCh38
rs531036526 Chr.5: 41142438-41142438 on Build GRCh38
rs961159277 Chr.5: 41142352-41142352 on Build GRCh38
rs933913165 Chr.5: 41142449-41142449 on Build GRCh38
rs570260298 Chr.5: 41142574-41142574 on Build GRCh38
rs756733060 Chr.5: 41142564-41142564 on Build GRCh38
rs1423416 Chr.5: 41142530-41142530 on Build GRCh38
rs551637633 Chr.5: 41142531-41142531 on Build GRCh38
rs992283347 Chr.5: 41142584-41142584 on Build GRCh38
rs10067242 Chr.5: 41142471-41142471 on Build GRCh38
rs1040510228 Chr.5: 41142489-41142489 on Build GRCh38
rs948020061 Chr.5: 41142291-41142291 on Build GRCh38

View Full Product Details