Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 42796419-42796923 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC152
Entrez Gene ID
100129792
Gene Name
coiled-coil domain containing 152
Gene Aliases
CH5400
UniGene
Hs.745017
Gene Chromosome Location
Chr.5: 42756818-42802437 on Build GRCh38
Transcripts
NM_001134848.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4980177 c.500C>A p.A167E Chr.5: 42796898-42796898 on Build GRCh38
3735878 c.481G>A p.E161K Chr.5: 42796879-42796879 on Build GRCh38
4821197 c.457G>C p.E153Q Chr.5: 42796855-42796855 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs933772304 Chr.5: 42796846-42796846 on Build GRCh38
rs936004024 Chr.5: 42796600-42796600 on Build GRCh38
rs575347001 Chr.5: 42796791-42796791 on Build GRCh38
rs1033079532 Chr.5: 42796560-42796560 on Build GRCh38
rs771822112 Chr.5: 42796831-42796831 on Build GRCh38
rs933824747 Chr.5: 42796499-42796499 on Build GRCh38
rs909853004 Chr.5: 42796771-42796771 on Build GRCh38
rs952424559 Chr.5: 42796831-42796831 on Build GRCh38
rs1024395554 Chr.5: 42796699-42796699 on Build GRCh38
rs897992326 Chr.5: 42796739-42796739 on Build GRCh38
rs970838080 Chr.5: 42796718-42796718 on Build GRCh38
rs192015308 Chr.5: 42796466-42796466 on Build GRCh38
rs748059143 Chr.5: 42796654-42796654 on Build GRCh38
rs778663883 Chr.5: 42796740-42796740 on Build GRCh38
rs1027886903 Chr.5: 42796751-42796751 on Build GRCh38
rs749198403 Chr.5: 42796896-42796896 on Build GRCh38
rs759218527 Chr.5: 42796806-42796809 on Build GRCh38
rs1056213493 Chr.5: 42796615-42796615 on Build GRCh38
rs1021176431 Chr.5: 42796606-42796606 on Build GRCh38
rs889534660 Chr.5: 42796548-42796548 on Build GRCh38
rs945911967 Chr.5: 42796534-42796534 on Build GRCh38
rs904463761 Chr.5: 42796582-42796582 on Build GRCh38
rs1041626565 Chr.5: 42796575-42796575 on Build GRCh38
rs532990379 Chr.5: 42796650-42796650 on Build GRCh38
rs984248497 Chr.5: 42796768-42796768 on Build GRCh38
rs3866437 Chr.5: 42796773-42796774 on Build GRCh38
rs972616046 Chr.5: 42796775-42796775 on Build GRCh38
rs922431732 Chr.5: 42796527-42796527 on Build GRCh38
rs1014297325 Chr.5: 42796675-42796675 on Build GRCh38
rs768042354 Chr.5: 42796632-42796632 on Build GRCh38
rs775317228 Chr.5: 42796877-42796877 on Build GRCh38
rs774350076 Chr.5: 42796494-42796494 on Build GRCh38
rs1049456391 Chr.5: 42796515-42796515 on Build GRCh38
rs999987679 Chr.5: 42796551-42796551 on Build GRCh38

View Full Product Details