Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 43445002-43445501 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM267
Entrez Gene ID
64417
Gene Name
transmembrane protein 267
Gene Aliases
C5orf28
UniGene
Hs.732093
Gene Chromosome Location
Chr.5: 43444252-43484660 on Build GRCh38
Transcripts
NM_022483.4 XM_005248333.4 XM_005248335.3 XM_006714486.3 XM_011514074.2 XM_011514075.2

SNP Detail

SNP ID Chromosome Location
rs368679925 Chr.5: 43445077-43445077 on Build GRCh38
rs904365257 Chr.5: 43445165-43445165 on Build GRCh38
rs904269229 Chr.5: 43445382-43445382 on Build GRCh38
rs112488837 Chr.5: 43445305-43445305 on Build GRCh38
rs1034268751 Chr.5: 43445179-43445179 on Build GRCh38
rs575737470 Chr.5: 43445419-43445419 on Build GRCh38
rs76385838 Chr.5: 43445142-43445142 on Build GRCh38
rs76745956 Chr.5: 43445316-43445316 on Build GRCh38
rs972180828 Chr.5: 43445257-43445257 on Build GRCh38
rs1040722259 Chr.5: 43445130-43445130 on Build GRCh38
rs1028542651 Chr.5: 43445289-43445289 on Build GRCh38
rs555701001 Chr.5: 43445392-43445392 on Build GRCh38
rs1004693019 Chr.5: 43445205-43445205 on Build GRCh38
rs542925506 Chr.5: 43445246-43445246 on Build GRCh38
rs545080373 Chr.5: 43445355-43445355 on Build GRCh38
rs956509884 Chr.5: 43445401-43445401 on Build GRCh38
rs980399888 Chr.5: 43445451-43445451 on Build GRCh38
rs910837507 Chr.5: 43445313-43445313 on Build GRCh38
rs887237533 Chr.5: 43445195-43445195 on Build GRCh38
rs926528798 Chr.5: 43445473-43445473 on Build GRCh38
rs79193495 Chr.5: 43445315-43445315 on Build GRCh38
rs946637795 Chr.5: 43445137-43445137 on Build GRCh38
rs905082228 Chr.5: 43445162-43445162 on Build GRCh38
rs942379881 Chr.5: 43445128-43445128 on Build GRCh38
rs1056660810 Chr.5: 43445176-43445176 on Build GRCh38
rs944923931 Chr.5: 43445329-43445329 on Build GRCh38
rs544748560 Chr.5: 43445436-43445436 on Build GRCh38
rs922277807 Chr.5: 43445353-43445353 on Build GRCh38
rs1028113094 Chr.5: 43445280-43445280 on Build GRCh38
rs894384067 Chr.5: 43445183-43445183 on Build GRCh38
rs796164094 Chr.5: 43445105-43445105 on Build GRCh38
rs536179828 Chr.5: 43445222-43445222 on Build GRCh38
rs935074945 Chr.5: 43445044-43445044 on Build GRCh38
rs1008436666 Chr.5: 43445332-43445332 on Build GRCh38
rs914875898 Chr.5: 43445120-43445120 on Build GRCh38
rs952283445 Chr.5: 43445301-43445301 on Build GRCh38
rs182804602 Chr.5: 43445249-43445249 on Build GRCh38
rs935884301 Chr.5: 43445391-43445392 on Build GRCh38
rs79228254 Chr.5: 43445318-43445318 on Build GRCh38
rs967717236 Chr.5: 43445450-43445450 on Build GRCh38
rs750249264 Chr.5: 43445025-43445025 on Build GRCh38
rs962324955 Chr.5: 43445254-43445254 on Build GRCh38
rs945670843 Chr.5: 43445354-43445354 on Build GRCh38
rs939174523 Chr.5: 43445166-43445166 on Build GRCh38
rs1041832470 Chr.5: 43445147-43445147 on Build GRCh38
rs150031883 Chr.5: 43445380-43445380 on Build GRCh38

View Full Product Details