Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 43453532-43454052 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM267
Entrez Gene ID
64417
Gene Name
transmembrane protein 267
Gene Aliases
C5orf28
UniGene
Hs.732093
Gene Chromosome Location
Chr.5: 43444252-43484660 on Build GRCh38
Transcripts
NM_022483.4 XM_005248333.4 XM_005248335.3 XM_006714486.3 XM_011514074.2 XM_011514075.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
738717 c.209G>C p.G70A Chr.5: 43453761-43453761 on Build GRCh38
4155831 c.200T>A p.V67E Chr.5: 43453770-43453770 on Build GRCh38
5555824 c.182G>C p.G61A Chr.5: 43453788-43453788 on Build GRCh38
5900891 c.299C>T p.S100F Chr.5: 43453671-43453671 on Build GRCh38
174802 c.21G>T p.K7N Chr.5: 43453949-43453949 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764564128 Chr.5: 43453761-43453761 on Build GRCh38
rs777651825 Chr.5: 43453896-43453896 on Build GRCh38
rs1041154800 Chr.5: 43453854-43453854 on Build GRCh38
rs776722443 Chr.5: 43453718-43453718 on Build GRCh38
rs754632792 Chr.5: 43453880-43453880 on Build GRCh38
rs958230088 Chr.5: 43453895-43453895 on Build GRCh38
rs751366891 Chr.5: 43453980-43453980 on Build GRCh38
rs752069538 Chr.5: 43453869-43453869 on Build GRCh38
rs764769279 Chr.5: 43453961-43453961 on Build GRCh38
rs754613044 Chr.5: 43453774-43453774 on Build GRCh38
rs776450879 Chr.5: 43453973-43453973 on Build GRCh38
rs753068208 Chr.5: 43453748-43453748 on Build GRCh38
rs968725364 Chr.5: 43453975-43453975 on Build GRCh38
rs746963577 Chr.5: 43453908-43453908 on Build GRCh38
rs867752992 Chr.5: 43453889-43453889 on Build GRCh38
rs764010888 Chr.5: 43453969-43453969 on Build GRCh38
rs755293659 Chr.5: 43453611-43453611 on Build GRCh38
rs886313782 Chr.5: 43453671-43453671 on Build GRCh38
rs538015811 Chr.5: 43453557-43453557 on Build GRCh38
rs544046875 Chr.5: 43453945-43453945 on Build GRCh38
rs150067423 Chr.5: 43453690-43453690 on Build GRCh38
rs749993631 Chr.5: 43453771-43453771 on Build GRCh38
rs1006452354 Chr.5: 43453695-43453695 on Build GRCh38
rs772100712 Chr.5: 43453698-43453698 on Build GRCh38
rs759744689 Chr.5: 43453729-43453729 on Build GRCh38
rs931208291 Chr.5: 43453689-43453689 on Build GRCh38
rs766690171 Chr.5: 43453717-43453717 on Build GRCh38
rs1047476545 Chr.5: 43453628-43453628 on Build GRCh38
rs1036650852 Chr.5: 43453601-43453601 on Build GRCh38
rs753343868 Chr.5: 43453954-43453954 on Build GRCh38
rs763915505 Chr.5: 43453971-43453974 on Build GRCh38
rs779155225 Chr.5: 43453589-43453589 on Build GRCh38
rs745939959 Chr.5: 43453927-43453927 on Build GRCh38
rs201366361 Chr.5: 43453772-43453772 on Build GRCh38
rs761593719 Chr.5: 43453974-43453976 on Build GRCh38
rs775686061 Chr.5: 43453933-43453933 on Build GRCh38
rs758271611 Chr.5: 43453892-43453892 on Build GRCh38
rs916732523 Chr.5: 43453953-43453953 on Build GRCh38
rs540944830 Chr.5: 43453773-43453773 on Build GRCh38
rs760820309 Chr.5: 43453716-43453716 on Build GRCh38
rs748120615 Chr.5: 43453697-43453697 on Build GRCh38
rs757191385 Chr.5: 43453987-43453987 on Build GRCh38
rs776723482 Chr.5: 43453825-43453825 on Build GRCh38
rs773471558 Chr.5: 43453706-43453706 on Build GRCh38
rs144425963 Chr.5: 43453843-43453843 on Build GRCh38
rs564786776 Chr.5: 43453881-43453881 on Build GRCh38
rs755813985 Chr.5: 43453674-43453674 on Build GRCh38
rs990038097 Chr.5: 43453940-43453940 on Build GRCh38
rs745352757 Chr.5: 43453672-43453672 on Build GRCh38
rs768043764 Chr.5: 43453947-43453947 on Build GRCh38
rs774653322 Chr.5: 43453944-43453944 on Build GRCh38
rs758851091 Chr.5: 43453648-43453648 on Build GRCh38
rs763098840 Chr.5: 43453935-43453935 on Build GRCh38
rs780713965 Chr.5: 43453669-43453669 on Build GRCh38
rs777845546 Chr.5: 43453804-43453804 on Build GRCh38
rs770859448 Chr.5: 43453921-43453921 on Build GRCh38
rs770928533 Chr.5: 43453824-43453824 on Build GRCh38
rs759706845 Chr.5: 43453834-43453834 on Build GRCh38
rs781177999 Chr.5: 43454007-43454007 on Build GRCh38
rs752411258 Chr.5: 43453965-43453965 on Build GRCh38
rs763291898 Chr.5: 43453846-43453846 on Build GRCh38
rs764652340 Chr.5: 43453866-43453866 on Build GRCh38
rs781171417 Chr.5: 43453925-43453925 on Build GRCh38
rs758945061 Chr.5: 43453758-43453758 on Build GRCh38
rs753352277 Chr.5: 43453877-43453877 on Build GRCh38
rs765540114 Chr.5: 43453740-43453740 on Build GRCh38
rs753155963 Chr.5: 43453618-43453618 on Build GRCh38
rs530630676 Chr.5: 43453928-43453928 on Build GRCh38
rs148412581 Chr.5: 43453876-43453876 on Build GRCh38
rs759078331 Chr.5: 43453960-43453960 on Build GRCh38
rs778610303 Chr.5: 43453778-43453778 on Build GRCh38
rs747932144 Chr.5: 43453796-43453796 on Build GRCh38
rs779559729 Chr.5: 43453686-43453686 on Build GRCh38
rs917626525 Chr.5: 43453568-43453568 on Build GRCh38
rs572796924 Chr.5: 43453839-43453839 on Build GRCh38
rs774141821 Chr.5: 43453691-43453691 on Build GRCh38
rs1010269451 Chr.5: 43453901-43453901 on Build GRCh38
rs201364406 Chr.5: 43453614-43453614 on Build GRCh38
rs201911245 Chr.5: 43453829-43453829 on Build GRCh38
rs552981404 Chr.5: 43453813-43453813 on Build GRCh38
rs537091081 Chr.5: 43453709-43453709 on Build GRCh38
rs752601534 Chr.5: 43453888-43453888 on Build GRCh38
rs1044472860 Chr.5: 43453579-43453579 on Build GRCh38
rs769026206 Chr.5: 43453936-43453936 on Build GRCh38
rs760611335 Chr.5: 43453747-43453749 on Build GRCh38
rs749178169 Chr.5: 43453687-43453687 on Build GRCh38
rs371101482 Chr.5: 43453966-43453966 on Build GRCh38

View Full Product Details