Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 43486623-43487139 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C5orf34
Entrez Gene ID
375444
Gene Name
chromosome 5 open reading frame 34
Gene Aliases
-
UniGene
Hs.661641
Gene Chromosome Location
Chr.5: 43486699-43515433 on Build GRCh38
Transcripts
NM_198566.3 XM_011514038.2 XM_017009445.1 XM_017009446.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1246877 c.1882G>A p.D628N Chr.5: 43486950-43486950 on Build GRCh38
738715 c.1780T>C p.S594P Chr.5: 43487052-43487052 on Build GRCh38
1437673 c.1864_1865insA p.T622fs*3 Chr.5: 43486967-43486968 on Build GRCh38
738716 c.1898T>G p.L633R Chr.5: 43486934-43486934 on Build GRCh38
5868560 c.1750C>T p.Q584* Chr.5: 43487082-43487082 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761561838 Chr.5: 43487111-43487111 on Build GRCh38
rs747581523 Chr.5: 43486974-43486974 on Build GRCh38
rs773770959 Chr.5: 43486890-43486890 on Build GRCh38
rs766284107 Chr.5: 43487030-43487030 on Build GRCh38
rs192180879 Chr.5: 43486717-43486717 on Build GRCh38
rs538642655 Chr.5: 43486814-43486814 on Build GRCh38
rs971179668 Chr.5: 43486848-43486848 on Build GRCh38
rs747312427 Chr.5: 43487065-43487065 on Build GRCh38
rs901803644 Chr.5: 43486946-43486946 on Build GRCh38
rs779100365 Chr.5: 43486954-43486954 on Build GRCh38
rs943402560 Chr.5: 43486931-43486931 on Build GRCh38
rs748529637 Chr.5: 43486883-43486883 on Build GRCh38
rs915681020 Chr.5: 43486839-43486839 on Build GRCh38
rs373065343 Chr.5: 43486663-43486663 on Build GRCh38
rs568229815 Chr.5: 43486862-43486862 on Build GRCh38
rs548312003 Chr.5: 43486937-43486937 on Build GRCh38
rs750391776 Chr.5: 43486933-43486933 on Build GRCh38
rs775188344 Chr.5: 43486988-43486988 on Build GRCh38
rs761314920 Chr.5: 43487010-43487010 on Build GRCh38
rs749338613 Chr.5: 43486950-43486950 on Build GRCh38
rs1005924588 Chr.5: 43486671-43486671 on Build GRCh38
rs762539928 Chr.5: 43486989-43486989 on Build GRCh38
rs757071885 Chr.5: 43486914-43486914 on Build GRCh38
rs772495959 Chr.5: 43486967-43486967 on Build GRCh38
rs140365787 Chr.5: 43487108-43487108 on Build GRCh38
rs761028856 Chr.5: 43486891-43486891 on Build GRCh38
rs80313047 Chr.5: 43486704-43486704 on Build GRCh38
rs549749867 Chr.5: 43486824-43486824 on Build GRCh38
rs147780634 Chr.5: 43486878-43486878 on Build GRCh38
rs574902419 Chr.5: 43486908-43486908 on Build GRCh38
rs989536949 Chr.5: 43487004-43487004 on Build GRCh38
rs746311253 Chr.5: 43487106-43487106 on Build GRCh38
rs897627879 Chr.5: 43486658-43486658 on Build GRCh38
rs760647777 Chr.5: 43487018-43487018 on Build GRCh38
rs909110300 Chr.5: 43486900-43486900 on Build GRCh38
rs751370528 Chr.5: 43486909-43486909 on Build GRCh38
rs757988454 Chr.5: 43486720-43486720 on Build GRCh38
rs748248548 Chr.5: 43486958-43486958 on Build GRCh38
rs575807724 Chr.5: 43486829-43486829 on Build GRCh38
rs977931391 Chr.5: 43486934-43486934 on Build GRCh38
rs375128668 Chr.5: 43486895-43486895 on Build GRCh38
rs924059924 Chr.5: 43486730-43486730 on Build GRCh38
rs559036373 Chr.5: 43486815-43486815 on Build GRCh38
rs752984794 Chr.5: 43487038-43487038 on Build GRCh38
rs764861448 Chr.5: 43486893-43486893 on Build GRCh38
rs114878845 Chr.5: 43486935-43486935 on Build GRCh38
rs144358871 Chr.5: 43487041-43487041 on Build GRCh38
rs772519443 Chr.5: 43486885-43486885 on Build GRCh38
rs780140915 Chr.5: 43486948-43486948 on Build GRCh38
rs567072222 Chr.5: 43486713-43486713 on Build GRCh38
rs959642530 Chr.5: 43487071-43487071 on Build GRCh38
rs767370833 Chr.5: 43487012-43487012 on Build GRCh38
rs544594256 Chr.5: 43487094-43487094 on Build GRCh38
rs771685963 Chr.5: 43486758-43486758 on Build GRCh38
rs527262832 Chr.5: 43486651-43486651 on Build GRCh38
rs764928417 Chr.5: 43486962-43486963 on Build GRCh38
rs755052206 Chr.5: 43487031-43487031 on Build GRCh38
rs965011721 Chr.5: 43486705-43486705 on Build GRCh38
rs532874327 Chr.5: 43487033-43487033 on Build GRCh38
rs76623878 Chr.5: 43486761-43486761 on Build GRCh38
rs780055107 Chr.5: 43486864-43486864 on Build GRCh38
rs774058086 Chr.5: 43487000-43487000 on Build GRCh38
rs763908529 Chr.5: 43486994-43486994 on Build GRCh38
rs757853125 Chr.5: 43487007-43487009 on Build GRCh38
rs750022308 Chr.5: 43486987-43486988 on Build GRCh38
rs552956108 Chr.5: 43486745-43486745 on Build GRCh38
rs145111126 Chr.5: 43486927-43486927 on Build GRCh38
rs750149096 Chr.5: 43487013-43487013 on Build GRCh38
rs768828300 Chr.5: 43486874-43486874 on Build GRCh38
rs923153728 Chr.5: 43486716-43486716 on Build GRCh38
rs886228700 Chr.5: 43486866-43486866 on Build GRCh38
rs531304003 Chr.5: 43486793-43486793 on Build GRCh38
rs771532139 Chr.5: 43486981-43486981 on Build GRCh38
rs765876893 Chr.5: 43487091-43487096 on Build GRCh38
rs147598915 Chr.5: 43486951-43486951 on Build GRCh38
rs142072137 Chr.5: 43486968-43486968 on Build GRCh38
rs368191439 Chr.5: 43486904-43486904 on Build GRCh38
rs777552661 Chr.5: 43486787-43486787 on Build GRCh38
rs151200604 Chr.5: 43487100-43487100 on Build GRCh38
rs957287711 Chr.5: 43486815-43486815 on Build GRCh38
rs760221047 Chr.5: 43487056-43487056 on Build GRCh38
rs1053737438 Chr.5: 43486772-43486772 on Build GRCh38
rs749538471 Chr.5: 43486867-43486867 on Build GRCh38

View Full Product Details