Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 43486997-43487497 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C5orf34
Entrez Gene ID
375444
Gene Name
chromosome 5 open reading frame 34
Gene Aliases
-
UniGene
Hs.661641
Gene Chromosome Location
Chr.5: 43486699-43515433 on Build GRCh38
Transcripts
NM_198566.3 XM_011514038.2 XM_017009445.1 XM_017009446.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
738715 c.1780T>C p.S594P Chr.5: 43487052-43487052 on Build GRCh38
3410291 c.1738G>T p.G580C Chr.5: 43487094-43487094 on Build GRCh38
5868560 c.1750C>T p.Q584* Chr.5: 43487082-43487082 on Build GRCh38
6367386 c.1745T>C p.L582P Chr.5: 43487087-43487087 on Build GRCh38
1068428 c.1825G>T p.E609* Chr.5: 43487007-43487007 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761561838 Chr.5: 43487111-43487111 on Build GRCh38
rs766284107 Chr.5: 43487030-43487030 on Build GRCh38
rs1047286470 Chr.5: 43487209-43487209 on Build GRCh38
rs1039340733 Chr.5: 43487383-43487387 on Build GRCh38
rs995866752 Chr.5: 43487277-43487277 on Build GRCh38
rs771594872 Chr.5: 43487123-43487123 on Build GRCh38
rs529201356 Chr.5: 43487311-43487311 on Build GRCh38
rs747312427 Chr.5: 43487065-43487065 on Build GRCh38
rs780985632 Chr.5: 43487326-43487326 on Build GRCh38
rs150954842 Chr.5: 43487351-43487351 on Build GRCh38
rs969869853 Chr.5: 43487332-43487332 on Build GRCh38
rs367677335 Chr.5: 43487293-43487293 on Build GRCh38
rs765751101 Chr.5: 43487366-43487366 on Build GRCh38
rs140365787 Chr.5: 43487108-43487108 on Build GRCh38
rs1040245330 Chr.5: 43487452-43487452 on Build GRCh38
rs888602989 Chr.5: 43487228-43487228 on Build GRCh38
rs928271542 Chr.5: 43487116-43487116 on Build GRCh38
rs927937514 Chr.5: 43487352-43487352 on Build GRCh38
rs4866831 Chr.5: 43487406-43487406 on Build GRCh38
rs773112145 Chr.5: 43487125-43487125 on Build GRCh38
rs909154485 Chr.5: 43487346-43487346 on Build GRCh38
rs746311253 Chr.5: 43487106-43487106 on Build GRCh38
rs529820274 Chr.5: 43487325-43487325 on Build GRCh38
rs957883018 Chr.5: 43487323-43487323 on Build GRCh38
rs1022630658 Chr.5: 43487206-43487206 on Build GRCh38
rs530427689 Chr.5: 43487141-43487141 on Build GRCh38
rs941514019 Chr.5: 43487451-43487451 on Build GRCh38
rs375813679 Chr.5: 43487162-43487162 on Build GRCh38
rs1037475283 Chr.5: 43487260-43487260 on Build GRCh38
rs752984794 Chr.5: 43487038-43487038 on Build GRCh38
rs971445160 Chr.5: 43487234-43487234 on Build GRCh38
rs902600017 Chr.5: 43487449-43487449 on Build GRCh38
rs941482874 Chr.5: 43487257-43487257 on Build GRCh38
rs144358871 Chr.5: 43487041-43487041 on Build GRCh38
rs921739649 Chr.5: 43487453-43487453 on Build GRCh38
rs766093281 Chr.5: 43487140-43487140 on Build GRCh38
rs567036705 Chr.5: 43487282-43487282 on Build GRCh38
rs900213879 Chr.5: 43487264-43487264 on Build GRCh38
rs959642530 Chr.5: 43487071-43487071 on Build GRCh38
rs544594256 Chr.5: 43487094-43487094 on Build GRCh38
rs1030406670 Chr.5: 43487291-43487291 on Build GRCh38
rs890394081 Chr.5: 43487292-43487292 on Build GRCh38
rs755052206 Chr.5: 43487031-43487031 on Build GRCh38
rs532874327 Chr.5: 43487033-43487033 on Build GRCh38
rs923311625 Chr.5: 43487403-43487403 on Build GRCh38
rs141100670 Chr.5: 43487126-43487126 on Build GRCh38
rs999384237 Chr.5: 43487297-43487297 on Build GRCh38
rs983045934 Chr.5: 43487429-43487429 on Build GRCh38
rs550219438 Chr.5: 43487238-43487238 on Build GRCh38
rs1042519150 Chr.5: 43487440-43487440 on Build GRCh38
rs910113377 Chr.5: 43487448-43487448 on Build GRCh38
rs765876893 Chr.5: 43487091-43487096 on Build GRCh38
rs35899284 Chr.5: 43487468-43487468 on Build GRCh38
rs151200604 Chr.5: 43487100-43487100 on Build GRCh38
rs760221047 Chr.5: 43487056-43487056 on Build GRCh38

View Full Product Details