Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 44310345-44310842 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGF10
Entrez Gene ID
2255
Gene Name
fibroblast growth factor 10
Gene Aliases
-
UniGene
Hs.248049
Gene Chromosome Location
Chr.5: 44301655-44389780 on Build GRCh38
Transcripts
NM_004465.1 XM_005248264.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3342868 c.386A>G p.N129S Chr.5: 44310470-44310470 on Build GRCh38
1207048 c.383G>A p.S128N Chr.5: 44310473-44310473 on Build GRCh38
376837 c.418C>A p.L140I Chr.5: 44310438-44310438 on Build GRCh38
3725711 c.358G>C p.V120L Chr.5: 44310498-44310498 on Build GRCh38
5438751 c.429+4A>C p.? Chr.5: 44310423-44310423 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs190450475 Chr.5: 44310398-44310398 on Build GRCh38
rs202058604 Chr.5: 44310433-44310433 on Build GRCh38
rs538839467 Chr.5: 44310613-44310613 on Build GRCh38
rs908814240 Chr.5: 44310570-44310570 on Build GRCh38
rs961216513 Chr.5: 44310530-44310530 on Build GRCh38
rs1002382613 Chr.5: 44310768-44310768 on Build GRCh38
rs201775225 Chr.5: 44310408-44310408 on Build GRCh38
rs1049725853 Chr.5: 44310616-44310616 on Build GRCh38
rs373458282 Chr.5: 44310377-44310377 on Build GRCh38
rs960888020 Chr.5: 44310820-44310820 on Build GRCh38
rs890998686 Chr.5: 44310376-44310376 on Build GRCh38
rs776683911 Chr.5: 44310489-44310489 on Build GRCh38
rs200885171 Chr.5: 44310431-44310431 on Build GRCh38
rs201649838 Chr.5: 44310382-44310382 on Build GRCh38
rs202114515 Chr.5: 44310514-44310514 on Build GRCh38
rs200244129 Chr.5: 44310553-44310553 on Build GRCh38
rs932393427 Chr.5: 44310654-44310654 on Build GRCh38
rs759880918 Chr.5: 44310777-44310777 on Build GRCh38
rs752268349 Chr.5: 44310579-44310579 on Build GRCh38
rs201664708 Chr.5: 44310487-44310487 on Build GRCh38
rs149119098 Chr.5: 44310559-44310559 on Build GRCh38
rs763873261 Chr.5: 44310795-44310795 on Build GRCh38
rs376823432 Chr.5: 44310758-44310758 on Build GRCh38
rs1022253671 Chr.5: 44310763-44310763 on Build GRCh38
rs754691079 Chr.5: 44310567-44310567 on Build GRCh38
rs200636378 Chr.5: 44310565-44310565 on Build GRCh38
rs891740468 Chr.5: 44310755-44310755 on Build GRCh38
rs145888320 Chr.5: 44310467-44310467 on Build GRCh38
rs761123609 Chr.5: 44310461-44310461 on Build GRCh38
rs566636962 Chr.5: 44310601-44310601 on Build GRCh38
rs968069825 Chr.5: 44310767-44310767 on Build GRCh38
rs956713353 Chr.5: 44310410-44310410 on Build GRCh38
rs376948496 Chr.5: 44310374-44310374 on Build GRCh38
rs558436601 Chr.5: 44310643-44310643 on Build GRCh38
rs111597502 Chr.5: 44310439-44310439 on Build GRCh38
rs537997753 Chr.5: 44310706-44310706 on Build GRCh38
rs960967610 Chr.5: 44310397-44310397 on Build GRCh38
rs758911531 Chr.5: 44310627-44310627 on Build GRCh38
rs185954950 Chr.5: 44310732-44310732 on Build GRCh38
rs6881797 Chr.5: 44310390-44310390 on Build GRCh38
rs1040348903 Chr.5: 44310690-44310690 on Build GRCh38
rs919673268 Chr.5: 44310624-44310624 on Build GRCh38
rs779571947 Chr.5: 44310378-44310378 on Build GRCh38
rs769710686 Chr.5: 44310498-44310498 on Build GRCh38
rs1033313628 Chr.5: 44310793-44310793 on Build GRCh38
rs764576422 Chr.5: 44310539-44310539 on Build GRCh38
rs201385181 Chr.5: 44310502-44310502 on Build GRCh38
rs104893887 Chr.5: 44310447-44310447 on Build GRCh38
rs1049994055 Chr.5: 44310668-44310668 on Build GRCh38
rs748059190 Chr.5: 44310419-44310419 on Build GRCh38
rs1002256083 Chr.5: 44310776-44310776 on Build GRCh38
rs17234513 Chr.5: 44310718-44310718 on Build GRCh38
rs746766662 Chr.5: 44310452-44310452 on Build GRCh38
rs778500773 Chr.5: 44310578-44310578 on Build GRCh38
rs1043484947 Chr.5: 44310739-44310739 on Build GRCh38
rs149851674 Chr.5: 44310430-44310430 on Build GRCh38
rs104893889 Chr.5: 44310443-44310443 on Build GRCh38
rs199742541 Chr.5: 44310562-44310562 on Build GRCh38
rs771316349 Chr.5: 44310540-44310540 on Build GRCh38
rs776922938 Chr.5: 44310606-44310606 on Build GRCh38
rs752130742 Chr.5: 44310547-44310547 on Build GRCh38
rs200392834 Chr.5: 44310490-44310490 on Build GRCh38
rs180898727 Chr.5: 44310705-44310705 on Build GRCh38
rs866583243 Chr.5: 44310444-44310444 on Build GRCh38

View Full Product Details