Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 55267287-55267797 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHX29
Entrez Gene ID
54505
Gene Name
DEAH-box helicase 29
Gene Aliases
DDX29
UniGene
Hs.593268
Gene Chromosome Location
Chr.5: 55256245-55307693 on Build GRCh38
Transcripts
NM_019030.2 XM_006714653.2 XM_017009588.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6504349 c.3410C>T p.T1137M Chr.5: 55267707-55267707 on Build GRCh38
4456154 c.3367_3368CT>TA p.L1123Y Chr.5: 55267749-55267750 on Build GRCh38
4440013 c.3431+1G>T p.? Chr.5: 55267685-55267685 on Build GRCh38
1437854 c.3351A>G p.R1117R Chr.5: 55267766-55267766 on Build GRCh38
5868623 c.3391G>A p.A1131T Chr.5: 55267726-55267726 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs17647768 Chr.5: 55267652-55267652 on Build GRCh38
rs533672124 Chr.5: 55267515-55267515 on Build GRCh38
rs903745488 Chr.5: 55267469-55267469 on Build GRCh38
rs966928112 Chr.5: 55267379-55267379 on Build GRCh38
rs770981357 Chr.5: 55267701-55267701 on Build GRCh38
rs754873065 Chr.5: 55267744-55267744 on Build GRCh38
rs551655345 Chr.5: 55267724-55267724 on Build GRCh38
rs747972357 Chr.5: 55267681-55267681 on Build GRCh38
rs758163649 Chr.5: 55267767-55267767 on Build GRCh38
rs575769043 Chr.5: 55267379-55267384 on Build GRCh38
rs753201999 Chr.5: 55267657-55267657 on Build GRCh38
rs997642729 Chr.5: 55267563-55267563 on Build GRCh38
rs1012456863 Chr.5: 55267417-55267417 on Build GRCh38
rs772960911 Chr.5: 55267641-55267641 on Build GRCh38
rs752518515 Chr.5: 55267751-55267751 on Build GRCh38
rs754925417 Chr.5: 55267663-55267663 on Build GRCh38
rs963626488 Chr.5: 55267677-55267677 on Build GRCh38
rs1045024421 Chr.5: 55267435-55267435 on Build GRCh38
rs777902525 Chr.5: 55267694-55267694 on Build GRCh38
rs377130178 Chr.5: 55267723-55267723 on Build GRCh38
rs747076813 Chr.5: 55267698-55267698 on Build GRCh38
rs372274520 Chr.5: 55267762-55267762 on Build GRCh38
rs930625362 Chr.5: 55267570-55267570 on Build GRCh38
rs1030796148 Chr.5: 55267622-55267622 on Build GRCh38
rs560999128 Chr.5: 55267485-55267485 on Build GRCh38
rs560420688 Chr.5: 55267572-55267572 on Build GRCh38
rs929782830 Chr.5: 55267342-55267342 on Build GRCh38
rs185772146 Chr.5: 55267618-55267618 on Build GRCh38
rs1055277134 Chr.5: 55267699-55267699 on Build GRCh38
rs753353592 Chr.5: 55267736-55267736 on Build GRCh38
rs974820337 Chr.5: 55267505-55267505 on Build GRCh38
rs765818944 Chr.5: 55267729-55267729 on Build GRCh38
rs929598363 Chr.5: 55267732-55267732 on Build GRCh38
rs1031030535 Chr.5: 55267502-55267502 on Build GRCh38
rs35928396 Chr.5: 55267475-55267476 on Build GRCh38
rs749927828 Chr.5: 55267646-55267646 on Build GRCh38
rs747250997 Chr.5: 55267773-55267773 on Build GRCh38
rs566797481 Chr.5: 55267726-55267726 on Build GRCh38
rs753831407 Chr.5: 55267659-55267659 on Build GRCh38
rs368247028 Chr.5: 55267725-55267725 on Build GRCh38
rs987615766 Chr.5: 55267405-55267405 on Build GRCh38
rs3810849 Chr.5: 55267414-55267414 on Build GRCh38
rs544181447 Chr.5: 55267383-55267383 on Build GRCh38
rs778048404 Chr.5: 55267768-55267768 on Build GRCh38
rs181126528 Chr.5: 55267548-55267548 on Build GRCh38
rs113232371 Chr.5: 55267645-55267645 on Build GRCh38
rs539032528 Chr.5: 55267708-55267708 on Build GRCh38
rs112102767 Chr.5: 55267372-55267372 on Build GRCh38
rs964758732 Chr.5: 55267483-55267483 on Build GRCh38
rs920638032 Chr.5: 55267535-55267535 on Build GRCh38
rs61757584 Chr.5: 55267706-55267706 on Build GRCh38
rs1000772522 Chr.5: 55267495-55267500 on Build GRCh38
rs199765941 Chr.5: 55267707-55267707 on Build GRCh38
rs771586438 Chr.5: 55267424-55267424 on Build GRCh38
rs765184974 Chr.5: 55267750-55267750 on Build GRCh38
rs879615555 Chr.5: 55267571-55267571 on Build GRCh38
rs755620960 Chr.5: 55267651-55267651 on Build GRCh38
rs188969569 Chr.5: 55267669-55267669 on Build GRCh38
rs772292296 Chr.5: 55267686-55267686 on Build GRCh38

View Full Product Details