Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 55994699-55995191 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLJ31104
Entrez Gene ID
441072
Gene Name
uncharacterized LOC441072
Gene Aliases
-
UniGene
Hs.482141
Gene Chromosome Location
Chr.5: 55995167-56003649 on Build GRCh38
Transcripts
NR_102755.1 NM_001190981.1 NM_002184.3 NM_175767.2 NR_120480.1 XM_017009441.1 XM_017009442.1

Gene Symbol
IL6ST
Entrez Gene ID
3572
Gene Name
interleukin 6 signal transducer
Gene Aliases
CD130, CDW130, GP130, IL-6RB
UniGene
Hs.532082
Gene Chromosome Location
Chr.5: 55935095-55994993 on Build GRCh38
Transcripts
NR_102755.1 NM_001190981.1 NM_002184.3 NM_175767.2 NR_120480.1 XM_017009441.1 XM_017009442.1

SNP Detail

SNP ID Chromosome Location
rs958431475 Chr.5: 55994888-55994888 on Build GRCh38
rs1033697726 Chr.5: 55995030-55995051 on Build GRCh38
rs562271733 Chr.5: 55994830-55994830 on Build GRCh38
rs908350176 Chr.5: 55995128-55995128 on Build GRCh38
rs927791522 Chr.5: 55994873-55994873 on Build GRCh38
rs372387775 Chr.5: 55994988-55994988 on Build GRCh38
rs1041284759 Chr.5: 55995080-55995080 on Build GRCh38
rs1022259645 Chr.5: 55994752-55994752 on Build GRCh38
rs757660839 Chr.5: 55994961-55994961 on Build GRCh38
rs965053139 Chr.5: 55995063-55995063 on Build GRCh38
rs535927064 Chr.5: 55994975-55994975 on Build GRCh38
rs781442015 Chr.5: 55995164-55995164 on Build GRCh38
rs1019781092 Chr.5: 55994770-55994770 on Build GRCh38
rs1024611638 Chr.5: 55994840-55994840 on Build GRCh38
rs961302474 Chr.5: 55994865-55994865 on Build GRCh38
rs1011578960 Chr.5: 55994728-55994728 on Build GRCh38
rs990680000 Chr.5: 55994987-55994987 on Build GRCh38
rs924632936 Chr.5: 55995041-55995041 on Build GRCh38
rs952449530 Chr.5: 55994817-55994817 on Build GRCh38
rs1041361044 Chr.5: 55994926-55994926 on Build GRCh38
rs982021960 Chr.5: 55994870-55994870 on Build GRCh38
rs901303755 Chr.5: 55994930-55994930 on Build GRCh38
rs987859579 Chr.5: 55995035-55995035 on Build GRCh38
rs371791925 Chr.5: 55994724-55994724 on Build GRCh38
rs1005291707 Chr.5: 55995123-55995123 on Build GRCh38
rs952514824 Chr.5: 55994945-55994945 on Build GRCh38
rs965570787 Chr.5: 55994772-55994772 on Build GRCh38
rs1037808830 Chr.5: 55994769-55994769 on Build GRCh38
rs888643042 Chr.5: 55995114-55995114 on Build GRCh38
rs1003169009 Chr.5: 55995167-55995167 on Build GRCh38
rs1038178387 Chr.5: 55994973-55994973 on Build GRCh38
rs1024913557 Chr.5: 55995008-55995008 on Build GRCh38
rs373078515 Chr.5: 55994972-55994972 on Build GRCh38
rs999101055 Chr.5: 55994922-55994922 on Build GRCh38
rs998999853 Chr.5: 55994761-55994761 on Build GRCh38
rs1015245054 Chr.5: 55994959-55994959 on Build GRCh38
rs995380817 Chr.5: 55994776-55994776 on Build GRCh38
rs563161343 Chr.5: 55995099-55995099 on Build GRCh38
rs954895068 Chr.5: 55995043-55995043 on Build GRCh38
rs901439776 Chr.5: 55995092-55995092 on Build GRCh38
rs1043568927 Chr.5: 55995032-55995032 on Build GRCh38
rs887062515 Chr.5: 55994821-55994821 on Build GRCh38
rs1047844395 Chr.5: 55995110-55995110 on Build GRCh38
rs1005739159 Chr.5: 55994947-55994947 on Build GRCh38
rs947346678 Chr.5: 55994904-55994904 on Build GRCh38
rs540315405 Chr.5: 55994910-55994910 on Build GRCh38
rs987654637 Chr.5: 55995044-55995044 on Build GRCh38
rs932466727 Chr.5: 55994944-55994944 on Build GRCh38
rs910833297 Chr.5: 55995051-55995051 on Build GRCh38
rs551203512 Chr.5: 55994855-55994855 on Build GRCh38
rs543917712 Chr.5: 55994809-55994809 on Build GRCh38
rs987685737 Chr.5: 55994935-55994935 on Build GRCh38
rs527596237 Chr.5: 55994898-55994898 on Build GRCh38
rs530238337 Chr.5: 55995143-55995143 on Build GRCh38
rs988368094 Chr.5: 55994895-55994895 on Build GRCh38
rs569406527 Chr.5: 55994990-55994990 on Build GRCh38
rs1051749468 Chr.5: 55994741-55994741 on Build GRCh38
rs937884770 Chr.5: 55994878-55994878 on Build GRCh38
rs920666582 Chr.5: 55995097-55995098 on Build GRCh38
rs943409417 Chr.5: 55994759-55994759 on Build GRCh38
rs113243265 Chr.5: 55994934-55994934 on Build GRCh38
rs1002376913 Chr.5: 55995010-55995024 on Build GRCh38
rs368794162 Chr.5: 55994857-55994857 on Build GRCh38
rs1013248947 Chr.5: 55994824-55994824 on Build GRCh38
rs916004593 Chr.5: 55994897-55994897 on Build GRCh38
rs898294114 Chr.5: 55994774-55994774 on Build GRCh38
rs528468667 Chr.5: 55994743-55994744 on Build GRCh38
rs1028076712 Chr.5: 55994790-55994790 on Build GRCh38
rs996994962 Chr.5: 55995098-55995098 on Build GRCh38
rs934652803 Chr.5: 55995048-55995048 on Build GRCh38
rs114216969 Chr.5: 55994753-55994753 on Build GRCh38
rs943397262 Chr.5: 55994953-55994954 on Build GRCh38
rs376325207 Chr.5: 55994998-55994998 on Build GRCh38
rs1055765261 Chr.5: 55994882-55994882 on Build GRCh38
rs540028080 Chr.5: 55994996-55994996 on Build GRCh38
rs914150129 Chr.5: 55994896-55994896 on Build GRCh38
rs867878436 Chr.5: 55994948-55994948 on Build GRCh38
rs532945522 Chr.5: 55994839-55994839 on Build GRCh38
rs1013468772 Chr.5: 55994995-55994995 on Build GRCh38
rs931136755 Chr.5: 55994983-55994983 on Build GRCh38
rs1049905670 Chr.5: 55995100-55995100 on Build GRCh38
rs1001902576 Chr.5: 55995026-55995026 on Build GRCh38
rs1051188104 Chr.5: 55995011-55995011 on Build GRCh38
rs959164731 Chr.5: 55994980-55994980 on Build GRCh38
rs573360530 Chr.5: 55994787-55994787 on Build GRCh38
rs983818213 Chr.5: 55994828-55994828 on Build GRCh38
rs960987621 Chr.5: 55995150-55995150 on Build GRCh38
rs903211966 Chr.5: 55994918-55994918 on Build GRCh38
rs961060290 Chr.5: 55994963-55994963 on Build GRCh38

View Full Product Details