Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 60597447-60597982 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DEPDC1B
Entrez Gene ID
55789
Gene Name
DEP domain containing 1B
Gene Aliases
BRCC3, XTP1
UniGene
Hs.482233
Gene Chromosome Location
Chr.5: 60596912-60700166 on Build GRCh38
Transcripts
NM_001145208.1 NM_018369.2 XM_011543509.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3855779 c.1539A>G p.A513A Chr.5: 60597804-60597804 on Build GRCh38
3345419 c.1477G>A p.E493K Chr.5: 60597866-60597866 on Build GRCh38
3345417 c.1528C>T p.L510L Chr.5: 60597815-60597815 on Build GRCh38
1069198 c.1446T>C p.P482P Chr.5: 60597897-60597897 on Build GRCh38
1069197 c.1582C>T p.R528* Chr.5: 60597761-60597761 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs961493430 Chr.5: 60597905-60597905 on Build GRCh38
rs966461421 Chr.5: 60597710-60597710 on Build GRCh38
rs906009666 Chr.5: 60597614-60597614 on Build GRCh38
rs923142437 Chr.5: 60597803-60597803 on Build GRCh38
rs1014493115 Chr.5: 60597520-60597520 on Build GRCh38
rs749788502 Chr.5: 60597828-60597828 on Build GRCh38
rs556463378 Chr.5: 60597747-60597747 on Build GRCh38
rs770495076 Chr.5: 60597847-60597847 on Build GRCh38
rs1038191172 Chr.5: 60597953-60597953 on Build GRCh38
rs768244156 Chr.5: 60597633-60597633 on Build GRCh38
rs770577370 Chr.5: 60597735-60597735 on Build GRCh38
rs749735121 Chr.5: 60597715-60597715 on Build GRCh38
rs201403911 Chr.5: 60597954-60597954 on Build GRCh38
rs748495095 Chr.5: 60597835-60597835 on Build GRCh38
rs750658887 Chr.5: 60597881-60597881 on Build GRCh38
rs779417976 Chr.5: 60597941-60597941 on Build GRCh38
rs903337857 Chr.5: 60597570-60597570 on Build GRCh38
rs369283471 Chr.5: 60597860-60597860 on Build GRCh38
rs974694045 Chr.5: 60597770-60597770 on Build GRCh38
rs769126100 Chr.5: 60597717-60597717 on Build GRCh38
rs757447437 Chr.5: 60597801-60597801 on Build GRCh38
rs943323753 Chr.5: 60597492-60597492 on Build GRCh38
rs1013167823 Chr.5: 60597673-60597673 on Build GRCh38
rs774034083 Chr.5: 60597869-60597869 on Build GRCh38
rs866755598 Chr.5: 60597539-60597539 on Build GRCh38
rs781414143 Chr.5: 60597807-60597807 on Build GRCh38
rs745442721 Chr.5: 60597855-60597855 on Build GRCh38
rs367566056 Chr.5: 60597956-60597956 on Build GRCh38
rs760211801 Chr.5: 60597722-60597722 on Build GRCh38
rs755431500 Chr.5: 60597829-60597829 on Build GRCh38
rs749416215 Chr.5: 60597936-60597936 on Build GRCh38
rs987101234 Chr.5: 60597846-60597846 on Build GRCh38
rs773825743 Chr.5: 60597677-60597677 on Build GRCh38
rs147993338 Chr.5: 60597814-60597814 on Build GRCh38
rs376138587 Chr.5: 60597575-60597575 on Build GRCh38
rs375582832 Chr.5: 60597880-60597880 on Build GRCh38
rs942186721 Chr.5: 60597608-60597608 on Build GRCh38
rs563776701 Chr.5: 60597863-60597863 on Build GRCh38
rs886498354 Chr.5: 60597597-60597597 on Build GRCh38
rs761496449 Chr.5: 60597942-60597943 on Build GRCh38
rs372551359 Chr.5: 60597868-60597868 on Build GRCh38
rs751333107 Chr.5: 60597799-60597799 on Build GRCh38
rs776026620 Chr.5: 60597743-60597743 on Build GRCh38
rs1047843115 Chr.5: 60597566-60597566 on Build GRCh38
rs756307854 Chr.5: 60597914-60597914 on Build GRCh38
rs766535102 Chr.5: 60597924-60597924 on Build GRCh38
rs776342890 Chr.5: 60597850-60597850 on Build GRCh38
rs575323127 Chr.5: 60597761-60597761 on Build GRCh38
rs769240764 Chr.5: 60597620-60597620 on Build GRCh38
rs756309095 Chr.5: 60597709-60597711 on Build GRCh38
rs755463243 Chr.5: 60597933-60597933 on Build GRCh38
rs750530507 Chr.5: 60597809-60597809 on Build GRCh38
rs762912245 Chr.5: 60597865-60597865 on Build GRCh38
rs753927078 Chr.5: 60597925-60597925 on Build GRCh38
rs777872765 Chr.5: 60597917-60597917 on Build GRCh38
rs536280731 Chr.5: 60597744-60597744 on Build GRCh38
rs543968003 Chr.5: 60597786-60597786 on Build GRCh38
rs933273636 Chr.5: 60597557-60597557 on Build GRCh38
rs543172318 Chr.5: 60597720-60597720 on Build GRCh38
rs550002273 Chr.5: 60597867-60597867 on Build GRCh38
rs574305437 Chr.5: 60597714-60597714 on Build GRCh38
rs780761454 Chr.5: 60597824-60597824 on Build GRCh38
rs1036347445 Chr.5: 60597639-60597639 on Build GRCh38
rs896164572 Chr.5: 60597655-60597655 on Build GRCh38
rs975061949 Chr.5: 60597526-60597526 on Build GRCh38
rs1037905254 Chr.5: 60597610-60597610 on Build GRCh38
rs781503102 Chr.5: 60597630-60597630 on Build GRCh38
rs752619351 Chr.5: 60597760-60597760 on Build GRCh38
rs554458770 Chr.5: 60597700-60597700 on Build GRCh38
rs1001835217 Chr.5: 60597638-60597638 on Build GRCh38

View Full Product Details