Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 60638659-60639168 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DEPDC1B
Entrez Gene ID
55789
Gene Name
DEP domain containing 1B
Gene Aliases
BRCC3, XTP1
UniGene
Hs.482233
Gene Chromosome Location
Chr.5: 60596912-60700166 on Build GRCh38
Transcripts
NM_001145208.1 NM_018369.2 XM_011543509.2 XM_011543510.2 XM_017009629.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1069201 c.813C>A p.V271V Chr.5: 60638835-60638835 on Build GRCh38
1069200 c.875T>G p.F292C Chr.5: 60638773-60638773 on Build GRCh38
1249984 c.758-1G>T p.? Chr.5: 60638891-60638891 on Build GRCh38
5434324 c.868C>A p.H290N Chr.5: 60638780-60638780 on Build GRCh38
3617314 c.851A>C p.E284A Chr.5: 60638797-60638797 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770406676 Chr.5: 60638886-60638886 on Build GRCh38
rs1045608627 Chr.5: 60639017-60639017 on Build GRCh38
rs776537987 Chr.5: 60638772-60638772 on Build GRCh38
rs772916014 Chr.5: 60638877-60638877 on Build GRCh38
rs1005697131 Chr.5: 60638827-60638827 on Build GRCh38
rs373860185 Chr.5: 60638919-60638919 on Build GRCh38
rs764307976 Chr.5: 60638812-60638812 on Build GRCh38
rs985980942 Chr.5: 60638964-60638964 on Build GRCh38
rs768571105 Chr.5: 60639101-60639101 on Build GRCh38
rs759947974 Chr.5: 60638840-60638841 on Build GRCh38
rs562905475 Chr.5: 60638971-60638971 on Build GRCh38
rs1022415574 Chr.5: 60638933-60638933 on Build GRCh38
rs746560300 Chr.5: 60638881-60638881 on Build GRCh38
rs756653073 Chr.5: 60638861-60638861 on Build GRCh38
rs1028390599 Chr.5: 60639130-60639130 on Build GRCh38
rs866212546 Chr.5: 60639133-60639133 on Build GRCh38
rs371478415 Chr.5: 60638882-60638882 on Build GRCh38
rs374813881 Chr.5: 60638860-60638860 on Build GRCh38
rs576317214 Chr.5: 60638952-60638952 on Build GRCh38
rs751781074 Chr.5: 60638928-60638928 on Build GRCh38
rs762802095 Chr.5: 60638922-60638922 on Build GRCh38
rs372125843 Chr.5: 60638781-60638781 on Build GRCh38
rs776591200 Chr.5: 60638889-60638889 on Build GRCh38
rs147980798 Chr.5: 60638938-60638938 on Build GRCh38
rs908982288 Chr.5: 60639088-60639088 on Build GRCh38
rs1003510109 Chr.5: 60638892-60638892 on Build GRCh38
rs767732303 Chr.5: 60638930-60638930 on Build GRCh38
rs767918174 Chr.5: 60638890-60638891 on Build GRCh38
rs957639136 Chr.5: 60639110-60639110 on Build GRCh38
rs752487712 Chr.5: 60638793-60638793 on Build GRCh38
rs894216941 Chr.5: 60638970-60638970 on Build GRCh38
rs577690538 Chr.5: 60639089-60639089 on Build GRCh38
rs368712606 Chr.5: 60638701-60638701 on Build GRCh38
rs545207789 Chr.5: 60639039-60639039 on Build GRCh38
rs908820537 Chr.5: 60638716-60638716 on Build GRCh38
rs551278399 Chr.5: 60638905-60638905 on Build GRCh38
rs577259802 Chr.5: 60638820-60638820 on Build GRCh38
rs888608324 Chr.5: 60638787-60638787 on Build GRCh38
rs746289152 Chr.5: 60638702-60638702 on Build GRCh38
rs770257076 Chr.5: 60638708-60638708 on Build GRCh38
rs749611819 Chr.5: 60638865-60638865 on Build GRCh38
rs761406952 Chr.5: 60638744-60638744 on Build GRCh38
rs139641058 Chr.5: 60638850-60638850 on Build GRCh38
rs778526118 Chr.5: 60638685-60638685 on Build GRCh38
rs757449266 Chr.5: 60638929-60638929 on Build GRCh38
rs941502022 Chr.5: 60639112-60639112 on Build GRCh38
rs751654885 Chr.5: 60638825-60638825 on Build GRCh38
rs918917165 Chr.5: 60639031-60639031 on Build GRCh38
rs771709707 Chr.5: 60638747-60638747 on Build GRCh38
rs975738073 Chr.5: 60638999-60638999 on Build GRCh38
rs902989147 Chr.5: 60639055-60639055 on Build GRCh38
rs146323592 Chr.5: 60638765-60638765 on Build GRCh38
rs918981770 Chr.5: 60638718-60638718 on Build GRCh38
rs780591901 Chr.5: 60638864-60638864 on Build GRCh38
rs565090899 Chr.5: 60639085-60639085 on Build GRCh38
rs758560757 Chr.5: 60638798-60638798 on Build GRCh38
rs781775948 Chr.5: 60638851-60638851 on Build GRCh38
rs774182578 Chr.5: 60638724-60638724 on Build GRCh38
rs996848368 Chr.5: 60639125-60639125 on Build GRCh38
rs759492525 Chr.5: 60638893-60638893 on Build GRCh38
rs780088279 Chr.5: 60638974-60638974 on Build GRCh38
rs764277937 Chr.5: 60638923-60638923 on Build GRCh38
rs965498581 Chr.5: 60638725-60638725 on Build GRCh38
rs766371871 Chr.5: 60638767-60638767 on Build GRCh38
rs774616488 Chr.5: 60638737-60638737 on Build GRCh38
rs765047737 Chr.5: 60638790-60638790 on Build GRCh38
rs771631290 Chr.5: 60638874-60638874 on Build GRCh38
rs910442136 Chr.5: 60638969-60638969 on Build GRCh38
rs772613719 Chr.5: 60638753-60638753 on Build GRCh38
rs201620154 Chr.5: 60638916-60638916 on Build GRCh38

View Full Product Details