Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 60753046-60753550 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELOVL7
Entrez Gene ID
79993
Gene Name
ELOVL fatty acid elongase 7
Gene Aliases
-
UniGene
Hs.274256
Gene Chromosome Location
Chr.5: 60751789-60844292 on Build GRCh38
Transcripts
NM_001104558.1 NM_001297617.1 NM_001297618.1 NM_024930.2 XM_005248606.4 XM_005248607.3 XM_006714695.3 XM_011543651.2 XM_017009885.1 XM_017009886.1 XM_017009887.1

SNP Detail

SNP ID Chromosome Location
rs1029653370 Chr.5: 60753342-60753342 on Build GRCh38
rs980596343 Chr.5: 60753441-60753441 on Build GRCh38
rs909930126 Chr.5: 60753373-60753373 on Build GRCh38
rs926548943 Chr.5: 60753204-60753218 on Build GRCh38
rs373855389 Chr.5: 60753314-60753314 on Build GRCh38
rs570605053 Chr.5: 60753215-60753216 on Build GRCh38
rs903615081 Chr.5: 60753240-60753240 on Build GRCh38
rs1022439139 Chr.5: 60753228-60753229 on Build GRCh38
rs1056661756 Chr.5: 60753100-60753100 on Build GRCh38
rs768536243 Chr.5: 60753493-60753493 on Build GRCh38
rs904269827 Chr.5: 60753162-60753162 on Build GRCh38
rs899520119 Chr.5: 60753271-60753271 on Build GRCh38
rs746022323 Chr.5: 60753076-60753076 on Build GRCh38
rs576029297 Chr.5: 60753145-60753146 on Build GRCh38
rs35840031 Chr.5: 60753225-60753226 on Build GRCh38
rs1044927046 Chr.5: 60753160-60753160 on Build GRCh38
rs981290275 Chr.5: 60753355-60753355 on Build GRCh38
rs914042208 Chr.5: 60753106-60753106 on Build GRCh38
rs913838370 Chr.5: 60753500-60753503 on Build GRCh38
rs551819564 Chr.5: 60753199-60753199 on Build GRCh38
rs1000286950 Chr.5: 60753260-60753260 on Build GRCh38
rs73759411 Chr.5: 60753413-60753413 on Build GRCh38
rs1056418111 Chr.5: 60753476-60753476 on Build GRCh38
rs192016127 Chr.5: 60753334-60753334 on Build GRCh38
rs774249282 Chr.5: 60753496-60753496 on Build GRCh38
rs960710582 Chr.5: 60753390-60753390 on Build GRCh38
rs1018401928 Chr.5: 60753273-60753276 on Build GRCh38
rs892154813 Chr.5: 60753216-60753216 on Build GRCh38
rs1004047462 Chr.5: 60753319-60753319 on Build GRCh38
rs2219333 Chr.5: 60753292-60753292 on Build GRCh38
rs565722148 Chr.5: 60753444-60753444 on Build GRCh38
rs1009299347 Chr.5: 60753228-60753228 on Build GRCh38
rs183295547 Chr.5: 60753473-60753473 on Build GRCh38
rs998077878 Chr.5: 60753280-60753280 on Build GRCh38
rs149021475 Chr.5: 60753155-60753155 on Build GRCh38
rs796284274 Chr.5: 60753353-60753353 on Build GRCh38
rs905070732 Chr.5: 60753173-60753173 on Build GRCh38
rs939287221 Chr.5: 60753183-60753183 on Build GRCh38
rs925472058 Chr.5: 60753504-60753504 on Build GRCh38
rs1039356217 Chr.5: 60753217-60753217 on Build GRCh38
rs1000384202 Chr.5: 60753170-60753170 on Build GRCh38
rs926623362 Chr.5: 60753363-60753363 on Build GRCh38
rs575747711 Chr.5: 60753296-60753296 on Build GRCh38
rs573155671 Chr.5: 60753139-60753139 on Build GRCh38
rs930071290 Chr.5: 60753092-60753092 on Build GRCh38
rs542201081 Chr.5: 60753144-60753144 on Build GRCh38
rs756304695 Chr.5: 60753136-60753136 on Build GRCh38
rs562284340 Chr.5: 60753145-60753145 on Build GRCh38
rs548161158 Chr.5: 60753508-60753508 on Build GRCh38

View Full Product Details