Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 60753339-60753838 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELOVL7
Entrez Gene ID
79993
Gene Name
ELOVL fatty acid elongase 7
Gene Aliases
-
UniGene
Hs.274256
Gene Chromosome Location
Chr.5: 60751789-60844292 on Build GRCh38
Transcripts
NM_001104558.1 NM_001297617.1 NM_001297618.1 NM_024930.2 XM_005248606.4 XM_005248607.3 XM_006714695.3 XM_011543651.2 XM_017009885.1 XM_017009886.1 XM_017009887.1

SNP Detail

SNP ID Chromosome Location
rs1034186416 Chr.5: 60753745-60753745 on Build GRCh38
rs980596343 Chr.5: 60753441-60753441 on Build GRCh38
rs909930126 Chr.5: 60753373-60753373 on Build GRCh38
rs568299789 Chr.5: 60753531-60753531 on Build GRCh38
rs899468919 Chr.5: 60753584-60753584 on Build GRCh38
rs933648209 Chr.5: 60753600-60753600 on Build GRCh38
rs1039876194 Chr.5: 60753541-60753541 on Build GRCh38
rs768536243 Chr.5: 60753493-60753493 on Build GRCh38
rs565722148 Chr.5: 60753444-60753444 on Build GRCh38
rs887016787 Chr.5: 60753619-60753619 on Build GRCh38
rs935744785 Chr.5: 60753527-60753527 on Build GRCh38
rs183295547 Chr.5: 60753473-60753473 on Build GRCh38
rs1016751610 Chr.5: 60753624-60753624 on Build GRCh38
rs998377203 Chr.5: 60753795-60753795 on Build GRCh38
rs769045553 Chr.5: 60753765-60753765 on Build GRCh38
rs945007840 Chr.5: 60753612-60753612 on Build GRCh38
rs913838370 Chr.5: 60753500-60753503 on Build GRCh38
rs925472058 Chr.5: 60753504-60753504 on Build GRCh38
rs926623362 Chr.5: 60753363-60753363 on Build GRCh38
rs1002265786 Chr.5: 60753744-60753744 on Build GRCh38
rs772783179 Chr.5: 60753776-60753776 on Build GRCh38
rs1030305348 Chr.5: 60753807-60753807 on Build GRCh38
rs1050788287 Chr.5: 60753613-60753613 on Build GRCh38
rs73759411 Chr.5: 60753413-60753413 on Build GRCh38
rs1007303300 Chr.5: 60753741-60753741 on Build GRCh38
rs13358053 Chr.5: 60753750-60753750 on Build GRCh38
rs1004161965 Chr.5: 60753623-60753623 on Build GRCh38
rs903561796 Chr.5: 60753634-60753634 on Build GRCh38
rs1056418111 Chr.5: 60753476-60753476 on Build GRCh38
rs774249282 Chr.5: 60753496-60753496 on Build GRCh38
rs960710582 Chr.5: 60753390-60753390 on Build GRCh38
rs898343133 Chr.5: 60753725-60753725 on Build GRCh38
rs548161158 Chr.5: 60753508-60753508 on Build GRCh38

View Full Product Details