Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 60844082-60844586 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELOVL7
Entrez Gene ID
79993
Gene Name
ELOVL fatty acid elongase 7
Gene Aliases
-
UniGene
Hs.274256
Gene Chromosome Location
Chr.5: 60751789-60844292 on Build GRCh38
Transcripts
NM_001104558.1 NM_001297617.1 NM_001297618.1 NM_024930.2 XM_005248606.4 XM_006714695.3 XM_017009886.1 XM_017009888.1 XM_017009889.1

SNP Detail

SNP ID Chromosome Location
rs370561056 Chr.5: 60844403-60844403 on Build GRCh38
rs10036497 Chr.5: 60844250-60844250 on Build GRCh38
rs759413908 Chr.5: 60844168-60844168 on Build GRCh38
rs890466998 Chr.5: 60844172-60844172 on Build GRCh38
rs775486968 Chr.5: 60844506-60844506 on Build GRCh38
rs932109702 Chr.5: 60844150-60844150 on Build GRCh38
rs1029941434 Chr.5: 60844488-60844488 on Build GRCh38
rs976963082 Chr.5: 60844291-60844291 on Build GRCh38
rs1000383408 Chr.5: 60844274-60844274 on Build GRCh38
rs1018423668 Chr.5: 60844283-60844283 on Build GRCh38
rs1037040330 Chr.5: 60844221-60844221 on Build GRCh38
rs1053652918 Chr.5: 60844115-60844115 on Build GRCh38
rs555875051 Chr.5: 60844476-60844476 on Build GRCh38
rs941422169 Chr.5: 60844356-60844356 on Build GRCh38
rs995402802 Chr.5: 60844556-60844556 on Build GRCh38
rs573874292 Chr.5: 60844374-60844374 on Build GRCh38
rs552263448 Chr.5: 60844342-60844342 on Build GRCh38
rs148426841 Chr.5: 60844542-60844542 on Build GRCh38
rs149714182 Chr.5: 60844188-60844188 on Build GRCh38
rs555199410 Chr.5: 60844401-60844401 on Build GRCh38
rs903613699 Chr.5: 60844420-60844420 on Build GRCh38
rs922011073 Chr.5: 60844141-60844141 on Build GRCh38
rs926421462 Chr.5: 60844383-60844383 on Build GRCh38
rs529867461 Chr.5: 60844196-60844196 on Build GRCh38
rs909948154 Chr.5: 60844185-60844185 on Build GRCh38
rs537216828 Chr.5: 60844468-60844468 on Build GRCh38
rs1051678124 Chr.5: 60844151-60844151 on Build GRCh38
rs35453042 Chr.5: 60844234-60844234 on Build GRCh38
rs998343875 Chr.5: 60844471-60844471 on Build GRCh38
rs968184855 Chr.5: 60844262-60844262 on Build GRCh38
rs762547915 Chr.5: 60844244-60844244 on Build GRCh38
rs899438156 Chr.5: 60844225-60844225 on Build GRCh38
rs964242943 Chr.5: 60844465-60844465 on Build GRCh38
rs547611532 Chr.5: 60844112-60844112 on Build GRCh38
rs1003383848 Chr.5: 60844245-60844245 on Build GRCh38
rs892170691 Chr.5: 60844254-60844254 on Build GRCh38
rs1018456259 Chr.5: 60844459-60844459 on Build GRCh38
rs1043761845 Chr.5: 60844415-60844415 on Build GRCh38
rs533734355 Chr.5: 60844487-60844487 on Build GRCh38
rs34598791 Chr.5: 60844259-60844259 on Build GRCh38
rs922839960 Chr.5: 60844337-60844337 on Build GRCh38
rs749124609 Chr.5: 60844187-60844187 on Build GRCh38
rs983174127 Chr.5: 60844350-60844350 on Build GRCh38
rs756892546 Chr.5: 60844422-60844422 on Build GRCh38
rs1044855161 Chr.5: 60844202-60844202 on Build GRCh38
rs939186164 Chr.5: 60844389-60844389 on Build GRCh38
rs909987193 Chr.5: 60844352-60844352 on Build GRCh38
rs1009639513 Chr.5: 60844257-60844257 on Build GRCh38
rs144568172 Chr.5: 60844357-60844357 on Build GRCh38
rs1056419051 Chr.5: 60844390-60844390 on Build GRCh38

View Full Product Details