Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 60904698-60905233 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERCC8
Entrez Gene ID
1161
Gene Name
ERCC excision repair 8, CSA ubiquitin ligase complex subunit
Gene Aliases
CKN1, CSA, UVSS2
UniGene
Hs.435237
Gene Chromosome Location
Chr.5: 60873832-60945078 on Build GRCh38
Transcripts
NM_000082.3 NM_001007233.2 NM_001007234.2 NM_001290285.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7410676 c.461C>G p.T154S Chr.5: 60904812-60904812 on Build GRCh38
1251477 c.421G>C p.E141Q Chr.5: 60904852-60904852 on Build GRCh38
4802400 c.428C>T p.T143I Chr.5: 60904845-60904845 on Build GRCh38
3761270 c.435T>C p.Y145Y Chr.5: 60904838-60904838 on Build GRCh38
6656042 c.465G>A p.K155K Chr.5: 60904808-60904808 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770131914 Chr.5: 60904893-60904894 on Build GRCh38
rs957986922 Chr.5: 60904949-60904949 on Build GRCh38
rs192695896 Chr.5: 60904843-60904843 on Build GRCh38
rs4647100 Chr.5: 60904838-60904838 on Build GRCh38
rs540789025 Chr.5: 60905200-60905200 on Build GRCh38
rs920006003 Chr.5: 60905177-60905177 on Build GRCh38
rs906640349 Chr.5: 60905161-60905161 on Build GRCh38
rs370547822 Chr.5: 60904916-60904916 on Build GRCh38
rs895140713 Chr.5: 60904862-60904862 on Build GRCh38
rs184972008 Chr.5: 60905102-60905102 on Build GRCh38
rs938019755 Chr.5: 60904840-60904840 on Build GRCh38
rs1042660283 Chr.5: 60904898-60904898 on Build GRCh38
rs183305163 Chr.5: 60904929-60904929 on Build GRCh38
rs121434325 Chr.5: 60904794-60904794 on Build GRCh38
rs769378958 Chr.5: 60904850-60904850 on Build GRCh38
rs1028621331 Chr.5: 60904964-60904964 on Build GRCh38
rs373930268 Chr.5: 60904975-60904975 on Build GRCh38
rs367886044 Chr.5: 60904867-60904867 on Build GRCh38
rs953034082 Chr.5: 60904975-60904978 on Build GRCh38
rs1010767625 Chr.5: 60904954-60904954 on Build GRCh38
rs768229773 Chr.5: 60904859-60904859 on Build GRCh38
rs754096373 Chr.5: 60904914-60904914 on Build GRCh38
rs374143395 Chr.5: 60904758-60904758 on Build GRCh38
rs780302746 Chr.5: 60904755-60904755 on Build GRCh38
rs1028435505 Chr.5: 60905082-60905082 on Build GRCh38
rs747559514 Chr.5: 60904748-60904749 on Build GRCh38
rs554352832 Chr.5: 60904944-60904945 on Build GRCh38
rs567326648 Chr.5: 60905027-60905027 on Build GRCh38
rs1037302131 Chr.5: 60904820-60904820 on Build GRCh38
rs4647098 Chr.5: 60905029-60905029 on Build GRCh38
rs775226445 Chr.5: 60904851-60904851 on Build GRCh38
rs778737089 Chr.5: 60905128-60905128 on Build GRCh38
rs373783755 Chr.5: 60904854-60904854 on Build GRCh38
rs972921662 Chr.5: 60905057-60905057 on Build GRCh38
rs371402610 Chr.5: 60904846-60904846 on Build GRCh38
rs1644460 Chr.5: 60904806-60904806 on Build GRCh38
rs973937645 Chr.5: 60905174-60905174 on Build GRCh38
rs769008937 Chr.5: 60904765-60904765 on Build GRCh38
rs987644056 Chr.5: 60904979-60904982 on Build GRCh38
rs778897710 Chr.5: 60904766-60904766 on Build GRCh38
rs144440143 Chr.5: 60904956-60904956 on Build GRCh38
rs4647101 Chr.5: 60904772-60904772 on Build GRCh38
rs281875222 Chr.5: 60904795-60904795 on Build GRCh38
rs192837494 Chr.5: 60905055-60905055 on Build GRCh38
rs561001438 Chr.5: 60904801-60904801 on Build GRCh38
rs766619032 Chr.5: 60904908-60904909 on Build GRCh38
rs576287182 Chr.5: 60905084-60905084 on Build GRCh38
rs150952570 Chr.5: 60904864-60904864 on Build GRCh38
rs4647099 Chr.5: 60904915-60904915 on Build GRCh38
rs949699727 Chr.5: 60904891-60904891 on Build GRCh38
rs755117208 Chr.5: 60904765-60904765 on Build GRCh38
rs187635667 Chr.5: 60905014-60905014 on Build GRCh38
rs4647097 Chr.5: 60905166-60905166 on Build GRCh38
rs999562535 Chr.5: 60905165-60905165 on Build GRCh38
rs533924190 Chr.5: 60905008-60905008 on Build GRCh38
rs558369377 Chr.5: 60905119-60905119 on Build GRCh38
rs748481426 Chr.5: 60904825-60904826 on Build GRCh38
rs931586872 Chr.5: 60905070-60905070 on Build GRCh38
rs373296539 Chr.5: 60904920-60904921 on Build GRCh38
rs763257985 Chr.5: 60904897-60904909 on Build GRCh38
rs766049153 Chr.5: 60904905-60904905 on Build GRCh38
rs996504833 Chr.5: 60905059-60905059 on Build GRCh38
rs767533851 Chr.5: 60904866-60904866 on Build GRCh38
rs374611680 Chr.5: 60904784-60904784 on Build GRCh38
rs750293312 Chr.5: 60904893-60904893 on Build GRCh38
rs760543594 Chr.5: 60904904-60904904 on Build GRCh38
rs1048644424 Chr.5: 60905074-60905074 on Build GRCh38
rs961167241 Chr.5: 60905168-60905168 on Build GRCh38
rs148393161 Chr.5: 60904792-60904792 on Build GRCh38
rs167037 Chr.5: 60904824-60904824 on Build GRCh38
rs938117957 Chr.5: 60905187-60905187 on Build GRCh38

View Full Product Details