Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 61159088-61159585 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMIM15
Entrez Gene ID
643155
Gene Name
small integral membrane protein 15
Gene Aliases
C5orf43
UniGene
Hs.508479
Gene Chromosome Location
Chr.5: 61157709-61162475 on Build GRCh38
Transcripts
NM_001048249.3

SNP Detail

SNP ID Chromosome Location
rs751143919 Chr.5: 61159543-61159543 on Build GRCh38
rs556238411 Chr.5: 61159513-61159513 on Build GRCh38
rs900529519 Chr.5: 61159216-61159216 on Build GRCh38
rs192242164 Chr.5: 61159268-61159268 on Build GRCh38
rs368037794 Chr.5: 61159302-61159302 on Build GRCh38
rs1042565410 Chr.5: 61159545-61159545 on Build GRCh38
rs963152809 Chr.5: 61159424-61159424 on Build GRCh38
rs549433993 Chr.5: 61159132-61159132 on Build GRCh38
rs996469452 Chr.5: 61159261-61159261 on Build GRCh38
rs958410783 Chr.5: 61159484-61159484 on Build GRCh38
rs544893518 Chr.5: 61159268-61159269 on Build GRCh38
rs749157246 Chr.5: 61159230-61159230 on Build GRCh38
rs148341488 Chr.5: 61159469-61159471 on Build GRCh38
rs918216490 Chr.5: 61159543-61159546 on Build GRCh38
rs973044955 Chr.5: 61159428-61159428 on Build GRCh38
rs1014674160 Chr.5: 61159364-61159364 on Build GRCh38
rs1053739104 Chr.5: 61159201-61159201 on Build GRCh38
rs567594273 Chr.5: 61159499-61159499 on Build GRCh38
rs534244797 Chr.5: 61159260-61159260 on Build GRCh38
rs774187769 Chr.5: 61159394-61159394 on Build GRCh38
rs563892796 Chr.5: 61159512-61159512 on Build GRCh38
rs571814252 Chr.5: 61159243-61159243 on Build GRCh38
rs762997000 Chr.5: 61159126-61159126 on Build GRCh38
rs991443144 Chr.5: 61159542-61159542 on Build GRCh38
rs753312568 Chr.5: 61159228-61159228 on Build GRCh38

View Full Product Details