Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 62391996-62392541 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DIMT1
Entrez Gene ID
27292
Gene Name
DIM1 dimethyladenosine transferase 1 homolog
Gene Aliases
DIM1, DIMT1L, HSA9761, HUSSY5
UniGene
Hs.731665
Gene Chromosome Location
Chr.5: 62388523-62403939 on Build GRCh38
Transcripts
NM_014473.2 XM_011543323.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1438091 c.761delA p.N254fs*12 Chr.5: 62392202-62392202 on Build GRCh38
5061143 c.755A>G p.E252G Chr.5: 62392208-62392208 on Build GRCh38
1438090 c.789T>C p.N263N Chr.5: 62392174-62392174 on Build GRCh38
4791561 c.742C>T p.Q248* Chr.5: 62392221-62392221 on Build GRCh38
738427 c.784C>A p.H262N Chr.5: 62392179-62392179 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs142896452 Chr.5: 62392288-62392291 on Build GRCh38
rs965159069 Chr.5: 62392037-62392043 on Build GRCh38
rs1025388392 Chr.5: 62392496-62392496 on Build GRCh38
rs530420923 Chr.5: 62392396-62392396 on Build GRCh38
rs939366307 Chr.5: 62392376-62392376 on Build GRCh38
rs746060462 Chr.5: 62392216-62392216 on Build GRCh38
rs897332571 Chr.5: 62392350-62392350 on Build GRCh38
rs764940913 Chr.5: 62392272-62392272 on Build GRCh38
rs945901571 Chr.5: 62392455-62392455 on Build GRCh38
rs763995129 Chr.5: 62392282-62392282 on Build GRCh38
rs868250510 Chr.5: 62392093-62392093 on Build GRCh38
rs61368921 Chr.5: 62392512-62392513 on Build GRCh38
rs1052965917 Chr.5: 62392176-62392176 on Build GRCh38
rs900810467 Chr.5: 62392488-62392488 on Build GRCh38
rs774865131 Chr.5: 62392275-62392275 on Build GRCh38
rs756187928 Chr.5: 62392430-62392433 on Build GRCh38
rs201635129 Chr.5: 62392268-62392268 on Build GRCh38
rs529832708 Chr.5: 62392152-62392152 on Build GRCh38
rs35382223 Chr.5: 62392509-62392510 on Build GRCh38
rs1015598299 Chr.5: 62392321-62392321 on Build GRCh38
rs766147037 Chr.5: 62392045-62392045 on Build GRCh38
rs190142959 Chr.5: 62392229-62392229 on Build GRCh38
rs948054208 Chr.5: 62392128-62392128 on Build GRCh38
rs762466520 Chr.5: 62392278-62392278 on Build GRCh38
rs147264869 Chr.5: 62392370-62392370 on Build GRCh38
rs771935080 Chr.5: 62392286-62392288 on Build GRCh38
rs749529154 Chr.5: 62392475-62392476 on Build GRCh38
rs776415026 Chr.5: 62392052-62392052 on Build GRCh38
rs748522701 Chr.5: 62392253-62392253 on Build GRCh38
rs772530191 Chr.5: 62392264-62392264 on Build GRCh38
rs566196993 Chr.5: 62392474-62392474 on Build GRCh38
rs930154560 Chr.5: 62392053-62392053 on Build GRCh38
rs910973257 Chr.5: 62392030-62392030 on Build GRCh38
rs182372726 Chr.5: 62392469-62392469 on Build GRCh38
rs756287685 Chr.5: 62392131-62392134 on Build GRCh38
rs764024705 Chr.5: 62392154-62392154 on Build GRCh38
rs566689559 Chr.5: 62392420-62392420 on Build GRCh38
rs536115783 Chr.5: 62392147-62392147 on Build GRCh38
rs998260837 Chr.5: 62392493-62392493 on Build GRCh38
rs753864718 Chr.5: 62392163-62392163 on Build GRCh38
rs745676755 Chr.5: 62392274-62392277 on Build GRCh38
rs948333688 Chr.5: 62392316-62392319 on Build GRCh38
rs116552056 Chr.5: 62392477-62392477 on Build GRCh38
rs904482283 Chr.5: 62392225-62392225 on Build GRCh38
rs757109196 Chr.5: 62392202-62392202 on Build GRCh38
rs200168584 Chr.5: 62392238-62392238 on Build GRCh38
rs572025988 Chr.5: 62392314-62392314 on Build GRCh38
rs548588604 Chr.5: 62392482-62392482 on Build GRCh38
rs551176697 Chr.5: 62392140-62392143 on Build GRCh38
rs752199854 Chr.5: 62392136-62392136 on Build GRCh38
rs923643291 Chr.5: 62392513-62392513 on Build GRCh38
rs776468160 Chr.5: 62392265-62392265 on Build GRCh38
rs142197144 Chr.5: 62392170-62392170 on Build GRCh38
rs752854202 Chr.5: 62392034-62392034 on Build GRCh38
rs976906139 Chr.5: 62392042-62392042 on Build GRCh38
rs532800603 Chr.5: 62392462-62392462 on Build GRCh38
rs778950670 Chr.5: 62392212-62392213 on Build GRCh38
rs762174473 Chr.5: 62392040-62392040 on Build GRCh38
rs200709755 Chr.5: 62392188-62392188 on Build GRCh38
rs369036493 Chr.5: 62392510-62392512 on Build GRCh38
rs959635993 Chr.5: 62392461-62392461 on Build GRCh38
rs1042638534 Chr.5: 62392063-62392063 on Build GRCh38
rs759154729 Chr.5: 62392121-62392121 on Build GRCh38
rs762551722 Chr.5: 62392065-62392065 on Build GRCh38
rs774866777 Chr.5: 62392251-62392251 on Build GRCh38
rs984734411 Chr.5: 62392297-62392297 on Build GRCh38
rs764814442 Chr.5: 62392127-62392127 on Build GRCh38
rs563961928 Chr.5: 62392459-62392459 on Build GRCh38
rs756237416 Chr.5: 62392223-62392223 on Build GRCh38
rs977842450 Chr.5: 62392503-62392503 on Build GRCh38
rs780022009 Chr.5: 62392224-62392224 on Build GRCh38
rs1017982652 Chr.5: 62392260-62392262 on Build GRCh38
rs900393285 Chr.5: 62392244-62392244 on Build GRCh38
rs573611624 Chr.5: 62392085-62392085 on Build GRCh38
rs749350723 Chr.5: 62392233-62392233 on Build GRCh38
rs757032349 Chr.5: 62392185-62392185 on Build GRCh38
rs552372560 Chr.5: 62392348-62392348 on Build GRCh38

View Full Product Details