Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 62392726-62393232 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DIMT1
Entrez Gene ID
27292
Gene Name
DIM1 dimethyladenosine transferase 1 homolog
Gene Aliases
DIM1, DIMT1L, HSA9761, HUSSY5
UniGene
Hs.731665
Gene Chromosome Location
Chr.5: 62388523-62403939 on Build GRCh38
Transcripts
NM_014473.2 XM_011543323.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6644020 c.704_705insA p.N235fs*9 Chr.5: 62392949-62392950 on Build GRCh38
1620446 c.664-6T>C p.? Chr.5: 62392996-62392996 on Build GRCh38
6513079 c.673G>T p.G225C Chr.5: 62392981-62392981 on Build GRCh38
7180904 c.684G>A p.R228R Chr.5: 62392970-62392970 on Build GRCh38
1203600 c.710C>T p.T237I Chr.5: 62392944-62392944 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs913915194 Chr.5: 62393030-62393030 on Build GRCh38
rs376566041 Chr.5: 62393181-62393181 on Build GRCh38
rs944055136 Chr.5: 62393025-62393025 on Build GRCh38
rs897268724 Chr.5: 62393040-62393040 on Build GRCh38
rs746635967 Chr.5: 62392942-62392943 on Build GRCh38
rs35014 Chr.5: 62393065-62393065 on Build GRCh38
rs1036980583 Chr.5: 62393026-62393026 on Build GRCh38
rs112654288 Chr.5: 62392904-62392904 on Build GRCh38
rs777758833 Chr.5: 62392875-62392875 on Build GRCh38
rs540721465 Chr.5: 62392816-62392816 on Build GRCh38
rs1046660700 Chr.5: 62393152-62393152 on Build GRCh38
rs957758230 Chr.5: 62392873-62392873 on Build GRCh38
rs552177389 Chr.5: 62392858-62392858 on Build GRCh38
rs1030787653 Chr.5: 62392861-62392861 on Build GRCh38
rs922500298 Chr.5: 62393146-62393147 on Build GRCh38
rs543436948 Chr.5: 62392968-62392968 on Build GRCh38
rs1030672927 Chr.5: 62393183-62393183 on Build GRCh38
rs559659986 Chr.5: 62393201-62393201 on Build GRCh38
rs768268621 Chr.5: 62392982-62392983 on Build GRCh38
rs1005224660 Chr.5: 62393198-62393198 on Build GRCh38
rs1012491680 Chr.5: 62392797-62392797 on Build GRCh38
rs1047197144 Chr.5: 62392828-62392829 on Build GRCh38
rs757482607 Chr.5: 62393074-62393074 on Build GRCh38
rs200893056 Chr.5: 62392901-62392901 on Build GRCh38
rs910647744 Chr.5: 62393077-62393077 on Build GRCh38
rs779857339 Chr.5: 62392937-62392938 on Build GRCh38
rs780259566 Chr.5: 62392917-62392917 on Build GRCh38
rs968094215 Chr.5: 62393051-62393051 on Build GRCh38
rs1012787661 Chr.5: 62393132-62393132 on Build GRCh38
rs370303380 Chr.5: 62392874-62392874 on Build GRCh38
rs1025612773 Chr.5: 62392805-62392805 on Build GRCh38
rs142291348 Chr.5: 62392933-62392933 on Build GRCh38
rs757083591 Chr.5: 62392888-62392888 on Build GRCh38
rs572121315 Chr.5: 62392814-62392814 on Build GRCh38
rs1048577814 Chr.5: 62393104-62393104 on Build GRCh38
rs1041560024 Chr.5: 62392795-62392809 on Build GRCh38
rs999217071 Chr.5: 62393162-62393162 on Build GRCh38
rs1039608832 Chr.5: 62392793-62392793 on Build GRCh38
rs767278025 Chr.5: 62392889-62392889 on Build GRCh38
rs139623518 Chr.5: 62393129-62393129 on Build GRCh38
rs150822218 Chr.5: 62392999-62392999 on Build GRCh38
rs376095015 Chr.5: 62392842-62392842 on Build GRCh38
rs778310826 Chr.5: 62392971-62392971 on Build GRCh38
rs779324117 Chr.5: 62392956-62392956 on Build GRCh38
rs755076198 Chr.5: 62392940-62392940 on Build GRCh38
rs564338862 Chr.5: 62393172-62393172 on Build GRCh38
rs895323449 Chr.5: 62393110-62393110 on Build GRCh38
rs746023889 Chr.5: 62393158-62393158 on Build GRCh38
rs747308911 Chr.5: 62392997-62392997 on Build GRCh38
rs957809018 Chr.5: 62393184-62393184 on Build GRCh38
rs748639651 Chr.5: 62392965-62392965 on Build GRCh38
rs779210414 Chr.5: 62393138-62393138 on Build GRCh38
rs970420080 Chr.5: 62392810-62392810 on Build GRCh38
rs942507325 Chr.5: 62393116-62393116 on Build GRCh38
rs750172035 Chr.5: 62392894-62392894 on Build GRCh38

View Full Product Details