Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 64690202-64690694 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM159B
Entrez Gene ID
100132916
Gene Name
family with sequence similarity 159 member B
Gene Aliases
-
UniGene
Hs.591776
Gene Chromosome Location
Chr.5: 64690308-64718190 on Build GRCh38
Transcripts
NM_001164442.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1735126 c.20T>C p.L7P Chr.5: 64690643-64690643 on Build GRCh38
4629194 c.12C>A p.A4A Chr.5: 64690635-64690635 on Build GRCh38
4638340 c.54C>T p.F18F Chr.5: 64690677-64690677 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs970998679 Chr.5: 64690228-64690228 on Build GRCh38
rs1019659113 Chr.5: 64690669-64690669 on Build GRCh38
rs932921704 Chr.5: 64690317-64690317 on Build GRCh38
rs369187634 Chr.5: 64690267-64690267 on Build GRCh38
rs748358422 Chr.5: 64690654-64690654 on Build GRCh38
rs752987930 Chr.5: 64690376-64690376 on Build GRCh38
rs530421732 Chr.5: 64690446-64690446 on Build GRCh38
rs758727370 Chr.5: 64690443-64690443 on Build GRCh38
rs936203130 Chr.5: 64690275-64690275 on Build GRCh38
rs932764707 Chr.5: 64690595-64690595 on Build GRCh38
rs527924776 Chr.5: 64690366-64690366 on Build GRCh38
rs771193726 Chr.5: 64690668-64690668 on Build GRCh38
rs1037131740 Chr.5: 64690365-64690365 on Build GRCh38
rs939815701 Chr.5: 64690401-64690401 on Build GRCh38
rs898489184 Chr.5: 64690630-64690630 on Build GRCh38
rs1051342729 Chr.5: 64690320-64690320 on Build GRCh38
rs1045644522 Chr.5: 64690671-64690671 on Build GRCh38
rs567832968 Chr.5: 64690395-64690395 on Build GRCh38
rs990749471 Chr.5: 64690279-64690279 on Build GRCh38
rs989906192 Chr.5: 64690546-64690546 on Build GRCh38
rs939982143 Chr.5: 64690348-64690348 on Build GRCh38
rs995585610 Chr.5: 64690394-64690394 on Build GRCh38
rs965487361 Chr.5: 64690488-64690488 on Build GRCh38
rs753190415 Chr.5: 64690470-64690470 on Build GRCh38
rs549866556 Chr.5: 64690523-64690523 on Build GRCh38
rs538743469 Chr.5: 64690558-64690558 on Build GRCh38
rs574604775 Chr.5: 64690634-64690634 on Build GRCh38
rs768033781 Chr.5: 64690404-64690404 on Build GRCh38
rs921514862 Chr.5: 64690519-64690519 on Build GRCh38
rs767120993 Chr.5: 64690585-64690585 on Build GRCh38
rs977209263 Chr.5: 64690237-64690237 on Build GRCh38
rs945720265 Chr.5: 64690554-64690554 on Build GRCh38
rs977469301 Chr.5: 64690506-64690506 on Build GRCh38
rs891132661 Chr.5: 64690332-64690332 on Build GRCh38
rs1002720144 Chr.5: 64690276-64690276 on Build GRCh38
rs534491670 Chr.5: 64690608-64690608 on Build GRCh38
rs1034231740 Chr.5: 64690301-64690301 on Build GRCh38
rs958612499 Chr.5: 64690319-64690319 on Build GRCh38
rs898549496 Chr.5: 64690379-64690379 on Build GRCh38
rs182652187 Chr.5: 64690356-64690356 on Build GRCh38
rs376261361 Chr.5: 64690596-64690596 on Build GRCh38
rs878954612 Chr.5: 64690598-64690598 on Build GRCh38
rs10471637 Chr.5: 64690524-64690524 on Build GRCh38
rs951385819 Chr.5: 64690638-64690638 on Build GRCh38
rs16893053 Chr.5: 64690636-64690636 on Build GRCh38
rs906957102 Chr.5: 64690568-64690568 on Build GRCh38
rs761263223 Chr.5: 64690495-64690495 on Build GRCh38
rs971102618 Chr.5: 64690496-64690496 on Build GRCh38
rs547982780 Chr.5: 64690374-64690374 on Build GRCh38
rs975556080 Chr.5: 64690517-64690517 on Build GRCh38
rs912653735 Chr.5: 64690476-64690476 on Build GRCh38
rs921417406 Chr.5: 64690313-64690313 on Build GRCh38
rs558653318 Chr.5: 64690574-64690574 on Build GRCh38
rs562261255 Chr.5: 64690552-64690552 on Build GRCh38
rs28582519 Chr.5: 64690622-64690622 on Build GRCh38
rs1012788833 Chr.5: 64690485-64690485 on Build GRCh38
rs560084547 Chr.5: 64690599-64690599 on Build GRCh38
rs763449955 Chr.5: 64690658-64690658 on Build GRCh38
rs982864838 Chr.5: 64690419-64690419 on Build GRCh38
rs779055665 Chr.5: 64690643-64690643 on Build GRCh38

View Full Product Details