Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 64768787-64769325 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CWC27
Entrez Gene ID
10283
Gene Name
CWC27 spliceosome associated protein homolog
Gene Aliases
NY-CO-10, SDCCAG-10, SDCCAG10
UniGene
Hs.371372
Gene Chromosome Location
Chr.5: 64768918-65018763 on Build GRCh38
Transcripts
NM_001297644.1 NM_001297645.1 NM_001318000.1 NM_005869.3 XM_011543095.2 XM_011543096.1 XM_017008943.1 XM_011543344.2

Gene Symbol
SREK1IP1
Entrez Gene ID
285672
Gene Name
SREK1 interacting protein 1
Gene Aliases
P18SRP, SFRS12IP1
UniGene
Hs.69504
Gene Chromosome Location
Chr.5: 64718148-64768849 on Build GRCh38
Transcripts
NM_001297644.1 NM_001297645.1 NM_001318000.1 NM_005869.3 XM_011543095.2 XM_011543096.1 XM_017008943.1 XM_011543344.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3749937 c.21G>A p.Q7Q Chr.5: 64769167-64769167 on Build GRCh38
6203858 c.2T>C p.M1T Chr.5: 64769148-64769148 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs199894099 Chr.5: 64769214-64769214 on Build GRCh38
rs1036200993 Chr.5: 64768889-64768889 on Build GRCh38
rs887080705 Chr.5: 64769253-64769253 on Build GRCh38
rs1011691856 Chr.5: 64768928-64768928 on Build GRCh38
rs764177551 Chr.5: 64769233-64769233 on Build GRCh38
rs138792608 Chr.5: 64769166-64769166 on Build GRCh38
rs1043607490 Chr.5: 64768978-64768978 on Build GRCh38
rs369134861 Chr.5: 64769226-64769226 on Build GRCh38
rs529010023 Chr.5: 64769178-64769178 on Build GRCh38
rs1035909603 Chr.5: 64769049-64769049 on Build GRCh38
rs992485226 Chr.5: 64769296-64769296 on Build GRCh38
rs555540258 Chr.5: 64768923-64768923 on Build GRCh38
rs575419615 Chr.5: 64769006-64769006 on Build GRCh38
rs761057727 Chr.5: 64768815-64768815 on Build GRCh38
rs1039957303 Chr.5: 64768906-64768906 on Build GRCh38
rs777071657 Chr.5: 64769098-64769098 on Build GRCh38
rs781748324 Chr.5: 64769239-64769239 on Build GRCh38
rs770263200 Chr.5: 64769177-64769177 on Build GRCh38
rs1004753068 Chr.5: 64768981-64768981 on Build GRCh38
rs1016518836 Chr.5: 64768986-64768986 on Build GRCh38
rs544511154 Chr.5: 64769030-64769030 on Build GRCh38
rs759935061 Chr.5: 64769099-64769099 on Build GRCh38
rs768458110 Chr.5: 64769106-64769106 on Build GRCh38
rs1025757723 Chr.5: 64768963-64768963 on Build GRCh38
rs760666501 Chr.5: 64769130-64769130 on Build GRCh38
rs553078416 Chr.5: 64768824-64768824 on Build GRCh38
rs761460877 Chr.5: 64769111-64769111 on Build GRCh38
rs764622997 Chr.5: 64769074-64769074 on Build GRCh38
rs374415915 Chr.5: 64769144-64769144 on Build GRCh38
rs757849403 Chr.5: 64769021-64769021 on Build GRCh38
rs933294812 Chr.5: 64768907-64768907 on Build GRCh38
rs746278592 Chr.5: 64769168-64769168 on Build GRCh38
rs760062719 Chr.5: 64769204-64769204 on Build GRCh38
rs753697923 Chr.5: 64769143-64769143 on Build GRCh38
rs376182536 Chr.5: 64769200-64769200 on Build GRCh38
rs113652835 Chr.5: 64769285-64769285 on Build GRCh38
rs752418945 Chr.5: 64769227-64769227 on Build GRCh38
rs960289448 Chr.5: 64769083-64769083 on Build GRCh38
rs747799215 Chr.5: 64769188-64769188 on Build GRCh38
rs927183282 Chr.5: 64769058-64769058 on Build GRCh38
rs758537226 Chr.5: 64769156-64769156 on Build GRCh38
rs1056820740 Chr.5: 64768876-64768876 on Build GRCh38
rs564032231 Chr.5: 64769280-64769280 on Build GRCh38
rs746999665 Chr.5: 64769162-64769162 on Build GRCh38
rs754734474 Chr.5: 64769145-64769145 on Build GRCh38
rs752901182 Chr.5: 64769154-64769154 on Build GRCh38
rs139284919 Chr.5: 64769271-64769271 on Build GRCh38
rs963202058 Chr.5: 64768989-64768989 on Build GRCh38
rs769990002 Chr.5: 64769224-64769225 on Build GRCh38
rs373204384 Chr.5: 64769237-64769237 on Build GRCh38
rs191791715 Chr.5: 64769235-64769235 on Build GRCh38
rs774118971 Chr.5: 64769108-64769108 on Build GRCh38
rs923168895 Chr.5: 64769242-64769242 on Build GRCh38
rs980009211 Chr.5: 64768811-64768811 on Build GRCh38
rs951440475 Chr.5: 64768967-64768967 on Build GRCh38
rs184310976 Chr.5: 64769289-64769289 on Build GRCh38
rs774206993 Chr.5: 64769179-64769179 on Build GRCh38
rs959943210 Chr.5: 64769290-64769290 on Build GRCh38
rs767326972 Chr.5: 64768879-64768879 on Build GRCh38
rs200669820 Chr.5: 64769140-64769140 on Build GRCh38
rs188192349 Chr.5: 64769121-64769121 on Build GRCh38
rs759382170 Chr.5: 64769222-64769222 on Build GRCh38
rs746676291 Chr.5: 64769272-64769272 on Build GRCh38
rs903389815 Chr.5: 64769026-64769026 on Build GRCh38
rs757808697 Chr.5: 64769165-64769165 on Build GRCh38
rs1363953 Chr.5: 64769116-64769116 on Build GRCh38
rs1006053202 Chr.5: 64769259-64769259 on Build GRCh38
rs1042775900 Chr.5: 64769125-64769125 on Build GRCh38
rs777909747 Chr.5: 64769158-64769158 on Build GRCh38
rs776525808 Chr.5: 64769211-64769211 on Build GRCh38
rs959725958 Chr.5: 64769072-64769072 on Build GRCh38
rs182629036 Chr.5: 64768918-64768918 on Build GRCh38
rs771558152 Chr.5: 64769196-64769196 on Build GRCh38
rs766178172 Chr.5: 64769138-64769138 on Build GRCh38
rs572699150 Chr.5: 64768883-64768883 on Build GRCh38
rs1007559155 Chr.5: 64768892-64768892 on Build GRCh38
rs749999452 Chr.5: 64769115-64769115 on Build GRCh38
rs765489883 Chr.5: 64769149-64769149 on Build GRCh38
rs901408487 Chr.5: 64769229-64769229 on Build GRCh38
rs912968636 Chr.5: 64768878-64768878 on Build GRCh38
rs758655759 Chr.5: 64769228-64769228 on Build GRCh38
rs765885255 Chr.5: 64768973-64768973 on Build GRCh38
rs550256610 Chr.5: 64768891-64768891 on Build GRCh38
rs2290821 Chr.5: 64769087-64769087 on Build GRCh38
rs766379370 Chr.5: 64769209-64769209 on Build GRCh38

View Full Product Details