Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 65528337-65528840 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CENPK
Entrez Gene ID
64105
Gene Name
centromere protein K
Gene Aliases
AF5alpha, CENP-K, FKSG14, P33, Solt
UniGene
Hs.529778
Gene Chromosome Location
Chr.5: 65488919-65563194 on Build GRCh38
Transcripts
NM_001267038.1 NM_022145.4 XM_011543534.2 XM_011543535.2 XM_011543536.2 XM_011543537.2 XM_011543538.2 XM_011543539.2 XM_011543540.2 XM_011543541.2 XM_011543542.2 XM_017009690.1 XM_017009691.1 XM_017009692.1 XM_017009693.1 XM_017009694.1 XM_017009695.1 XM_017009696.1 XM_017009697.1 XM_017009698.1 XM_017009699.1 XM_017009700.1 XM_017009701.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1718389 c.565C>T p.P189S Chr.5: 65528484-65528484 on Build GRCh38
7137696 c.528C>G p.L176L Chr.5: 65528521-65528521 on Build GRCh38
4407485 c.481G>T p.E161* Chr.5: 65528568-65528568 on Build GRCh38
6104437 c.587A>G p.K196R Chr.5: 65528462-65528462 on Build GRCh38
1486897 c.496A>T p.M166L Chr.5: 65528553-65528553 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs867341199 Chr.5: 65528700-65528700 on Build GRCh38
rs372297020 Chr.5: 65528650-65528650 on Build GRCh38
rs750085640 Chr.5: 65528598-65528599 on Build GRCh38
rs757372616 Chr.5: 65528457-65528457 on Build GRCh38
rs759021447 Chr.5: 65528467-65528467 on Build GRCh38
rs538146502 Chr.5: 65528506-65528506 on Build GRCh38
rs765688833 Chr.5: 65528426-65528426 on Build GRCh38
rs758865586 Chr.5: 65528537-65528537 on Build GRCh38
rs761761092 Chr.5: 65528416-65528418 on Build GRCh38
rs370050260 Chr.5: 65528450-65528450 on Build GRCh38
rs765061204 Chr.5: 65528589-65528589 on Build GRCh38
rs766865224 Chr.5: 65528695-65528697 on Build GRCh38
rs780289086 Chr.5: 65528615-65528615 on Build GRCh38
rs761469552 Chr.5: 65528750-65528750 on Build GRCh38
rs111554721 Chr.5: 65528437-65528437 on Build GRCh38
rs911787066 Chr.5: 65528689-65528689 on Build GRCh38
rs910073611 Chr.5: 65528745-65528745 on Build GRCh38
rs774913472 Chr.5: 65528449-65528449 on Build GRCh38
rs868259372 Chr.5: 65528505-65528505 on Build GRCh38
rs765028924 Chr.5: 65528532-65528532 on Build GRCh38
rs180691282 Chr.5: 65528701-65528701 on Build GRCh38
rs781320311 Chr.5: 65528582-65528582 on Build GRCh38
rs781204320 Chr.5: 65528468-65528468 on Build GRCh38
rs745986169 Chr.5: 65528595-65528595 on Build GRCh38
rs34902707 Chr.5: 65528665-65528665 on Build GRCh38
rs752112029 Chr.5: 65528553-65528553 on Build GRCh38
rs758102685 Chr.5: 65528618-65528619 on Build GRCh38
rs753079695 Chr.5: 65528536-65528536 on Build GRCh38
rs773227049 Chr.5: 65528493-65528493 on Build GRCh38
rs1045332424 Chr.5: 65528803-65528803 on Build GRCh38
rs986090100 Chr.5: 65528682-65528682 on Build GRCh38
rs919241391 Chr.5: 65528384-65528384 on Build GRCh38
rs569355500 Chr.5: 65528495-65528495 on Build GRCh38
rs751083168 Chr.5: 65528623-65528626 on Build GRCh38
rs151068525 Chr.5: 65528471-65528471 on Build GRCh38
rs747020010 Chr.5: 65528451-65528451 on Build GRCh38
rs758950351 Chr.5: 65528438-65528438 on Build GRCh38
rs1048951812 Chr.5: 65528738-65528738 on Build GRCh38
rs915223608 Chr.5: 65528697-65528700 on Build GRCh38
rs866918568 Chr.5: 65528484-65528484 on Build GRCh38
rs576367160 Chr.5: 65528754-65528754 on Build GRCh38
rs934470564 Chr.5: 65528735-65528735 on Build GRCh38
rs965364667 Chr.5: 65528695-65528695 on Build GRCh38
rs376864120 Chr.5: 65528530-65528530 on Build GRCh38
rs895702458 Chr.5: 65528799-65528799 on Build GRCh38
rs757773089 Chr.5: 65528581-65528581 on Build GRCh38
rs753538360 Chr.5: 65528419-65528419 on Build GRCh38
rs756300956 Chr.5: 65528608-65528608 on Build GRCh38
rs765895811 Chr.5: 65528411-65528411 on Build GRCh38
rs779524964 Chr.5: 65528617-65528618 on Build GRCh38
rs751173961 Chr.5: 65528728-65528728 on Build GRCh38
rs753169464 Chr.5: 65528428-65528428 on Build GRCh38
rs1045704909 Chr.5: 65528414-65528414 on Build GRCh38
rs771962047 Chr.5: 65528483-65528483 on Build GRCh38
rs1048517866 Chr.5: 65528374-65528374 on Build GRCh38
rs774114264 Chr.5: 65528481-65528481 on Build GRCh38
rs554755425 Chr.5: 65528655-65528655 on Build GRCh38
rs765914401 Chr.5: 65528499-65528499 on Build GRCh38
rs10940040 Chr.5: 65528703-65528703 on Build GRCh38
rs755228399 Chr.5: 65528422-65528422 on Build GRCh38
rs764451391 Chr.5: 65528538-65528538 on Build GRCh38
rs770061194 Chr.5: 65528473-65528473 on Build GRCh38
rs377239133 Chr.5: 65528494-65528494 on Build GRCh38
rs906225786 Chr.5: 65528427-65528427 on Build GRCh38
rs565655873 Chr.5: 65528378-65528378 on Build GRCh38
rs1012548043 Chr.5: 65528366-65528366 on Build GRCh38

View Full Product Details