Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 65647043-65647590 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRAPPC13
Entrez Gene ID
80006
Gene Name
trafficking protein particle complex 13
Gene Aliases
C5orf44
UniGene
Hs.591760
Gene Chromosome Location
Chr.5: 65624731-65666136 on Build GRCh38
Transcripts
NM_001093755.1 NM_001093756.1 NM_001243737.1 NM_024941.3 XM_011543652.2 XM_017009890.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5516942 c.326G>A p.R109H Chr.5: 65647080-65647080 on Build GRCh38
1438181 c.334C>A p.L112I Chr.5: 65647088-65647088 on Build GRCh38
5516940 c.326G>A p.R109H Chr.5: 65647080-65647080 on Build GRCh38
5640567 c.401A>G p.H134R Chr.5: 65647155-65647155 on Build GRCh38
3947655 c.325C>T p.R109C Chr.5: 65647079-65647079 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779352321 Chr.5: 65647539-65647539 on Build GRCh38
rs34118433 Chr.5: 65647482-65647482 on Build GRCh38
rs528320156 Chr.5: 65647114-65647114 on Build GRCh38
rs754937655 Chr.5: 65647070-65647070 on Build GRCh38
rs563169448 Chr.5: 65647100-65647100 on Build GRCh38
rs373482922 Chr.5: 65647130-65647130 on Build GRCh38
rs187453330 Chr.5: 65647423-65647423 on Build GRCh38
rs371603608 Chr.5: 65647096-65647096 on Build GRCh38
rs879840078 Chr.5: 65647520-65647520 on Build GRCh38
rs971791402 Chr.5: 65647073-65647073 on Build GRCh38
rs551912160 Chr.5: 65647189-65647189 on Build GRCh38
rs1021181042 Chr.5: 65647369-65647369 on Build GRCh38
rs958198564 Chr.5: 65647248-65647251 on Build GRCh38
rs183146614 Chr.5: 65647098-65647098 on Build GRCh38
rs774609307 Chr.5: 65647155-65647155 on Build GRCh38
rs996637035 Chr.5: 65647217-65647217 on Build GRCh38
rs931132712 Chr.5: 65647557-65647557 on Build GRCh38
rs527763711 Chr.5: 65647418-65647418 on Build GRCh38
rs990065911 Chr.5: 65647290-65647290 on Build GRCh38
rs951729629 Chr.5: 65647559-65647559 on Build GRCh38
rs75250458 Chr.5: 65647126-65647126 on Build GRCh38
rs754826487 Chr.5: 65647216-65647216 on Build GRCh38
rs746307546 Chr.5: 65647113-65647113 on Build GRCh38
rs1003859941 Chr.5: 65647168-65647168 on Build GRCh38
rs192591624 Chr.5: 65647459-65647459 on Build GRCh38
rs1010888013 Chr.5: 65647234-65647234 on Build GRCh38
rs200873645 Chr.5: 65647207-65647207 on Build GRCh38
rs201346826 Chr.5: 65647220-65647220 on Build GRCh38
rs771376916 Chr.5: 65647108-65647108 on Build GRCh38
rs754415460 Chr.5: 65647224-65647224 on Build GRCh38
rs147986951 Chr.5: 65647455-65647455 on Build GRCh38
rs777169338 Chr.5: 65647111-65647111 on Build GRCh38
rs990826712 Chr.5: 65647466-65647466 on Build GRCh38
rs71626596 Chr.5: 65647566-65647566 on Build GRCh38
rs775916146 Chr.5: 65647125-65647125 on Build GRCh38
rs746693143 Chr.5: 65647103-65647103 on Build GRCh38
rs374854724 Chr.5: 65647223-65647223 on Build GRCh38
rs770434786 Chr.5: 65647124-65647124 on Build GRCh38
rs757726265 Chr.5: 65647421-65647421 on Build GRCh38
rs994160303 Chr.5: 65647497-65647497 on Build GRCh38
rs777454933 Chr.5: 65647102-65647102 on Build GRCh38
rs752616369 Chr.5: 65647222-65647222 on Build GRCh38
rs200281003 Chr.5: 65647085-65647085 on Build GRCh38
rs549907575 Chr.5: 65647550-65647550 on Build GRCh38
rs1043862587 Chr.5: 65647265-65647265 on Build GRCh38
rs757913989 Chr.5: 65647101-65647101 on Build GRCh38
rs531915793 Chr.5: 65647138-65647138 on Build GRCh38
rs377246348 Chr.5: 65647162-65647162 on Build GRCh38
rs938096052 Chr.5: 65647465-65647465 on Build GRCh38
rs889186823 Chr.5: 65647373-65647373 on Build GRCh38

View Full Product Details