Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 65994617-65995143 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERBIN
Entrez Gene ID
55914
Gene Name
erbb2 interacting protein
Gene Aliases
ERBB2IP, HEL-S-78, LAP2
UniGene
Hs.591774
Gene Chromosome Location
Chr.5: 65926554-66081024 on Build GRCh38
Transcripts
NM_001006600.2 NM_001253697.1 NM_001253698.1 NM_001253699.1 NM_001253701.1 NM_018695.3 XM_005248554.2 XM_005248555.3 XM_006714660.3 XM_017009635.1 XM_017009636.1 XM_017009637.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6655762 c.307+7_307+8insT p.? Chr.5: 65994871-65994872 on Build GRCh38
5649514 c.205C>G p.Q69E Chr.5: 65994762-65994762 on Build GRCh38
6567470 c.307+7C>G p.? Chr.5: 65994871-65994871 on Build GRCh38
6655761 c.307+7_307+8insT p.? Chr.5: 65994871-65994872 on Build GRCh38
4484593 c.282C>T p.L94L Chr.5: 65994839-65994839 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs75060345 Chr.5: 65994885-65994885 on Build GRCh38
rs779948462 Chr.5: 65995051-65995051 on Build GRCh38
rs1036295168 Chr.5: 65994944-65994944 on Build GRCh38
rs756742838 Chr.5: 65994805-65994805 on Build GRCh38
rs929810620 Chr.5: 65995107-65995107 on Build GRCh38
rs745795967 Chr.5: 65994848-65994848 on Build GRCh38
rs769758780 Chr.5: 65994849-65994849 on Build GRCh38
rs551754249 Chr.5: 65994732-65994732 on Build GRCh38
rs200234729 Chr.5: 65994766-65994766 on Build GRCh38
rs779027324 Chr.5: 65994829-65994829 on Build GRCh38
rs540601423 Chr.5: 65995109-65995109 on Build GRCh38
rs756683923 Chr.5: 65994886-65994886 on Build GRCh38
rs753861899 Chr.5: 65994871-65994872 on Build GRCh38
rs375539128 Chr.5: 65994721-65994721 on Build GRCh38
rs928419993 Chr.5: 65994674-65994674 on Build GRCh38
rs769998156 Chr.5: 65994701-65994701 on Build GRCh38
rs576137462 Chr.5: 65994986-65994986 on Build GRCh38
rs544733533 Chr.5: 65995012-65995012 on Build GRCh38
rs558534849 Chr.5: 65995047-65995047 on Build GRCh38
rs200559912 Chr.5: 65994896-65994896 on Build GRCh38
rs751899967 Chr.5: 65994771-65994771 on Build GRCh38
rs201231847 Chr.5: 65994860-65994860 on Build GRCh38
rs767979905 Chr.5: 65994883-65994883 on Build GRCh38
rs981987376 Chr.5: 65994652-65994652 on Build GRCh38
rs914274824 Chr.5: 65994719-65994719 on Build GRCh38
rs573516188 Chr.5: 65994924-65994924 on Build GRCh38
rs985502396 Chr.5: 65995022-65995022 on Build GRCh38
rs911159792 Chr.5: 65995049-65995049 on Build GRCh38
rs760438330 Chr.5: 65994725-65994725 on Build GRCh38
rs534110623 Chr.5: 65994841-65994841 on Build GRCh38
rs367993738 Chr.5: 65994899-65994899 on Build GRCh38
rs201576817 Chr.5: 65994950-65994950 on Build GRCh38
rs143846165 Chr.5: 65994832-65994832 on Build GRCh38
rs540539267 Chr.5: 65994927-65994927 on Build GRCh38
rs970690019 Chr.5: 65994921-65994921 on Build GRCh38
rs944682602 Chr.5: 65994968-65994968 on Build GRCh38
rs762208867 Chr.5: 65994878-65994878 on Build GRCh38
rs920671280 Chr.5: 65994672-65994672 on Build GRCh38
rs748389016 Chr.5: 65994903-65994903 on Build GRCh38
rs762632075 Chr.5: 65994757-65994757 on Build GRCh38
rs993573658 Chr.5: 65994877-65994877 on Build GRCh38
rs141142863 Chr.5: 65995066-65995066 on Build GRCh38
rs773877079 Chr.5: 65994874-65994874 on Build GRCh38
rs367775865 Chr.5: 65994750-65994750 on Build GRCh38
rs781736340 Chr.5: 65994788-65994788 on Build GRCh38
rs757725690 Chr.5: 65994785-65994785 on Build GRCh38
rs1011157742 Chr.5: 65995104-65995104 on Build GRCh38
rs1040955176 Chr.5: 65995093-65995093 on Build GRCh38
rs764743869 Chr.5: 65994749-65994749 on Build GRCh38
rs371566283 Chr.5: 65994914-65994914 on Build GRCh38
rs1049668272 Chr.5: 65994722-65994722 on Build GRCh38
rs367968861 Chr.5: 65994844-65994844 on Build GRCh38
rs780365015 Chr.5: 65994709-65994709 on Build GRCh38
rs1028181321 Chr.5: 65994776-65994776 on Build GRCh38
rs988598414 Chr.5: 65995004-65995004 on Build GRCh38
rs56195564 Chr.5: 65994716-65994716 on Build GRCh38
rs776496813 Chr.5: 65994836-65994836 on Build GRCh38
rs780177969 Chr.5: 65994814-65994814 on Build GRCh38
rs746768463 Chr.5: 65994831-65994831 on Build GRCh38
rs762388266 Chr.5: 65994865-65994865 on Build GRCh38
rs368543697 Chr.5: 65994726-65994726 on Build GRCh38
rs775038585 Chr.5: 65994857-65994857 on Build GRCh38
rs777797458 Chr.5: 65995112-65995112 on Build GRCh38
rs376647035 Chr.5: 65995021-65995021 on Build GRCh38
rs956185058 Chr.5: 65994970-65994970 on Build GRCh38
rs918456474 Chr.5: 65995103-65995103 on Build GRCh38
rs755427400 Chr.5: 65994700-65994700 on Build GRCh38
rs187619364 Chr.5: 65994904-65994904 on Build GRCh38
rs779153488 Chr.5: 65994901-65994901 on Build GRCh38
rs61596028 Chr.5: 65994715-65994715 on Build GRCh38
rs763759993 Chr.5: 65994873-65994873 on Build GRCh38
rs202002843 Chr.5: 65994729-65994729 on Build GRCh38
rs768902243 Chr.5: 65994828-65994828 on Build GRCh38
rs914284691 Chr.5: 65995013-65995013 on Build GRCh38
rs981749002 Chr.5: 65994880-65994880 on Build GRCh38
rs763700136 Chr.5: 65994759-65994759 on Build GRCh38
rs34832453 Chr.5: 65994819-65994819 on Build GRCh38
rs893615298 Chr.5: 65995082-65995082 on Build GRCh38
rs750979131 Chr.5: 65994792-65994792 on Build GRCh38

View Full Product Details