Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 67182528-67183036 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD180
Entrez Gene ID
4064
Gene Name
CD180 molecule
Gene Aliases
LY64, Ly78, RP105
UniGene
Hs.87205
Gene Chromosome Location
Chr.5: 67182275-67196789 on Build GRCh38
Transcripts
NM_005582.2 XM_005248504.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3828326 c.1856C>G p.S619C Chr.5: 67182987-67182987 on Build GRCh38
171064 c.1915C>T p.L639L Chr.5: 67182928-67182928 on Build GRCh38
1438246 c.1866G>A p.K622K Chr.5: 67182977-67182977 on Build GRCh38
1438245 c.1871C>T p.S624F Chr.5: 67182972-67182972 on Build GRCh38
4895130 c.1834T>A p.S612T Chr.5: 67183009-67183009 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747699790 Chr.5: 67182916-67182916 on Build GRCh38
rs1007894238 Chr.5: 67182551-67182551 on Build GRCh38
rs746362762 Chr.5: 67182985-67182985 on Build GRCh38
rs5744529 Chr.5: 67182564-67182564 on Build GRCh38
rs746684140 Chr.5: 67182975-67182975 on Build GRCh38
rs577689374 Chr.5: 67182550-67182550 on Build GRCh38
rs778906167 Chr.5: 67182709-67182709 on Build GRCh38
rs768796013 Chr.5: 67183001-67183001 on Build GRCh38
rs544924019 Chr.5: 67182983-67182983 on Build GRCh38
rs58890858 Chr.5: 67182738-67182738 on Build GRCh38
rs943191205 Chr.5: 67182789-67182789 on Build GRCh38
rs904464382 Chr.5: 67182799-67182799 on Build GRCh38
rs775679457 Chr.5: 67182988-67182988 on Build GRCh38
rs949324975 Chr.5: 67182885-67182885 on Build GRCh38
rs2230525 Chr.5: 67182798-67182798 on Build GRCh38
rs34803053 Chr.5: 67182724-67182725 on Build GRCh38
rs142133637 Chr.5: 67182632-67182632 on Build GRCh38
rs764261269 Chr.5: 67182844-67182844 on Build GRCh38
rs747294050 Chr.5: 67182812-67182812 on Build GRCh38
rs182074198 Chr.5: 67182703-67182703 on Build GRCh38
rs753770262 Chr.5: 67182899-67182899 on Build GRCh38
rs751830500 Chr.5: 67182866-67182866 on Build GRCh38
rs556865552 Chr.5: 67182755-67182755 on Build GRCh38
rs1024358677 Chr.5: 67182880-67182880 on Build GRCh38
rs575426264 Chr.5: 67182782-67182782 on Build GRCh38
rs1037525068 Chr.5: 67182941-67182941 on Build GRCh38
rs190682990 Chr.5: 67182647-67182647 on Build GRCh38
rs771130683 Chr.5: 67182976-67182976 on Build GRCh38
rs1037970131 Chr.5: 67182557-67182557 on Build GRCh38
rs776941829 Chr.5: 67182820-67182820 on Build GRCh38
rs1031659726 Chr.5: 67182565-67182565 on Build GRCh38
rs1014350532 Chr.5: 67182876-67182876 on Build GRCh38
rs1017269462 Chr.5: 67182708-67182708 on Build GRCh38
rs911724723 Chr.5: 67182779-67182779 on Build GRCh38
rs1017490160 Chr.5: 67182552-67182552 on Build GRCh38
rs1803440 Chr.5: 67182581-67182581 on Build GRCh38
rs760121114 Chr.5: 67182825-67182825 on Build GRCh38
rs890090671 Chr.5: 67182615-67182615 on Build GRCh38
rs749517827 Chr.5: 67182554-67182554 on Build GRCh38
rs1017499350 Chr.5: 67182917-67182917 on Build GRCh38
rs770471496 Chr.5: 67182986-67182986 on Build GRCh38
rs773597415 Chr.5: 67182806-67182806 on Build GRCh38
rs918192141 Chr.5: 67182859-67182859 on Build GRCh38
rs766078074 Chr.5: 67183013-67183013 on Build GRCh38
rs140321429 Chr.5: 67182832-67182832 on Build GRCh38
rs762162192 Chr.5: 67183011-67183011 on Build GRCh38
rs1052068312 Chr.5: 67182549-67182549 on Build GRCh38
rs771003235 Chr.5: 67182667-67182667 on Build GRCh38
rs980769431 Chr.5: 67182663-67182663 on Build GRCh38
rs766201593 Chr.5: 67182875-67182875 on Build GRCh38
rs564562758 Chr.5: 67182609-67182609 on Build GRCh38
rs369776137 Chr.5: 67182920-67182920 on Build GRCh38
rs769105461 Chr.5: 67183010-67183010 on Build GRCh38
rs891798277 Chr.5: 67182848-67182848 on Build GRCh38
rs376832414 Chr.5: 67182922-67182922 on Build GRCh38
rs5744527 Chr.5: 67182831-67182831 on Build GRCh38
rs368264252 Chr.5: 67182608-67182608 on Build GRCh38
rs866909606 Chr.5: 67182895-67182895 on Build GRCh38
rs776431180 Chr.5: 67183014-67183014 on Build GRCh38
rs2230524 Chr.5: 67182901-67182901 on Build GRCh38
rs538796184 Chr.5: 67182678-67182678 on Build GRCh38
rs565546385 Chr.5: 67182870-67182870 on Build GRCh38
rs759441508 Chr.5: 67183015-67183015 on Build GRCh38
rs5744528 Chr.5: 67182816-67182816 on Build GRCh38
rs763243563 Chr.5: 67182841-67182841 on Build GRCh38
rs777240115 Chr.5: 67182932-67182932 on Build GRCh38
rs762894875 Chr.5: 67182989-67182989 on Build GRCh38
rs1024933123 Chr.5: 67182617-67182617 on Build GRCh38
rs1007200083 Chr.5: 67182631-67182631 on Build GRCh38
rs747424517 Chr.5: 67182970-67182970 on Build GRCh38
rs904535841 Chr.5: 67182558-67182558 on Build GRCh38
rs897684612 Chr.5: 67182951-67182951 on Build GRCh38
rs539383079 Chr.5: 67183012-67183013 on Build GRCh38
rs1052094146 Chr.5: 67183008-67183008 on Build GRCh38
rs768039902 Chr.5: 67182881-67182881 on Build GRCh38
rs756092596 Chr.5: 67182954-67182954 on Build GRCh38
rs749470933 Chr.5: 67182664-67182664 on Build GRCh38
rs778376595 Chr.5: 67182904-67182904 on Build GRCh38
rs746429444 Chr.5: 67182773-67182773 on Build GRCh38
rs75456095 Chr.5: 67182961-67182961 on Build GRCh38

View Full Product Details