Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 69195723-69196255 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CENPH
Entrez Gene ID
64946
Gene Name
centromere protein H
Gene Aliases
-
UniGene
Hs.631967
Gene Chromosome Location
Chr.5: 69189548-69210357 on Build GRCh38
Transcripts
NM_022909.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6104363 c.262G>C p.E88Q Chr.5: 69195739-69195739 on Build GRCh38
3072066 c.268G>A p.E90K Chr.5: 69195745-69195745 on Build GRCh38
6104362 c.313A>T p.R105W Chr.5: 69195790-69195790 on Build GRCh38
1438373 c.314G>T p.R105M Chr.5: 69195791-69195791 on Build GRCh38
6900614 c.314G>A p.R105K Chr.5: 69195791-69195791 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs189095269 Chr.5: 69196086-69196086 on Build GRCh38
rs147308361 Chr.5: 69196074-69196075 on Build GRCh38
rs570728502 Chr.5: 69195863-69195863 on Build GRCh38
rs567040501 Chr.5: 69195924-69195924 on Build GRCh38
rs185845023 Chr.5: 69195805-69195805 on Build GRCh38
rs34230657 Chr.5: 69196142-69196143 on Build GRCh38
rs112911250 Chr.5: 69195893-69195894 on Build GRCh38
rs574535134 Chr.5: 69196023-69196023 on Build GRCh38
rs909161002 Chr.5: 69196114-69196115 on Build GRCh38
rs539822205 Chr.5: 69195848-69195848 on Build GRCh38
rs768998151 Chr.5: 69196175-69196175 on Build GRCh38
rs928168619 Chr.5: 69196218-69196218 on Build GRCh38
rs749014993 Chr.5: 69195849-69195849 on Build GRCh38
rs1007669943 Chr.5: 69196171-69196171 on Build GRCh38
rs887605074 Chr.5: 69195803-69195803 on Build GRCh38
rs763683166 Chr.5: 69195820-69195820 on Build GRCh38
rs996388142 Chr.5: 69196111-69196111 on Build GRCh38
rs945637095 Chr.5: 69195983-69195983 on Build GRCh38
rs552522714 Chr.5: 69195860-69195860 on Build GRCh38
rs1031770126 Chr.5: 69196222-69196222 on Build GRCh38
rs201323177 Chr.5: 69195777-69195777 on Build GRCh38
rs1046681136 Chr.5: 69195985-69195985 on Build GRCh38
rs762804222 Chr.5: 69195815-69195815 on Build GRCh38
rs549866264 Chr.5: 69196096-69196096 on Build GRCh38
rs535569782 Chr.5: 69196054-69196054 on Build GRCh38
rs185579653 Chr.5: 69196131-69196131 on Build GRCh38
rs904732127 Chr.5: 69196199-69196199 on Build GRCh38
rs761116827 Chr.5: 69196170-69196170 on Build GRCh38
rs571727374 Chr.5: 69196095-69196095 on Build GRCh38
rs1008801896 Chr.5: 69196092-69196092 on Build GRCh38
rs776014041 Chr.5: 69196138-69196138 on Build GRCh38
rs201108217 Chr.5: 69195808-69195808 on Build GRCh38
rs994476054 Chr.5: 69196031-69196031 on Build GRCh38
rs913893057 Chr.5: 69195947-69195947 on Build GRCh38
rs960079189 Chr.5: 69196075-69196076 on Build GRCh38
rs564449817 Chr.5: 69196116-69196116 on Build GRCh38
rs968903593 Chr.5: 69195996-69195996 on Build GRCh38
rs938144346 Chr.5: 69196001-69196001 on Build GRCh38
rs776874294 Chr.5: 69196192-69196192 on Build GRCh38
rs201808938 Chr.5: 69195887-69195887 on Build GRCh38
rs2126922 Chr.5: 69196077-69196077 on Build GRCh38
rs767679889 Chr.5: 69195771-69195771 on Build GRCh38
rs997276558 Chr.5: 69195873-69195873 on Build GRCh38
rs1051536746 Chr.5: 69196160-69196160 on Build GRCh38
rs1047408557 Chr.5: 69196037-69196037 on Build GRCh38
rs1018817423 Chr.5: 69195837-69195837 on Build GRCh38
rs1008750914 Chr.5: 69195828-69195828 on Build GRCh38
rs181139075 Chr.5: 69196108-69196108 on Build GRCh38
rs913946377 Chr.5: 69196174-69196174 on Build GRCh38
rs191266624 Chr.5: 69196168-69196168 on Build GRCh38
rs748574550 Chr.5: 69196128-69196129 on Build GRCh38
rs566115703 Chr.5: 69195832-69195832 on Build GRCh38
rs768652623 Chr.5: 69195749-69195749 on Build GRCh38
rs1054885646 Chr.5: 69196021-69196021 on Build GRCh38
rs752987588 Chr.5: 69195810-69195811 on Build GRCh38
rs897677247 Chr.5: 69196135-69196135 on Build GRCh38
rs2972375 Chr.5: 69195942-69195942 on Build GRCh38
rs776382802 Chr.5: 69195780-69195780 on Build GRCh38
rs775459868 Chr.5: 69195812-69195812 on Build GRCh38
rs556279973 Chr.5: 69195997-69195997 on Build GRCh38
rs751178961 Chr.5: 69195826-69195826 on Build GRCh38
rs898887421 Chr.5: 69196026-69196026 on Build GRCh38
rs376726420 Chr.5: 69195766-69195766 on Build GRCh38
rs397744259 Chr.5: 69195894-69195895 on Build GRCh38
rs921308345 Chr.5: 69195878-69195878 on Build GRCh38
rs376036036 Chr.5: 69195757-69195757 on Build GRCh38
rs11283275 Chr.5: 69195887-69195888 on Build GRCh38
rs35021233 Chr.5: 69195888-69195889 on Build GRCh38
rs933035017 Chr.5: 69196149-69196149 on Build GRCh38

View Full Product Details